Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100087754-100088251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SASS6
Entrez Gene ID
163786
Gene Name
SAS-6 centriolar assembly protein
Gene Aliases
MCPH14, SAS-6, SAS6
UniGene
Hs.591447
Gene Chromosome Location
Chr.1: 100082632-100133095 on Build GRCh38
Transcripts
NM_001304829.1 NM_194292.2 XM_017000486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4790867 c.1710A>T p.A570A Chr.1: 100088201-100088201 on Build GRCh38
6857467 c.1687T>G p.L563V Chr.1: 100088224-100088224 on Build GRCh38
7334291 c.1734A>T p.G578G Chr.1: 100088177-100088177 on Build GRCh38
5684300 c.1676T>A p.V559D Chr.1: 100088235-100088235 on Build GRCh38
892925 c.1676T>C p.V559A Chr.1: 100088235-100088235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891133514 Chr.1: 100087961-100087961 on Build GRCh38
rs373791132 Chr.1: 100088182-100088182 on Build GRCh38
rs755646247 Chr.1: 100088197-100088197 on Build GRCh38
rs775503778 Chr.1: 100087933-100087933 on Build GRCh38
rs986134518 Chr.1: 100087848-100087848 on Build GRCh38
rs565794639 Chr.1: 100087890-100087890 on Build GRCh38
rs754709915 Chr.1: 100088222-100088222 on Build GRCh38
rs762862602 Chr.1: 100088170-100088170 on Build GRCh38
rs767025470 Chr.1: 100088177-100088177 on Build GRCh38
rs111376606 Chr.1: 100087835-100087836 on Build GRCh38
rs113004403 Chr.1: 100088002-100088002 on Build GRCh38
rs111770751 Chr.1: 100088028-100088028 on Build GRCh38
rs990312812 Chr.1: 100087932-100087932 on Build GRCh38
rs534689555 Chr.1: 100087891-100087891 on Build GRCh38
rs955372094 Chr.1: 100087844-100087844 on Build GRCh38
rs375229473 Chr.1: 100088091-100088091 on Build GRCh38
rs879006118 Chr.1: 100087921-100087921 on Build GRCh38
rs536802866 Chr.1: 100088001-100088001 on Build GRCh38
rs779928526 Chr.1: 100088202-100088202 on Build GRCh38
rs184224576 Chr.1: 100087786-100087786 on Build GRCh38
rs1053409136 Chr.1: 100087978-100087979 on Build GRCh38
rs1021822282 Chr.1: 100088086-100088086 on Build GRCh38
rs764562864 Chr.1: 100088128-100088128 on Build GRCh38
rs369339821 Chr.1: 100088146-100088146 on Build GRCh38
rs895096140 Chr.1: 100088147-100088147 on Build GRCh38
rs941901806 Chr.1: 100087822-100087822 on Build GRCh38
rs945133812 Chr.1: 100087937-100087937 on Build GRCh38
rs927634857 Chr.1: 100088032-100088032 on Build GRCh38
rs556832031 Chr.1: 100088037-100088037 on Build GRCh38
rs974113909 Chr.1: 100087827-100087827 on Build GRCh38
rs548058679 Chr.1: 100087892-100087892 on Build GRCh38
rs763339900 Chr.1: 100088122-100088122 on Build GRCh38
rs567864008 Chr.1: 100087928-100087928 on Build GRCh38
rs776612464 Chr.1: 100088112-100088112 on Build GRCh38
rs760063746 Chr.1: 100088114-100088114 on Build GRCh38
rs576637206 Chr.1: 100088166-100088166 on Build GRCh38
rs994266018 Chr.1: 100088219-100088219 on Build GRCh38
rs944246019 Chr.1: 100087975-100087975 on Build GRCh38
rs753780384 Chr.1: 100088214-100088214 on Build GRCh38
rs770373477 Chr.1: 100088116-100088116 on Build GRCh38
rs749953313 Chr.1: 100088187-100088187 on Build GRCh38
rs778639092 Chr.1: 100088226-100088226 on Build GRCh38
rs957844482 Chr.1: 100087926-100087926 on Build GRCh38
rs776170009 Chr.1: 100088120-100088120 on Build GRCh38
rs563623594 Chr.1: 100087778-100087778 on Build GRCh38
rs771037455 Chr.1: 100088107-100088107 on Build GRCh38
rs977779586 Chr.1: 100088015-100088015 on Build GRCh38

View Full Product Details