Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 29762730-29763265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBMS3
Entrez Gene ID
27303
Gene Name
RNA binding motif single stranded interacting protein 3
Gene Aliases
-
UniGene
Hs.221436
Gene Chromosome Location
Chr.3: 29280860-30010395 on Build GRCh38
Transcripts
NM_001003792.2 NM_001003793.2 NM_001177711.1 NM_001177712.1 NM_014483.3 XM_005265060.2 XM_005265061.2 XM_005265062.1 XM_005265063.1 XM_005265065.4 XM_017006178.1 XM_017006179.1 XM_017006180.1 XM_017006181.1 XM_017006182.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3427502 c.613C>A p.L205M Chr.3: 29762965-29762965 on Build GRCh38
4002640 c.637+7A>T p.? Chr.3: 29762996-29762996 on Build GRCh38
4553321 c.586G>A p.V196I Chr.3: 29762938-29762938 on Build GRCh38
5824974 c.630C>A p.G210G Chr.3: 29762982-29762982 on Build GRCh38
5207219 c.611A>T p.Y204F Chr.3: 29762963-29762963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759571594 Chr.3: 29762978-29762978 on Build GRCh38
rs569969030 Chr.3: 29763226-29763226 on Build GRCh38
rs752186055 Chr.3: 29762886-29762886 on Build GRCh38
rs767117451 Chr.3: 29762885-29762885 on Build GRCh38
rs765691266 Chr.3: 29763003-29763003 on Build GRCh38
rs923375143 Chr.3: 29763237-29763237 on Build GRCh38
rs1049544172 Chr.3: 29762918-29762918 on Build GRCh38
rs760760665 Chr.3: 29762890-29762890 on Build GRCh38
rs768819331 Chr.3: 29762950-29762950 on Build GRCh38
rs899575258 Chr.3: 29762756-29762756 on Build GRCh38
rs1371839 Chr.3: 29762996-29762996 on Build GRCh38
rs890770226 Chr.3: 29763130-29763130 on Build GRCh38
rs146937617 Chr.3: 29762767-29762767 on Build GRCh38
rs758819558 Chr.3: 29762925-29762925 on Build GRCh38
rs765952109 Chr.3: 29763002-29763002 on Build GRCh38
rs150660396 Chr.3: 29763025-29763025 on Build GRCh38
rs763703307 Chr.3: 29762901-29762901 on Build GRCh38
rs898408040 Chr.3: 29763155-29763155 on Build GRCh38
rs956478696 Chr.3: 29763037-29763037 on Build GRCh38
rs766043006 Chr.3: 29762860-29762860 on Build GRCh38
rs765710298 Chr.3: 29763038-29763039 on Build GRCh38
rs1024466515 Chr.3: 29763077-29763077 on Build GRCh38
rs750732160 Chr.3: 29763013-29763013 on Build GRCh38
rs758988262 Chr.3: 29763015-29763015 on Build GRCh38
rs752066051 Chr.3: 29763024-29763024 on Build GRCh38
rs2577124 Chr.3: 29762904-29762904 on Build GRCh38
rs368732266 Chr.3: 29762935-29762935 on Build GRCh38
rs950274042 Chr.3: 29762851-29762851 on Build GRCh38
rs774007838 Chr.3: 29762977-29762977 on Build GRCh38
rs557257146 Chr.3: 29762898-29762898 on Build GRCh38
rs560451954 Chr.3: 29762780-29762780 on Build GRCh38
rs939920520 Chr.3: 29763134-29763134 on Build GRCh38
rs141691907 Chr.3: 29762949-29762949 on Build GRCh38
rs751006803 Chr.3: 29762923-29762923 on Build GRCh38
rs79180200 Chr.3: 29762792-29762792 on Build GRCh38
rs917442729 Chr.3: 29762825-29762825 on Build GRCh38
rs779082369 Chr.3: 29762910-29762910 on Build GRCh38
rs764532920 Chr.3: 29762898-29762898 on Build GRCh38
rs775454194 Chr.3: 29762980-29762980 on Build GRCh38
rs995770567 Chr.3: 29763171-29763171 on Build GRCh38
rs940774135 Chr.3: 29762981-29762981 on Build GRCh38
rs749973445 Chr.3: 29763041-29763041 on Build GRCh38
rs536348489 Chr.3: 29762897-29762898 on Build GRCh38
rs770761462 Chr.3: 29762967-29762967 on Build GRCh38
rs7340719 Chr.3: 29762809-29762809 on Build GRCh38
rs966953666 Chr.3: 29762921-29762921 on Build GRCh38
rs1040883172 Chr.3: 29763232-29763232 on Build GRCh38
rs767033513 Chr.3: 29763044-29763044 on Build GRCh38
rs984769073 Chr.3: 29763167-29763167 on Build GRCh38
rs759322513 Chr.3: 29762879-29762879 on Build GRCh38
rs995598157 Chr.3: 29762865-29762865 on Build GRCh38
rs898292197 Chr.3: 29762864-29762864 on Build GRCh38
rs376251270 Chr.3: 29763099-29763099 on Build GRCh38
rs75670194 Chr.3: 29762763-29762763 on Build GRCh38
rs550824334 Chr.3: 29763068-29763068 on Build GRCh38
rs959466741 Chr.3: 29762896-29762896 on Build GRCh38
rs980286051 Chr.3: 29763093-29763093 on Build GRCh38
rs753447532 Chr.3: 29762902-29762902 on Build GRCh38
rs545463681 Chr.3: 29763198-29763198 on Build GRCh38
rs760207983 Chr.3: 29762998-29762998 on Build GRCh38
rs748257009 Chr.3: 29762964-29762964 on Build GRCh38
rs1371840 Chr.3: 29763018-29763018 on Build GRCh38
rs931786049 Chr.3: 29763240-29763240 on Build GRCh38
rs763544501 Chr.3: 29763001-29763001 on Build GRCh38
rs368908564 Chr.3: 29762995-29762998 on Build GRCh38
rs774356085 Chr.3: 29762875-29762875 on Build GRCh38
rs745616097 Chr.3: 29763089-29763089 on Build GRCh38
rs199988238 Chr.3: 29762985-29762985 on Build GRCh38
rs781589599 Chr.3: 29762962-29762962 on Build GRCh38
rs1015453841 Chr.3: 29762828-29762828 on Build GRCh38
rs549984872 Chr.3: 29763204-29763204 on Build GRCh38
rs1036915031 Chr.3: 29763148-29763148 on Build GRCh38

View Full Product Details