Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 37052572-37053104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRFIP2
Entrez Gene ID
9209
Gene Name
LRR binding FLII interacting protein 2
Gene Aliases
HUFI-2
UniGene
Hs.740522
Gene Chromosome Location
Chr.3: 37052626-37176817 on Build GRCh38
Transcripts
NM_001134369.2 NM_001282691.1 NM_006309.3 NM_017724.2 XM_005265539.2 XM_005265540.1 XM_005265550.1 XM_005265551.1 XM_005265553.1 XM_005265554.2 XM_005265556.1 XM_006713385.1 XM_006713386.1 XM_006713387.1 XM_006713388.1 XM_006713389.1 XM_006713390.1 XM_006713392.1 XM_006713393.1 XM_006713394.1 XM_006713395.1 XM_006713396.1 XM_006713397.1 XM_006713400.1 XM_006713404.1 XM_011534217.1 XM_011534218.1 XM_011534219.1 XM_011534221.1 XM_011534222.1 XM_011534229.1 XM_017007465.1 XM_017007466.1 XM_017007467.1 XM_017007468.1 XM_017007469.1 XM_017007470.1 XM_017007471.1 XM_017007472.1 XM_017007473.1 XM_017007474.1 XM_017007475.1 XM_017007476.1 XM_017007477.1 XM_017007478.1 XM_017007479.1 XM_017007480.1 XM_017007481.1 XM_017007482.1 XM_017007483.1 XM_017007484.1

SNP Detail

SNP ID Chromosome Location
rs945659844 Chr.3: 37052921-37052923 on Build GRCh38
rs1033842377 Chr.3: 37052878-37052879 on Build GRCh38
rs958484501 Chr.3: 37052896-37052897 on Build GRCh38
rs943821963 Chr.3: 37052654-37052654 on Build GRCh38
rs1039576118 Chr.3: 37052668-37052668 on Build GRCh38
rs75908412 Chr.3: 37052836-37052836 on Build GRCh38
rs773429575 Chr.3: 37052976-37052976 on Build GRCh38
rs771548592 Chr.3: 37052600-37052601 on Build GRCh38
rs886716171 Chr.3: 37052880-37052880 on Build GRCh38
rs757862539 Chr.3: 37053027-37053028 on Build GRCh38
rs967295252 Chr.3: 37053015-37053015 on Build GRCh38
rs1028790038 Chr.3: 37053007-37053007 on Build GRCh38
rs995590719 Chr.3: 37052735-37052735 on Build GRCh38
rs1004541000 Chr.3: 37052974-37052974 on Build GRCh38
rs543999097 Chr.3: 37052613-37052613 on Build GRCh38
rs151247849 Chr.3: 37053080-37053080 on Build GRCh38
rs927973408 Chr.3: 37053056-37053056 on Build GRCh38
rs529403313 Chr.3: 37052795-37052795 on Build GRCh38
rs991305445 Chr.3: 37053077-37053077 on Build GRCh38
rs111226642 Chr.3: 37052616-37052616 on Build GRCh38
rs3774329 Chr.3: 37053031-37053031 on Build GRCh38
rs978520289 Chr.3: 37052623-37052623 on Build GRCh38
rs930833976 Chr.3: 37052846-37052846 on Build GRCh38
rs1049340143 Chr.3: 37052847-37052847 on Build GRCh38
rs541037863 Chr.3: 37052954-37052954 on Build GRCh38
rs928057269 Chr.3: 37052678-37052678 on Build GRCh38
rs912460031 Chr.3: 37053022-37053022 on Build GRCh38
rs190129295 Chr.3: 37053010-37053010 on Build GRCh38
rs569658747 Chr.3: 37053072-37053072 on Build GRCh38
rs989911017 Chr.3: 37052908-37052908 on Build GRCh38
rs1027523448 Chr.3: 37052768-37052768 on Build GRCh38
rs989979890 Chr.3: 37053012-37053012 on Build GRCh38
rs760815711 Chr.3: 37053004-37053004 on Build GRCh38
rs1055260847 Chr.3: 37052748-37052748 on Build GRCh38
rs772069172 Chr.3: 37052916-37052916 on Build GRCh38
rs201835937 Chr.3: 37052790-37052790 on Build GRCh38
rs913183132 Chr.3: 37052615-37052615 on Build GRCh38
rs945732124 Chr.3: 37052605-37052605 on Build GRCh38
rs148720085 Chr.3: 37053014-37053014 on Build GRCh38
rs202002930 Chr.3: 37052689-37052691 on Build GRCh38
rs939445231 Chr.3: 37052688-37052688 on Build GRCh38
rs1019042572 Chr.3: 37052975-37052975 on Build GRCh38
rs895324139 Chr.3: 37052822-37052822 on Build GRCh38
rs1025153390 Chr.3: 37052820-37052820 on Build GRCh38
rs746577374 Chr.3: 37052742-37052742 on Build GRCh38

View Full Product Details