Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39074648-39075156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR48
Entrez Gene ID
57599
Gene Name
WD repeat domain 48
Gene Aliases
P80, SPG60, UAF1
UniGene
Hs.109778
Gene Chromosome Location
Chr.3: 39051986-39096671 on Build GRCh38
Transcripts
NM_001303402.1 NM_001303403.1 NM_020839.3 XM_005265346.3 XM_011533971.1 XM_017006960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6421907 c.676C>A p.L226M Chr.3: 39074729-39074729 on Build GRCh38
5950309 c.770C>T p.A257V Chr.3: 39074823-39074823 on Build GRCh38
1044397 c.871T>G p.C291G Chr.3: 39074924-39074924 on Build GRCh38
6019994 c.817G>C p.D273H Chr.3: 39074870-39074870 on Build GRCh38
3593763 c.859C>T p.R287W Chr.3: 39074912-39074912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016504510 Chr.3: 39075072-39075072 on Build GRCh38
rs758573682 Chr.3: 39074789-39074789 on Build GRCh38
rs754409192 Chr.3: 39074984-39074984 on Build GRCh38
rs1013702056 Chr.3: 39074722-39074722 on Build GRCh38
rs765621635 Chr.3: 39074784-39074784 on Build GRCh38
rs745561822 Chr.3: 39074854-39074854 on Build GRCh38
rs986639974 Chr.3: 39074923-39074923 on Build GRCh38
rs781644309 Chr.3: 39074808-39074808 on Build GRCh38
rs778402538 Chr.3: 39074973-39074973 on Build GRCh38
rs913706214 Chr.3: 39074936-39074936 on Build GRCh38
rs771711364 Chr.3: 39074856-39074856 on Build GRCh38
rs762923499 Chr.3: 39074914-39074914 on Build GRCh38
rs960892403 Chr.3: 39074702-39074702 on Build GRCh38
rs750508981 Chr.3: 39074781-39074781 on Build GRCh38
rs9865014 Chr.3: 39075130-39075130 on Build GRCh38
rs778397814 Chr.3: 39074850-39074850 on Build GRCh38
rs760625198 Chr.3: 39074749-39074749 on Build GRCh38
rs765239917 Chr.3: 39074767-39074767 on Build GRCh38
rs760537101 Chr.3: 39074877-39074877 on Build GRCh38
rs1004670547 Chr.3: 39075068-39075068 on Build GRCh38
rs951045099 Chr.3: 39074786-39074786 on Build GRCh38
rs1026403642 Chr.3: 39074776-39074776 on Build GRCh38
rs759228561 Chr.3: 39074697-39074697 on Build GRCh38
rs551888994 Chr.3: 39075041-39075041 on Build GRCh38
rs764754294 Chr.3: 39074700-39074701 on Build GRCh38
rs756704105 Chr.3: 39074823-39074823 on Build GRCh38
rs775592713 Chr.3: 39074671-39074671 on Build GRCh38
rs766524631 Chr.3: 39074931-39074931 on Build GRCh38
rs767381237 Chr.3: 39074710-39074710 on Build GRCh38
rs532191635 Chr.3: 39075023-39075023 on Build GRCh38
rs764056166 Chr.3: 39075069-39075070 on Build GRCh38
rs1006909643 Chr.3: 39075064-39075064 on Build GRCh38
rs377765260 Chr.3: 39074953-39074953 on Build GRCh38
rs374426411 Chr.3: 39074887-39074887 on Build GRCh38
rs748619060 Chr.3: 39074815-39074815 on Build GRCh38
rs368832991 Chr.3: 39074806-39074806 on Build GRCh38
rs774111205 Chr.3: 39074686-39074686 on Build GRCh38
rs903880104 Chr.3: 39074941-39074941 on Build GRCh38
rs1002721171 Chr.3: 39074680-39074680 on Build GRCh38
rs559831281 Chr.3: 39074869-39074869 on Build GRCh38
rs1015769478 Chr.3: 39075077-39075077 on Build GRCh38
rs938004238 Chr.3: 39074996-39074996 on Build GRCh38
rs78462179 Chr.3: 39075044-39075044 on Build GRCh38
rs962726306 Chr.3: 39075084-39075084 on Build GRCh38
rs762664164 Chr.3: 39074938-39074938 on Build GRCh38
rs753053409 Chr.3: 39074835-39074835 on Build GRCh38
rs756602172 Chr.3: 39074964-39074964 on Build GRCh38
rs944601541 Chr.3: 39074995-39074995 on Build GRCh38
rs767936897 Chr.3: 39074947-39074947 on Build GRCh38
rs748792693 Chr.3: 39074832-39074832 on Build GRCh38
rs149913299 Chr.3: 39074824-39074824 on Build GRCh38
rs756678107 Chr.3: 39074988-39074988 on Build GRCh38
rs921007129 Chr.3: 39074848-39074848 on Build GRCh38
rs753118357 Chr.3: 39074954-39074954 on Build GRCh38
rs751691894 Chr.3: 39074937-39074937 on Build GRCh38
rs1043630964 Chr.3: 39074939-39074939 on Build GRCh38
rs116274141 Chr.3: 39075052-39075052 on Build GRCh38
rs568337906 Chr.3: 39075053-39075053 on Build GRCh38
rs548508187 Chr.3: 39075079-39075080 on Build GRCh38
rs376868647 Chr.3: 39074728-39074728 on Build GRCh38
rs181618364 Chr.3: 39075047-39075047 on Build GRCh38
rs933852453 Chr.3: 39074860-39074860 on Build GRCh38
rs776611526 Chr.3: 39074911-39074911 on Build GRCh38
rs144887987 Chr.3: 39074807-39074807 on Build GRCh38
rs770482987 Chr.3: 39074678-39074678 on Build GRCh38
rs766501475 Chr.3: 39074797-39074797 on Build GRCh38
rs961923186 Chr.3: 39074689-39074689 on Build GRCh38

View Full Product Details