Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49017506-49018003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DALRD3
Entrez Gene ID
55152
Gene Name
DALR anticodon binding domain containing 3
Gene Aliases
-
UniGene
Hs.654815
Gene Chromosome Location
Chr.3: 49015399-49021386 on Build GRCh38
Transcripts
NM_001009996.2 NM_001276405.1 NM_018114.5 XM_005265269.3 XM_006713219.2 XM_011533891.1 XM_011533892.1 XM_011533893.1 XM_017006723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
446685 c.36delG p.W12fs*13 Chr.3: 49017794-49017794 on Build GRCh38
4788804 c.485C>T p.A162V Chr.3: 49017846-49017846 on Build GRCh38
1745377 c.725_744del20 p.E242fs*2 Chr.3: 49017488-49017507 on Build GRCh38
2849263 c.657G>T p.K219N Chr.3: 49017674-49017674 on Build GRCh38
1753261 c.117G>A p.R39R Chr.3: 49017713-49017713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764337023 Chr.3: 49017863-49017863 on Build GRCh38
rs748828314 Chr.3: 49017656-49017656 on Build GRCh38
rs754850067 Chr.3: 49017916-49017916 on Build GRCh38
rs777608812 Chr.3: 49017585-49017585 on Build GRCh38
rs772043252 Chr.3: 49017668-49017668 on Build GRCh38
rs558459260 Chr.3: 49017555-49017555 on Build GRCh38
rs760668877 Chr.3: 49017576-49017576 on Build GRCh38
rs746058911 Chr.3: 49017665-49017665 on Build GRCh38
rs748939169 Chr.3: 49017844-49017844 on Build GRCh38
rs758689843 Chr.3: 49017814-49017814 on Build GRCh38
rs766668118 Chr.3: 49017889-49017889 on Build GRCh38
rs907268407 Chr.3: 49017590-49017590 on Build GRCh38
rs139210102 Chr.3: 49017750-49017750 on Build GRCh38
rs777247162 Chr.3: 49017873-49017873 on Build GRCh38
rs576118617 Chr.3: 49017812-49017812 on Build GRCh38
rs375570832 Chr.3: 49017785-49017785 on Build GRCh38
rs777724131 Chr.3: 49017640-49017640 on Build GRCh38
rs919477793 Chr.3: 49017957-49017957 on Build GRCh38
rs76091680 Chr.3: 49017717-49017717 on Build GRCh38
rs577338193 Chr.3: 49017947-49017947 on Build GRCh38
rs758716427 Chr.3: 49017886-49017886 on Build GRCh38
rs368553099 Chr.3: 49017975-49017975 on Build GRCh38
rs572118307 Chr.3: 49017705-49017705 on Build GRCh38
rs1020529069 Chr.3: 49017735-49017735 on Build GRCh38
rs1013273067 Chr.3: 49017880-49017880 on Build GRCh38
rs780008631 Chr.3: 49017821-49017821 on Build GRCh38
rs543609217 Chr.3: 49017841-49017841 on Build GRCh38
rs760990865 Chr.3: 49017858-49017858 on Build GRCh38
rs370888177 Chr.3: 49017654-49017654 on Build GRCh38
rs753901314 Chr.3: 49017645-49017645 on Build GRCh38
rs957193722 Chr.3: 49017820-49017820 on Build GRCh38
rs767842632 Chr.3: 49017622-49017622 on Build GRCh38
rs764893102 Chr.3: 49017599-49017599 on Build GRCh38
rs570753761 Chr.3: 49017569-49017569 on Build GRCh38
rs757399378 Chr.3: 49017802-49017802 on Build GRCh38
rs761345101 Chr.3: 49017723-49017723 on Build GRCh38
rs1038717265 Chr.3: 49017933-49017933 on Build GRCh38
rs757445839 Chr.3: 49017650-49017650 on Build GRCh38
rs1023998046 Chr.3: 49017727-49017727 on Build GRCh38
rs752989112 Chr.3: 49017758-49017758 on Build GRCh38
rs765426258 Chr.3: 49017883-49017883 on Build GRCh38
rs775610201 Chr.3: 49017575-49017575 on Build GRCh38
rs745684461 Chr.3: 49017676-49017677 on Build GRCh38
rs371270653 Chr.3: 49017884-49017884 on Build GRCh38
rs762578499 Chr.3: 49017743-49017743 on Build GRCh38
rs757284020 Chr.3: 49017567-49017567 on Build GRCh38
rs755934243 Chr.3: 49017980-49017980 on Build GRCh38
rs747010405 Chr.3: 49017676-49017676 on Build GRCh38
rs781227318 Chr.3: 49017838-49017838 on Build GRCh38
rs377632520 Chr.3: 49017550-49017550 on Build GRCh38
rs778837269 Chr.3: 49017810-49017810 on Build GRCh38
rs548329545 Chr.3: 49017832-49017832 on Build GRCh38
rs753899554 Chr.3: 49017787-49017787 on Build GRCh38
rs756231655 Chr.3: 49017545-49017545 on Build GRCh38
rs867448120 Chr.3: 49017771-49017771 on Build GRCh38
rs776140435 Chr.3: 49017583-49017583 on Build GRCh38
rs748420071 Chr.3: 49017882-49017887 on Build GRCh38
rs761188975 Chr.3: 49017768-49017768 on Build GRCh38
rs377402749 Chr.3: 49017579-49017579 on Build GRCh38
rs373966327 Chr.3: 49017847-49017847 on Build GRCh38
rs897066104 Chr.3: 49017946-49017946 on Build GRCh38
rs907304480 Chr.3: 49017979-49017979 on Build GRCh38
rs779982352 Chr.3: 49017669-49017669 on Build GRCh38
rs748620203 Chr.3: 49017541-49017541 on Build GRCh38
rs201113233 Chr.3: 49017553-49017553 on Build GRCh38
rs780833701 Chr.3: 49017921-49017921 on Build GRCh38
rs781465443 Chr.3: 49017877-49017878 on Build GRCh38
rs534636120 Chr.3: 49017570-49017570 on Build GRCh38
rs751898785 Chr.3: 49017600-49017600 on Build GRCh38
rs990845856 Chr.3: 49017685-49017685 on Build GRCh38
rs929992145 Chr.3: 49017965-49017965 on Build GRCh38
rs143496675 Chr.3: 49017840-49017840 on Build GRCh38
rs758198200 Chr.3: 49017805-49017810 on Build GRCh38
rs375464441 Chr.3: 49017906-49017906 on Build GRCh38
rs753257206 Chr.3: 49017630-49017630 on Build GRCh38
rs779157732 Chr.3: 49017651-49017651 on Build GRCh38
rs61743083 Chr.3: 49017639-49017639 on Build GRCh38
rs759932101 Chr.3: 49017609-49017609 on Build GRCh38
rs748752784 Chr.3: 49017981-49017981 on Build GRCh38
rs772964964 Chr.3: 49017739-49017739 on Build GRCh38
rs778514277 Chr.3: 49017656-49017657 on Build GRCh38
rs769154034 Chr.3: 49017729-49017729 on Build GRCh38
rs953463047 Chr.3: 49017862-49017862 on Build GRCh38
rs367782586 Chr.3: 49017849-49017849 on Build GRCh38
rs746750916 Chr.3: 49017826-49017826 on Build GRCh38

View Full Product Details