Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49098262-49098754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QARS
Entrez Gene ID
5859
Gene Name
glutaminyl-tRNA synthetase
Gene Aliases
GLNRS, MSCCA, PRO2195
UniGene
Hs.79322
Gene Chromosome Location
Chr.3: 49095932-49105129 on Build GRCh38
Transcripts
NM_001272073.1 NM_005051.2 NR_073590.1 XM_017006965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5864775 c.2050C>T p.P684S Chr.3: 49098387-49098387 on Build GRCh38
3595182 c.1904C>T p.P635L Chr.3: 49098652-49098652 on Build GRCh38
446689 c.1984G>C p.E662Q Chr.3: 49098453-49098453 on Build GRCh38
7087219 c.2060G>A p.C687Y Chr.3: 49098377-49098377 on Build GRCh38
6588789 c.1929delC p.Y643fs*1 Chr.3: 49098627-49098627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367835872 Chr.3: 49098490-49098490 on Build GRCh38
rs756380392 Chr.3: 49098419-49098419 on Build GRCh38
rs756075510 Chr.3: 49098516-49098516 on Build GRCh38
rs1017117201 Chr.3: 49098297-49098297 on Build GRCh38
rs79363335 Chr.3: 49098352-49098352 on Build GRCh38
rs775375268 Chr.3: 49098626-49098626 on Build GRCh38
rs761792999 Chr.3: 49098376-49098376 on Build GRCh38
rs374530633 Chr.3: 49098707-49098707 on Build GRCh38
rs376482152 Chr.3: 49098354-49098354 on Build GRCh38
rs979330776 Chr.3: 49098671-49098671 on Build GRCh38
rs767187216 Chr.3: 49098622-49098622 on Build GRCh38
rs753102798 Chr.3: 49098500-49098502 on Build GRCh38
rs777447293 Chr.3: 49098520-49098520 on Build GRCh38
rs145343705 Chr.3: 49098612-49098612 on Build GRCh38
rs964597269 Chr.3: 49098718-49098718 on Build GRCh38
rs574420266 Chr.3: 49098502-49098502 on Build GRCh38
rs201781920 Chr.3: 49098368-49098368 on Build GRCh38
rs976613108 Chr.3: 49098526-49098526 on Build GRCh38
rs753338846 Chr.3: 49098527-49098527 on Build GRCh38
rs747269470 Chr.3: 49098485-49098485 on Build GRCh38
rs1009354427 Chr.3: 49098497-49098497 on Build GRCh38
rs563148564 Chr.3: 49098534-49098534 on Build GRCh38
rs767548783 Chr.3: 49098513-49098513 on Build GRCh38
rs557972227 Chr.3: 49098362-49098362 on Build GRCh38
rs764306525 Chr.3: 49098688-49098688 on Build GRCh38
rs780778915 Chr.3: 49098332-49098332 on Build GRCh38
rs568924851 Chr.3: 49098323-49098323 on Build GRCh38
rs143557138 Chr.3: 49098300-49098300 on Build GRCh38
rs772994973 Chr.3: 49098356-49098356 on Build GRCh38
rs750619032 Chr.3: 49098391-49098391 on Build GRCh38
rs376994371 Chr.3: 49098289-49098289 on Build GRCh38
rs774424848 Chr.3: 49098616-49098616 on Build GRCh38
rs760709234 Chr.3: 49098293-49098293 on Build GRCh38
rs144316335 Chr.3: 49098373-49098373 on Build GRCh38
rs759914821 Chr.3: 49098283-49098283 on Build GRCh38
rs756620094 Chr.3: 49098699-49098699 on Build GRCh38
rs567271491 Chr.3: 49098286-49098286 on Build GRCh38
rs747555785 Chr.3: 49098350-49098350 on Build GRCh38
rs766197862 Chr.3: 49098417-49098417 on Build GRCh38
rs771253717 Chr.3: 49098615-49098615 on Build GRCh38
rs770657741 Chr.3: 49098479-49098479 on Build GRCh38
rs1013287948 Chr.3: 49098480-49098480 on Build GRCh38
rs761265228 Chr.3: 49098684-49098684 on Build GRCh38
rs764987448 Chr.3: 49098672-49098672 on Build GRCh38
rs1034813926 Chr.3: 49098545-49098545 on Build GRCh38
rs905390740 Chr.3: 49098510-49098510 on Build GRCh38
rs537960607 Chr.3: 49098684-49098684 on Build GRCh38
rs767673089 Chr.3: 49098326-49098326 on Build GRCh38
rs114149286 Chr.3: 49098331-49098331 on Build GRCh38
rs760702138 Chr.3: 49098627-49098627 on Build GRCh38
rs940163832 Chr.3: 49098433-49098433 on Build GRCh38
rs764054134 Chr.3: 49098633-49098633 on Build GRCh38
rs988027951 Chr.3: 49098307-49098307 on Build GRCh38
rs768876281 Chr.3: 49098563-49098563 on Build GRCh38
rs752320395 Chr.3: 49098450-49098450 on Build GRCh38
rs746293241 Chr.3: 49098357-49098357 on Build GRCh38
rs140225766 Chr.3: 49098446-49098446 on Build GRCh38
rs374312387 Chr.3: 49098435-49098435 on Build GRCh38
rs1037325022 Chr.3: 49098437-49098437 on Build GRCh38
rs865998076 Chr.3: 49098473-49098473 on Build GRCh38
rs532793779 Chr.3: 49098575-49098575 on Build GRCh38
rs779126108 Chr.3: 49098696-49098696 on Build GRCh38
rs765374731 Chr.3: 49098312-49098312 on Build GRCh38
rs750008591 Chr.3: 49098676-49098676 on Build GRCh38
rs752555108 Chr.3: 49098515-49098515 on Build GRCh38
rs139730889 Chr.3: 49098670-49098670 on Build GRCh38
rs879055592 Chr.3: 49098483-49098483 on Build GRCh38
rs980562659 Chr.3: 49098326-49098327 on Build GRCh38
rs13064784 Chr.3: 49098528-49098528 on Build GRCh38
rs753463624 Chr.3: 49098660-49098660 on Build GRCh38
rs1009235138 Chr.3: 49098611-49098611 on Build GRCh38
rs201648871 Chr.3: 49098313-49098313 on Build GRCh38
rs370934093 Chr.3: 49098369-49098369 on Build GRCh38
rs371725638 Chr.3: 49098491-49098491 on Build GRCh38
rs751375938 Chr.3: 49098494-49098494 on Build GRCh38
rs781741003 Chr.3: 49098572-49098572 on Build GRCh38
rs778251931 Chr.3: 49098706-49098706 on Build GRCh38
rs781435764 Chr.3: 49098681-49098681 on Build GRCh38
rs758172803 Chr.3: 49098314-49098314 on Build GRCh38
rs370587517 Chr.3: 49098438-49098438 on Build GRCh38
rs544950843 Chr.3: 49098686-49098686 on Build GRCh38
rs372388585 Chr.3: 49098732-49098732 on Build GRCh38
rs770241224 Chr.3: 49098583-49098583 on Build GRCh38
rs778797651 Chr.3: 49098366-49098366 on Build GRCh38
rs777618796 Chr.3: 49098464-49098464 on Build GRCh38
rs780478501 Chr.3: 49098484-49098484 on Build GRCh38
rs142441496 Chr.3: 49098404-49098404 on Build GRCh38
rs762780883 Chr.3: 49098409-49098409 on Build GRCh38
rs373351841 Chr.3: 49098310-49098310 on Build GRCh38
rs954037456 Chr.3: 49098533-49098533 on Build GRCh38
rs748978904 Chr.3: 49098465-49098465 on Build GRCh38
rs878982739 Chr.3: 49098486-49098486 on Build GRCh38
rs367776807 Chr.3: 49098489-49098489 on Build GRCh38
rs773296739 Chr.3: 49098605-49098605 on Build GRCh38
rs921379455 Chr.3: 49098363-49098363 on Build GRCh38
rs754208205 Chr.3: 49098719-49098720 on Build GRCh38
rs1032058964 Chr.3: 49098493-49098493 on Build GRCh38
rs755672487 Chr.3: 49098458-49098458 on Build GRCh38
rs765497781 Chr.3: 49098379-49098379 on Build GRCh38
rs370397423 Chr.3: 49098620-49098620 on Build GRCh38
rs747462635 Chr.3: 49098553-49098553 on Build GRCh38
rs746939236 Chr.3: 49098328-49098328 on Build GRCh38
rs138588649 Chr.3: 49098364-49098364 on Build GRCh38
rs112579679 Chr.3: 49098499-49098499 on Build GRCh38
rs758025209 Chr.3: 49098680-49098680 on Build GRCh38

View Full Product Details