Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52248546-52249047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPM1M
Entrez Gene ID
132160
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1M
Gene Aliases
PP2C-eta, PP2CE, PP2Ceta
UniGene
Hs.373560
Gene Chromosome Location
Chr.3: 52245725-52250599 on Build GRCh38
Transcripts
NM_001122870.2 NM_144641.3 XM_005264879.2 XM_005264880.3 XM_005264881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6887179 c.500G>A p.R167Q Chr.3: 52248960-52248960 on Build GRCh38
4325533 c.1032C>T p.L344L Chr.3: 52249009-52249009 on Build GRCh38
3993200 c.309C>T p.L103L Chr.3: 52248667-52248667 on Build GRCh38
4325530 c.938C>A p.S313* Chr.3: 52248660-52248660 on Build GRCh38
6165002 c.576C>T p.L192L Chr.3: 52249036-52249036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566965007 Chr.3: 52248647-52248647 on Build GRCh38
rs757739981 Chr.3: 52248665-52248665 on Build GRCh38
rs759473298 Chr.3: 52248884-52248884 on Build GRCh38
rs143262820 Chr.3: 52248979-52248980 on Build GRCh38
rs762914464 Chr.3: 52248932-52248932 on Build GRCh38
rs368889437 Chr.3: 52248643-52248643 on Build GRCh38
rs957172062 Chr.3: 52248772-52248772 on Build GRCh38
rs375538193 Chr.3: 52248631-52248631 on Build GRCh38
rs371849651 Chr.3: 52248668-52248668 on Build GRCh38
rs746439943 Chr.3: 52248992-52248992 on Build GRCh38
rs765291075 Chr.3: 52248600-52248600 on Build GRCh38
rs781445847 Chr.3: 52248633-52248633 on Build GRCh38
rs530607812 Chr.3: 52248578-52248578 on Build GRCh38
rs376715042 Chr.3: 52248923-52248923 on Build GRCh38
rs774352909 Chr.3: 52248945-52248945 on Build GRCh38
rs1025064092 Chr.3: 52248810-52248810 on Build GRCh38
rs1055181531 Chr.3: 52249017-52249017 on Build GRCh38
rs372600142 Chr.3: 52248685-52248685 on Build GRCh38
rs766892014 Chr.3: 52248971-52248971 on Build GRCh38
rs779491855 Chr.3: 52248986-52248986 on Build GRCh38
rs973636679 Chr.3: 52248948-52248948 on Build GRCh38
rs562217803 Chr.3: 52248960-52248960 on Build GRCh38
rs144497405 Chr.3: 52248661-52248661 on Build GRCh38
rs895108123 Chr.3: 52248757-52248757 on Build GRCh38
rs745624409 Chr.3: 52248924-52248924 on Build GRCh38
rs541562415 Chr.3: 52249010-52249010 on Build GRCh38
rs746663761 Chr.3: 52248749-52248749 on Build GRCh38
rs749653157 Chr.3: 52248662-52248678 on Build GRCh38
rs572900471 Chr.3: 52248998-52248998 on Build GRCh38
rs895162109 Chr.3: 52249021-52249021 on Build GRCh38
rs780718617 Chr.3: 52248640-52248640 on Build GRCh38
rs758479893 Chr.3: 52248611-52248611 on Build GRCh38
rs190246060 Chr.3: 52248981-52248981 on Build GRCh38
rs147068914 Chr.3: 52249008-52249008 on Build GRCh38
rs558517985 Chr.3: 52248940-52248940 on Build GRCh38
rs200280814 Chr.3: 52248720-52248720 on Build GRCh38
rs756549079 Chr.3: 52248742-52248742 on Build GRCh38
rs554185326 Chr.3: 52248648-52248648 on Build GRCh38
rs765094830 Chr.3: 52248687-52248687 on Build GRCh38
rs929493765 Chr.3: 52248988-52248988 on Build GRCh38
rs765988455 Chr.3: 52249006-52249006 on Build GRCh38
rs866501269 Chr.3: 52248795-52248795 on Build GRCh38
rs764790440 Chr.3: 52248745-52248745 on Build GRCh38
rs750757250 Chr.3: 52248728-52248728 on Build GRCh38
rs764471609 Chr.3: 52248591-52248591 on Build GRCh38
rs1039835582 Chr.3: 52248990-52248990 on Build GRCh38
rs760472064 Chr.3: 52248573-52248573 on Build GRCh38
rs999349190 Chr.3: 52248859-52248859 on Build GRCh38
rs778700956 Chr.3: 52248667-52248668 on Build GRCh38
rs762967758 Chr.3: 52248701-52248701 on Build GRCh38
rs970334497 Chr.3: 52248828-52248829 on Build GRCh38
rs760285460 Chr.3: 52248657-52248657 on Build GRCh38
rs1051652554 Chr.3: 52248967-52248967 on Build GRCh38
rs1029858184 Chr.3: 52248866-52248866 on Build GRCh38
rs538523142 Chr.3: 52248874-52248874 on Build GRCh38
rs1022185345 Chr.3: 52248843-52248843 on Build GRCh38
rs775381624 Chr.3: 52248929-52248929 on Build GRCh38
rs751749084 Chr.3: 52248627-52248627 on Build GRCh38
rs753777821 Chr.3: 52248583-52248583 on Build GRCh38
rs770755292 Chr.3: 52248659-52248659 on Build GRCh38
rs770435952 Chr.3: 52248920-52248920 on Build GRCh38
rs761966252 Chr.3: 52248951-52248951 on Build GRCh38
rs202067249 Chr.3: 52249009-52249009 on Build GRCh38
rs754393792 Chr.3: 52248980-52248980 on Build GRCh38
rs891143805 Chr.3: 52248968-52248968 on Build GRCh38
rs202243150 Chr.3: 52248613-52248613 on Build GRCh38
rs902364936 Chr.3: 52248856-52248856 on Build GRCh38
rs148950187 Chr.3: 52248985-52248985 on Build GRCh38
rs757950732 Chr.3: 52248746-52248746 on Build GRCh38
rs772773123 Chr.3: 52248572-52248572 on Build GRCh38
rs776351153 Chr.3: 52248660-52248660 on Build GRCh38
rs750148308 Chr.3: 52248921-52248921 on Build GRCh38
rs745565132 Chr.3: 52249007-52249007 on Build GRCh38
rs926356237 Chr.3: 52248906-52248906 on Build GRCh38
rs372310852 Chr.3: 52248607-52248607 on Build GRCh38
rs535627330 Chr.3: 52248652-52248652 on Build GRCh38
rs752660059 Chr.3: 52248697-52248697 on Build GRCh38
rs757486735 Chr.3: 52248723-52248723 on Build GRCh38
rs983630279 Chr.3: 52249003-52249003 on Build GRCh38
rs1037645894 Chr.3: 52248618-52248618 on Build GRCh38
rs138288630 Chr.3: 52248962-52248962 on Build GRCh38
rs764124601 Chr.3: 52248712-52248712 on Build GRCh38
rs201528882 Chr.3: 52248976-52248976 on Build GRCh38
rs746381519 Chr.3: 52248638-52248638 on Build GRCh38
rs980503462 Chr.3: 52248841-52248841 on Build GRCh38
rs1010837486 Chr.3: 52248781-52248781 on Build GRCh38
rs769597568 Chr.3: 52248925-52248925 on Build GRCh38
rs570488765 Chr.3: 52248570-52248570 on Build GRCh38
rs569513929 Chr.3: 52248814-52248814 on Build GRCh38
rs777215638 Chr.3: 52248748-52248748 on Build GRCh38
rs555993841 Chr.3: 52248743-52248743 on Build GRCh38

View Full Product Details