Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53184796-53185291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCD
Entrez Gene ID
5580
Gene Name
protein kinase C delta
Gene Aliases
ALPS3, CVID9, MAY1, PKCD, nPKC-delta
UniGene
Hs.155342
Gene Chromosome Location
Chr.3: 53161207-53192717 on Build GRCh38
Transcripts
NM_001316327.1 NM_006254.3 NM_212539.1 XM_006713257.3 XM_006713259.2 XM_017006855.1 XM_017006856.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889895 c.888G>T p.Q296H Chr.3: 53184974-53184974 on Build GRCh38
163733 c.865G>T p.E289* Chr.3: 53184951-53184951 on Build GRCh38
2780537 c.817C>T p.R273W Chr.3: 53184903-53184903 on Build GRCh38
7202966 c.852G>A p.Q284Q Chr.3: 53184938-53184938 on Build GRCh38
3729243 c.825G>T p.K275N Chr.3: 53184911-53184911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567614454 Chr.3: 53185199-53185199 on Build GRCh38
rs377760765 Chr.3: 53184825-53184825 on Build GRCh38
rs6798141 Chr.3: 53185262-53185262 on Build GRCh38
rs891947575 Chr.3: 53185060-53185060 on Build GRCh38
rs370600760 Chr.3: 53184980-53184980 on Build GRCh38
rs370808161 Chr.3: 53184956-53184956 on Build GRCh38
rs74437127 Chr.3: 53184821-53184821 on Build GRCh38
rs372488751 Chr.3: 53185128-53185128 on Build GRCh38
rs56406497 Chr.3: 53185133-53185133 on Build GRCh38
rs782180691 Chr.3: 53184959-53184959 on Build GRCh38
rs1011721379 Chr.3: 53185098-53185098 on Build GRCh38
rs551307318 Chr.3: 53185001-53185001 on Build GRCh38
rs782111405 Chr.3: 53184849-53184849 on Build GRCh38
rs146516302 Chr.3: 53185245-53185245 on Build GRCh38
rs372028715 Chr.3: 53184932-53184932 on Build GRCh38
rs782191905 Chr.3: 53184856-53184856 on Build GRCh38
rs41275527 Chr.3: 53185248-53185248 on Build GRCh38
rs782595191 Chr.3: 53184968-53184968 on Build GRCh38
rs111638508 Chr.3: 53185259-53185259 on Build GRCh38
rs782335632 Chr.3: 53184827-53184827 on Build GRCh38
rs145324216 Chr.3: 53184926-53184926 on Build GRCh38
rs377512214 Chr.3: 53184904-53184904 on Build GRCh38
rs1052890871 Chr.3: 53185057-53185057 on Build GRCh38
rs782012522 Chr.3: 53184861-53184861 on Build GRCh38
rs781942162 Chr.3: 53184993-53184993 on Build GRCh38
rs533821731 Chr.3: 53185092-53185092 on Build GRCh38
rs781957524 Chr.3: 53184845-53184845 on Build GRCh38
rs762017004 Chr.3: 53184862-53184862 on Build GRCh38
rs781877260 Chr.3: 53184930-53184930 on Build GRCh38
rs536443555 Chr.3: 53185213-53185213 on Build GRCh38
rs111525461 Chr.3: 53185035-53185035 on Build GRCh38
rs782535622 Chr.3: 53184950-53184950 on Build GRCh38
rs1030284837 Chr.3: 53185253-53185253 on Build GRCh38
rs562250420 Chr.3: 53185007-53185007 on Build GRCh38
rs781989661 Chr.3: 53185014-53185014 on Build GRCh38
rs373245386 Chr.3: 53184879-53184879 on Build GRCh38
rs782446907 Chr.3: 53184966-53184966 on Build GRCh38
rs782140747 Chr.3: 53185015-53185015 on Build GRCh38
rs782084034 Chr.3: 53184996-53184996 on Build GRCh38
rs112763010 Chr.3: 53184859-53184859 on Build GRCh38
rs782157692 Chr.3: 53184871-53184871 on Build GRCh38
rs80298191 Chr.3: 53184856-53184856 on Build GRCh38
rs557088485 Chr.3: 53185249-53185249 on Build GRCh38
rs782223934 Chr.3: 53184970-53184970 on Build GRCh38
rs782414501 Chr.3: 53184985-53184985 on Build GRCh38
rs782373708 Chr.3: 53184971-53184971 on Build GRCh38
rs370951928 Chr.3: 53185022-53185022 on Build GRCh38

View Full Product Details