Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53224964-53225471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TKT
Entrez Gene ID
7086
Gene Name
transketolase
Gene Aliases
HEL-S-48, HEL107, TK, TKT1
UniGene
Hs.89643
Gene Chromosome Location
Chr.3: 53224707-53256114 on Build GRCh38
Transcripts
NM_001135055.2 XM_011534054.1

SNP Detail

SNP ID Chromosome Location
rs781972407 Chr.3: 53225242-53225242 on Build GRCh38
rs374773808 Chr.3: 53225162-53225162 on Build GRCh38
rs17306163 Chr.3: 53225139-53225139 on Build GRCh38
rs1041203939 Chr.3: 53225363-53225363 on Build GRCh38
rs936370760 Chr.3: 53225298-53225298 on Build GRCh38
rs573673575 Chr.3: 53225202-53225202 on Build GRCh38
rs116990537 Chr.3: 53225079-53225079 on Build GRCh38
rs1019798836 Chr.3: 53225128-53225128 on Build GRCh38
rs902206326 Chr.3: 53225059-53225059 on Build GRCh38
rs917767256 Chr.3: 53225221-53225221 on Build GRCh38
rs889502137 Chr.3: 53225314-53225314 on Build GRCh38
rs370083498 Chr.3: 53225144-53225144 on Build GRCh38
rs1042027445 Chr.3: 53225033-53225033 on Build GRCh38
rs558072712 Chr.3: 53224998-53224998 on Build GRCh38
rs1007031314 Chr.3: 53225120-53225120 on Build GRCh38
rs13093936 Chr.3: 53224997-53224997 on Build GRCh38
rs544477026 Chr.3: 53225320-53225320 on Build GRCh38
rs1026352958 Chr.3: 53225380-53225380 on Build GRCh38
rs949328842 Chr.3: 53225022-53225022 on Build GRCh38
rs571469565 Chr.3: 53225008-53225008 on Build GRCh38
rs1000603653 Chr.3: 53225066-53225066 on Build GRCh38
rs952844141 Chr.3: 53225184-53225184 on Build GRCh38
rs952734635 Chr.3: 53225385-53225385 on Build GRCh38
rs191955582 Chr.3: 53225109-53225109 on Build GRCh38
rs1053458676 Chr.3: 53225313-53225313 on Build GRCh38
rs923628069 Chr.3: 53225289-53225289 on Build GRCh38
rs374874866 Chr.3: 53225234-53225234 on Build GRCh38
rs990573218 Chr.3: 53225216-53225216 on Build GRCh38
rs782194200 Chr.3: 53225193-53225193 on Build GRCh38
rs994409594 Chr.3: 53225372-53225372 on Build GRCh38
rs927415860 Chr.3: 53225200-53225200 on Build GRCh38
rs782338577 Chr.3: 53225214-53225214 on Build GRCh38
rs73840245 Chr.3: 53225355-53225355 on Build GRCh38
rs1032134300 Chr.3: 53225102-53225102 on Build GRCh38
rs527662452 Chr.3: 53225371-53225371 on Build GRCh38
rs575243119 Chr.3: 53225243-53225243 on Build GRCh38
rs782554346 Chr.3: 53225305-53225306 on Build GRCh38
rs35706165 Chr.3: 53225315-53225316 on Build GRCh38
rs1056076 Chr.3: 53225038-53225038 on Build GRCh38

View Full Product Details