Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109214460-109214960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARS
Entrez Gene ID
6301
Gene Name
seryl-tRNA synthetase
Gene Aliases
SERRS, SERS
UniGene
Hs.531176
Gene Chromosome Location
Chr.1: 109213893-109238182 on Build GRCh38
Transcripts
NM_006513.3 NR_034072.1 XM_006710813.2

SNP Detail

SNP ID Chromosome Location
rs910484543 Chr.1: 109214527-109214527 on Build GRCh38
rs1025253883 Chr.1: 109214923-109214923 on Build GRCh38
rs772170570 Chr.1: 109214664-109214664 on Build GRCh38
rs745898215 Chr.1: 109214632-109214632 on Build GRCh38
rs1044816626 Chr.1: 109214493-109214494 on Build GRCh38
rs968855750 Chr.1: 109214635-109214635 on Build GRCh38
rs3203683 Chr.1: 109214840-109214840 on Build GRCh38
rs993503927 Chr.1: 109214595-109214595 on Build GRCh38
rs754898767 Chr.1: 109214680-109214680 on Build GRCh38
rs1017928009 Chr.1: 109214631-109214631 on Build GRCh38
rs891285600 Chr.1: 109214853-109214853 on Build GRCh38
rs770989871 Chr.1: 109214561-109214561 on Build GRCh38
rs370690317 Chr.1: 109214658-109214658 on Build GRCh38
rs1044486982 Chr.1: 109214925-109214925 on Build GRCh38
rs1057120614 Chr.1: 109214886-109214886 on Build GRCh38
rs1023110501 Chr.1: 109214634-109214634 on Build GRCh38
rs896367515 Chr.1: 109214487-109214487 on Build GRCh38
rs148074553 Chr.1: 109214555-109214555 on Build GRCh38
rs530798829 Chr.1: 109214615-109214615 on Build GRCh38
rs973794513 Chr.1: 109214644-109214644 on Build GRCh38
rs537110176 Chr.1: 109214493-109214493 on Build GRCh38
rs933159027 Chr.1: 109214503-109214503 on Build GRCh38
rs553222967 Chr.1: 109214604-109214604 on Build GRCh38
rs879081946 Chr.1: 109214884-109214884 on Build GRCh38
rs1010792291 Chr.1: 109214586-109214586 on Build GRCh38
rs904723887 Chr.1: 109214927-109214927 on Build GRCh38
rs941858634 Chr.1: 109214761-109214761 on Build GRCh38
rs562391480 Chr.1: 109214596-109214596 on Build GRCh38
rs866617558 Chr.1: 109214671-109214671 on Build GRCh38
rs559150571 Chr.1: 109214483-109214483 on Build GRCh38
rs985726210 Chr.1: 109214610-109214610 on Build GRCh38
rs544806592 Chr.1: 109214708-109214708 on Build GRCh38
rs1054563525 Chr.1: 109214748-109214748 on Build GRCh38
rs1025655134 Chr.1: 109214870-109214870 on Build GRCh38
rs1007559466 Chr.1: 109214860-109214860 on Build GRCh38
rs778744262 Chr.1: 109214611-109214611 on Build GRCh38
rs557032357 Chr.1: 109214505-109214505 on Build GRCh38
rs747370312 Chr.1: 109214666-109214666 on Build GRCh38
rs1030047583 Chr.1: 109214698-109214698 on Build GRCh38
rs962370201 Chr.1: 109214893-109214893 on Build GRCh38
rs779120546 Chr.1: 109214623-109214623 on Build GRCh38
rs190110682 Chr.1: 109214488-109214488 on Build GRCh38
rs4970829 Chr.1: 109214673-109214673 on Build GRCh38
rs765652260 Chr.1: 109214803-109214803 on Build GRCh38
rs895916350 Chr.1: 109214891-109214891 on Build GRCh38
rs947985283 Chr.1: 109214492-109214492 on Build GRCh38
rs769053476 Chr.1: 109214763-109214763 on Build GRCh38
rs1044996984 Chr.1: 109214697-109214697 on Build GRCh38
rs761136922 Chr.1: 109214726-109214726 on Build GRCh38
rs575486242 Chr.1: 109214720-109214720 on Build GRCh38
rs1056404259 Chr.1: 109214486-109214487 on Build GRCh38
rs776929458 Chr.1: 109214825-109214825 on Build GRCh38
rs141792440 Chr.1: 109214921-109214921 on Build GRCh38
rs948006124 Chr.1: 109214696-109214696 on Build GRCh38
rs1013098554 Chr.1: 109214633-109214633 on Build GRCh38
rs941920250 Chr.1: 109214545-109214545 on Build GRCh38

View Full Product Details