Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112045663-112046164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS7
Entrez Gene ID
344805
Gene Name
transmembrane protease, serine 7
Gene Aliases
-
UniGene
Hs.435490
Gene Chromosome Location
Chr.3: 112039067-112081269 on Build GRCh38
Transcripts
NM_001042575.2 NR_026734.1 XM_011512754.1 XM_011512758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1417758 c.214C>T p.R72C Chr.3: 112045805-112045805 on Build GRCh38
4437484 c.175G>A p.G59S Chr.3: 112045766-112045766 on Build GRCh38
7262320 c.162-10G>A p.? Chr.3: 112045743-112045743 on Build GRCh38
6248598 c.304G>T p.G102W Chr.3: 112045895-112045895 on Build GRCh38
5720117 c.316G>A p.G106R Chr.3: 112045907-112045907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774693646 Chr.3: 112046088-112046088 on Build GRCh38
rs749652729 Chr.3: 112045978-112045978 on Build GRCh38
rs769381998 Chr.3: 112045805-112045805 on Build GRCh38
rs186760163 Chr.3: 112046064-112046064 on Build GRCh38
rs899507049 Chr.3: 112045773-112045773 on Build GRCh38
rs1010197166 Chr.3: 112046072-112046075 on Build GRCh38
rs920918040 Chr.3: 112046115-112046115 on Build GRCh38
rs116816360 Chr.3: 112046065-112046065 on Build GRCh38
rs938414430 Chr.3: 112045834-112045834 on Build GRCh38
rs377534244 Chr.3: 112046067-112046067 on Build GRCh38
rs755506679 Chr.3: 112045870-112045870 on Build GRCh38
rs1030137324 Chr.3: 112045730-112045730 on Build GRCh38
rs756571384 Chr.3: 112045806-112045806 on Build GRCh38
rs987929866 Chr.3: 112046017-112046017 on Build GRCh38
rs986628382 Chr.3: 112045773-112045787 on Build GRCh38
rs946496103 Chr.3: 112045759-112045759 on Build GRCh38
rs532108333 Chr.3: 112045712-112045713 on Build GRCh38
rs376478648 Chr.3: 112045884-112045884 on Build GRCh38
rs527984904 Chr.3: 112046083-112046083 on Build GRCh38
rs371377353 Chr.3: 112045903-112045903 on Build GRCh38
rs997070466 Chr.3: 112045797-112045797 on Build GRCh38
rs538810957 Chr.3: 112045700-112045700 on Build GRCh38
rs780510102 Chr.3: 112045822-112045822 on Build GRCh38
rs757337665 Chr.3: 112045765-112045765 on Build GRCh38
rs574230817 Chr.3: 112045902-112045902 on Build GRCh38
rs375279190 Chr.3: 112045927-112045927 on Build GRCh38
rs576309342 Chr.3: 112045691-112045691 on Build GRCh38
rs767673800 Chr.3: 112045780-112045780 on Build GRCh38
rs544020562 Chr.3: 112046096-112046096 on Build GRCh38
rs776475324 Chr.3: 112045987-112045987 on Build GRCh38
rs770713452 Chr.3: 112045970-112045970 on Build GRCh38
rs745544402 Chr.3: 112045687-112045687 on Build GRCh38
rs558597001 Chr.3: 112045738-112045738 on Build GRCh38
rs939200892 Chr.3: 112045714-112045714 on Build GRCh38
rs960633136 Chr.3: 112046013-112046013 on Build GRCh38
rs746604580 Chr.3: 112045956-112045956 on Build GRCh38
rs759400725 Chr.3: 112045742-112045742 on Build GRCh38
rs370919980 Chr.3: 112045885-112045885 on Build GRCh38
rs201851007 Chr.3: 112045917-112045917 on Build GRCh38
rs745528613 Chr.3: 112046122-112046122 on Build GRCh38
rs1033994574 Chr.3: 112046097-112046097 on Build GRCh38
rs980685387 Chr.3: 112045980-112045980 on Build GRCh38
rs147939107 Chr.3: 112045934-112045934 on Build GRCh38
rs866247414 Chr.3: 112045844-112045844 on Build GRCh38
rs750634407 Chr.3: 112045785-112045785 on Build GRCh38
rs927020265 Chr.3: 112045833-112045833 on Build GRCh38
rs749820920 Chr.3: 112045847-112045847 on Build GRCh38
rs771202123 Chr.3: 112045921-112045921 on Build GRCh38
rs1021530846 Chr.3: 112046085-112046085 on Build GRCh38
rs901267326 Chr.3: 112045999-112045999 on Build GRCh38
rs927870077 Chr.3: 112045996-112045996 on Build GRCh38
rs998240216 Chr.3: 112046047-112046047 on Build GRCh38
rs1057488596 Chr.3: 112045843-112045843 on Build GRCh38
rs181255206 Chr.3: 112045793-112045793 on Build GRCh38
rs565473222 Chr.3: 112046068-112046068 on Build GRCh38
rs765214425 Chr.3: 112045743-112045743 on Build GRCh38
rs572291797 Chr.3: 112045750-112045750 on Build GRCh38
rs955941842 Chr.3: 112045731-112045731 on Build GRCh38
rs932323496 Chr.3: 112046117-112046117 on Build GRCh38
rs996030573 Chr.3: 112045887-112045887 on Build GRCh38
rs562996735 Chr.3: 112046049-112046049 on Build GRCh38
rs942685800 Chr.3: 112045935-112045936 on Build GRCh38
rs57759994 Chr.3: 112046111-112046111 on Build GRCh38

View Full Product Details