Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112061486-112062013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS7
Entrez Gene ID
344805
Gene Name
transmembrane protease, serine 7
Gene Aliases
-
UniGene
Hs.435490
Gene Chromosome Location
Chr.3: 112039067-112081269 on Build GRCh38
Transcripts
NM_001042575.2 NR_026734.1 XM_011512754.1 XM_011512758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4983485 c.970C>T p.R324* Chr.3: 112061824-112061824 on Build GRCh38
7230669 c.1002C>G p.L334L Chr.3: 112061856-112061856 on Build GRCh38
5397802 c.980G>A p.S327N Chr.3: 112061834-112061834 on Build GRCh38
3118168 c.958C>T p.Q320* Chr.3: 112061812-112061812 on Build GRCh38
5346868 c.1029G>T p.K343N Chr.3: 112061883-112061883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952583569 Chr.3: 112061802-112061802 on Build GRCh38
rs965253867 Chr.3: 112061832-112061832 on Build GRCh38
rs757002153 Chr.3: 112061923-112061923 on Build GRCh38
rs770907508 Chr.3: 112061751-112061751 on Build GRCh38
rs753517442 Chr.3: 112061791-112061791 on Build GRCh38
rs762143422 Chr.3: 112061748-112061748 on Build GRCh38
rs777386661 Chr.3: 112061801-112061801 on Build GRCh38
rs934279955 Chr.3: 112061580-112061580 on Build GRCh38
rs750622452 Chr.3: 112061849-112061849 on Build GRCh38
rs771221099 Chr.3: 112061966-112061966 on Build GRCh38
rs772288893 Chr.3: 112061961-112061961 on Build GRCh38
rs372885822 Chr.3: 112061831-112061831 on Build GRCh38
rs973879002 Chr.3: 112061550-112061550 on Build GRCh38
rs766493367 Chr.3: 112061895-112061895 on Build GRCh38
rs773173912 Chr.3: 112061570-112061570 on Build GRCh38
rs372640419 Chr.3: 112061770-112061770 on Build GRCh38
rs900665920 Chr.3: 112061664-112061664 on Build GRCh38
rs182776846 Chr.3: 112061869-112061869 on Build GRCh38
rs550816319 Chr.3: 112061557-112061557 on Build GRCh38
rs373363349 Chr.3: 112061791-112061791 on Build GRCh38
rs200385589 Chr.3: 112061880-112061880 on Build GRCh38
rs1012499842 Chr.3: 112061539-112061539 on Build GRCh38
rs75223055 Chr.3: 112061589-112061589 on Build GRCh38
rs976359473 Chr.3: 112061717-112061717 on Build GRCh38
rs1016284642 Chr.3: 112061692-112061692 on Build GRCh38
rs769810585 Chr.3: 112061878-112061878 on Build GRCh38
rs747639255 Chr.3: 112061792-112061792 on Build GRCh38
rs73853301 Chr.3: 112061782-112061782 on Build GRCh38
rs148656864 Chr.3: 112061799-112061799 on Build GRCh38
rs997672914 Chr.3: 112061670-112061670 on Build GRCh38
rs73853300 Chr.3: 112061729-112061729 on Build GRCh38
rs771864037 Chr.3: 112061740-112061740 on Build GRCh38
rs16859132 Chr.3: 112061894-112061894 on Build GRCh38
rs761150028 Chr.3: 112061854-112061854 on Build GRCh38
rs371897494 Chr.3: 112061741-112061741 on Build GRCh38
rs755563495 Chr.3: 112061866-112061866 on Build GRCh38
rs759266488 Chr.3: 112061674-112061674 on Build GRCh38
rs369604109 Chr.3: 112061769-112061769 on Build GRCh38
rs947082562 Chr.3: 112061798-112061798 on Build GRCh38
rs751056782 Chr.3: 112061922-112061922 on Build GRCh38
rs761953101 Chr.3: 112061842-112061842 on Build GRCh38
rs566952680 Chr.3: 112061635-112061635 on Build GRCh38
rs200310602 Chr.3: 112061868-112061868 on Build GRCh38
rs767847233 Chr.3: 112061777-112061777 on Build GRCh38
rs114324071 Chr.3: 112061571-112061571 on Build GRCh38
rs542321006 Chr.3: 112061835-112061835 on Build GRCh38
rs994928388 Chr.3: 112061789-112061789 on Build GRCh38
rs144741324 Chr.3: 112061749-112061749 on Build GRCh38
rs764511706 Chr.3: 112061965-112061965 on Build GRCh38
rs753165112 Chr.3: 112061904-112061904 on Build GRCh38
rs371646957 Chr.3: 112061825-112061825 on Build GRCh38
rs772237307 Chr.3: 112061883-112061883 on Build GRCh38
rs745646914 Chr.3: 112061876-112061876 on Build GRCh38
rs1017835060 Chr.3: 112061661-112061661 on Build GRCh38
rs762184222 Chr.3: 112061772-112061772 on Build GRCh38
rs760491819 Chr.3: 112061583-112061583 on Build GRCh38
rs943725720 Chr.3: 112061706-112061706 on Build GRCh38
rs976411967 Chr.3: 112061837-112061837 on Build GRCh38
rs340142 Chr.3: 112061824-112061824 on Build GRCh38
rs865834027 Chr.3: 112061937-112061937 on Build GRCh38
rs995211767 Chr.3: 112061934-112061934 on Build GRCh38
rs566712186 Chr.3: 112061929-112061929 on Build GRCh38
rs765799417 Chr.3: 112061903-112061903 on Build GRCh38
rs200937737 Chr.3: 112061846-112061846 on Build GRCh38
rs763538426 Chr.3: 112061912-112061912 on Build GRCh38
rs774651051 Chr.3: 112061702-112061702 on Build GRCh38
rs757911010 Chr.3: 112061907-112061907 on Build GRCh38
rs553086504 Chr.3: 112061628-112061628 on Build GRCh38
rs747052700 Chr.3: 112061964-112061964 on Build GRCh38
rs370856661 Chr.3: 112061859-112061859 on Build GRCh38
rs915867834 Chr.3: 112061742-112061742 on Build GRCh38
rs747766716 Chr.3: 112061738-112061738 on Build GRCh38
rs766696441 Chr.3: 112061855-112061855 on Build GRCh38
rs535244607 Chr.3: 112061673-112061673 on Build GRCh38
rs770646460 Chr.3: 112061875-112061875 on Build GRCh38
rs573163699 Chr.3: 112061957-112061957 on Build GRCh38
rs754327766 Chr.3: 112061785-112061785 on Build GRCh38
rs764979638 Chr.3: 112061727-112061727 on Build GRCh38
rs546578563 Chr.3: 112061901-112061901 on Build GRCh38
rs746831777 Chr.3: 112061811-112061811 on Build GRCh38
rs73853302 Chr.3: 112061783-112061783 on Build GRCh38
rs746600592 Chr.3: 112061872-112061872 on Build GRCh38
rs948262927 Chr.3: 112061893-112061893 on Build GRCh38
rs777116498 Chr.3: 112061900-112061900 on Build GRCh38
rs192353447 Chr.3: 112061553-112061553 on Build GRCh38
rs948534435 Chr.3: 112061753-112061753 on Build GRCh38
rs199807220 Chr.3: 112061933-112061933 on Build GRCh38
rs538770557 Chr.3: 112061971-112061971 on Build GRCh38
rs773903549 Chr.3: 112061737-112061737 on Build GRCh38
rs374926360 Chr.3: 112061782-112061783 on Build GRCh38
rs187367994 Chr.3: 112061887-112061887 on Build GRCh38
rs750857353 Chr.3: 112061781-112061781 on Build GRCh38
rs1015031835 Chr.3: 112061572-112061572 on Build GRCh38
rs1030828858 Chr.3: 112061671-112061671 on Build GRCh38
rs777437309 Chr.3: 112061871-112061871 on Build GRCh38
rs776673366 Chr.3: 112061968-112061968 on Build GRCh38
rs754448505 Chr.3: 112061786-112061786 on Build GRCh38
rs770499697 Chr.3: 112061584-112061584 on Build GRCh38
rs201583103 Chr.3: 112061765-112061765 on Build GRCh38
rs749169362 Chr.3: 112061959-112061959 on Build GRCh38
rs773045252 Chr.3: 112061743-112061743 on Build GRCh38
rs751744890 Chr.3: 112061771-112061771 on Build GRCh38
rs373850617 Chr.3: 112061962-112061962 on Build GRCh38
rs780678214 Chr.3: 112061924-112061924 on Build GRCh38

View Full Product Details