Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112074098-112074605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS7
Entrez Gene ID
344805
Gene Name
transmembrane protease, serine 7
Gene Aliases
-
UniGene
Hs.435490
Gene Chromosome Location
Chr.3: 112039067-112081269 on Build GRCh38
Transcripts
NM_001042575.2 NR_026734.1 XM_011512754.1 XM_011512758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6162694 c.1393G>A p.E465K Chr.3: 112074400-112074400 on Build GRCh38
1036318 c.1359T>A p.C453* Chr.3: 112074366-112074366 on Build GRCh38
7233943 c.1389T>G p.S463R Chr.3: 112074396-112074396 on Build GRCh38
479210 c.1379C>A p.P460Q Chr.3: 112074386-112074386 on Build GRCh38
341336 c.928G>A p.G310R Chr.3: 112074370-112074370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750876841 Chr.3: 112074354-112074354 on Build GRCh38
rs773135339 Chr.3: 112074395-112074395 on Build GRCh38
rs762923495 Chr.3: 112074408-112074408 on Build GRCh38
rs188194931 Chr.3: 112074509-112074509 on Build GRCh38
rs576583178 Chr.3: 112074504-112074504 on Build GRCh38
rs752710198 Chr.3: 112074310-112074310 on Build GRCh38
rs769719396 Chr.3: 112074127-112074127 on Build GRCh38
rs371106755 Chr.3: 112074311-112074311 on Build GRCh38
rs748912100 Chr.3: 112074249-112074249 on Build GRCh38
rs529689914 Chr.3: 112074220-112074220 on Build GRCh38
rs781491048 Chr.3: 112074246-112074247 on Build GRCh38
rs143185999 Chr.3: 112074403-112074403 on Build GRCh38
rs926825579 Chr.3: 112074166-112074166 on Build GRCh38
rs918049263 Chr.3: 112074469-112074469 on Build GRCh38
rs1006734796 Chr.3: 112074173-112074173 on Build GRCh38
rs985045493 Chr.3: 112074480-112074480 on Build GRCh38
rs748246898 Chr.3: 112074370-112074370 on Build GRCh38
rs754343964 Chr.3: 112074223-112074223 on Build GRCh38
rs763207446 Chr.3: 112074267-112074267 on Build GRCh38
rs543410683 Chr.3: 112074275-112074275 on Build GRCh38
rs879055630 Chr.3: 112074559-112074559 on Build GRCh38
rs778511548 Chr.3: 112074374-112074374 on Build GRCh38
rs903084125 Chr.3: 112074291-112074291 on Build GRCh38
rs759561813 Chr.3: 112074285-112074285 on Build GRCh38
rs899696455 Chr.3: 112074179-112074179 on Build GRCh38
rs770145338 Chr.3: 112074461-112074462 on Build GRCh38
rs199812280 Chr.3: 112074365-112074365 on Build GRCh38
rs953277298 Chr.3: 112074317-112074317 on Build GRCh38
rs929355884 Chr.3: 112074146-112074146 on Build GRCh38
rs1048199227 Chr.3: 112074153-112074153 on Build GRCh38
rs1032838647 Chr.3: 112074304-112074304 on Build GRCh38
rs770772785 Chr.3: 112074397-112074397 on Build GRCh38
rs752074324 Chr.3: 112074213-112074213 on Build GRCh38
rs560843163 Chr.3: 112074200-112074200 on Build GRCh38
rs372741294 Chr.3: 112074352-112074352 on Build GRCh38
rs757647862 Chr.3: 112074334-112074334 on Build GRCh38
rs896442814 Chr.3: 112074235-112074235 on Build GRCh38
rs762274303 Chr.3: 112074454-112074454 on Build GRCh38
rs768393529 Chr.3: 112074372-112074372 on Build GRCh38
rs370180770 Chr.3: 112074508-112074508 on Build GRCh38
rs761816548 Chr.3: 112074246-112074246 on Build GRCh38
rs868314540 Chr.3: 112074431-112074431 on Build GRCh38
rs183473278 Chr.3: 112074280-112074280 on Build GRCh38
rs1041632162 Chr.3: 112074288-112074288 on Build GRCh38
rs760726716 Chr.3: 112074396-112074396 on Build GRCh38
rs201392669 Chr.3: 112074263-112074263 on Build GRCh38
rs752020708 Chr.3: 112074320-112074320 on Build GRCh38
rs191454 Chr.3: 112074469-112074469 on Build GRCh38
rs896737860 Chr.3: 112074130-112074130 on Build GRCh38
rs1017671963 Chr.3: 112074175-112074175 on Build GRCh38
rs768178999 Chr.3: 112074262-112074262 on Build GRCh38
rs966071277 Chr.3: 112074426-112074426 on Build GRCh38
rs779671840 Chr.3: 112074361-112074361 on Build GRCh38
rs747955696 Chr.3: 112074376-112074376 on Build GRCh38
rs750596951 Chr.3: 112074486-112074486 on Build GRCh38
rs771811062 Chr.3: 112074390-112074390 on Build GRCh38
rs765306367 Chr.3: 112074302-112074302 on Build GRCh38
rs368517173 Chr.3: 112074421-112074421 on Build GRCh38
rs773080251 Chr.3: 112074283-112074283 on Build GRCh38
rs1047951461 Chr.3: 112074471-112074471 on Build GRCh38
rs72942186 Chr.3: 112074572-112074572 on Build GRCh38
rs769308774 Chr.3: 112074245-112074245 on Build GRCh38
rs774675425 Chr.3: 112074436-112074436 on Build GRCh38
rs755590902 Chr.3: 112074357-112074357 on Build GRCh38
rs540964867 Chr.3: 112074162-112074162 on Build GRCh38
rs1029625260 Chr.3: 112074216-112074220 on Build GRCh38
rs988042962 Chr.3: 112074279-112074279 on Build GRCh38
rs774196425 Chr.3: 112074270-112074270 on Build GRCh38
rs764383058 Chr.3: 112074315-112074315 on Build GRCh38
rs368141776 Chr.3: 112074247-112074247 on Build GRCh38
rs971723998 Chr.3: 112074449-112074449 on Build GRCh38
rs952287713 Chr.3: 112074474-112074474 on Build GRCh38

View Full Product Details