Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120411763-120412240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL1
Entrez Gene ID
11167
Gene Name
follistatin like 1
Gene Aliases
FRP, FSL1, MIR198, OCC-1, OCC1, tsc36
UniGene
Hs.269512
Gene Chromosome Location
Chr.3: 120394214-120451071 on Build GRCh38
Transcripts
NM_007085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5949197 c.298+10G>C p.? Chr.3: 120411844-120411844 on Build GRCh38
5949169 c.203G>C p.S68T Chr.3: 120411949-120411949 on Build GRCh38
479266 c.202A>C p.S68R Chr.3: 120411950-120411950 on Build GRCh38
5949209 c.173G>A p.C58Y Chr.3: 120411979-120411979 on Build GRCh38
281258 c.298G>T p.E100* Chr.3: 120411854-120411854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147567351 Chr.3: 120412098-120412098 on Build GRCh38
rs374833559 Chr.3: 120411810-120411810 on Build GRCh38
rs550029447 Chr.3: 120412050-120412050 on Build GRCh38
rs767516382 Chr.3: 120412051-120412052 on Build GRCh38
rs767632221 Chr.3: 120411916-120411916 on Build GRCh38
rs377762680 Chr.3: 120412011-120412011 on Build GRCh38
rs755359984 Chr.3: 120411995-120411995 on Build GRCh38
rs529215697 Chr.3: 120412072-120412072 on Build GRCh38
rs59060347 Chr.3: 120412055-120412056 on Build GRCh38
rs368128153 Chr.3: 120411817-120411817 on Build GRCh38
rs969975605 Chr.3: 120412025-120412025 on Build GRCh38
rs1004418784 Chr.3: 120411829-120411829 on Build GRCh38
rs374114238 Chr.3: 120411962-120411962 on Build GRCh38
rs777920066 Chr.3: 120411836-120411837 on Build GRCh38
rs199556968 Chr.3: 120411869-120411869 on Build GRCh38
rs749258496 Chr.3: 120412006-120412009 on Build GRCh38
rs762080512 Chr.3: 120411910-120411910 on Build GRCh38
rs933137369 Chr.3: 120412029-120412058 on Build GRCh38
rs771916645 Chr.3: 120412004-120412004 on Build GRCh38
rs746444048 Chr.3: 120412009-120412009 on Build GRCh38
rs780379738 Chr.3: 120411961-120411961 on Build GRCh38
rs145054908 Chr.3: 120411864-120411864 on Build GRCh38
rs868766499 Chr.3: 120411814-120411814 on Build GRCh38
rs759715717 Chr.3: 120411870-120411870 on Build GRCh38
rs112243174 Chr.3: 120412039-120412040 on Build GRCh38
rs769168863 Chr.3: 120411856-120411856 on Build GRCh38
rs747786469 Chr.3: 120412001-120412001 on Build GRCh38
rs67083358 Chr.3: 120412037-120412058 on Build GRCh38
rs750406713 Chr.3: 120411917-120411917 on Build GRCh38
rs944436458 Chr.3: 120412151-120412151 on Build GRCh38
rs757298109 Chr.3: 120411813-120411813 on Build GRCh38
rs796132602 Chr.3: 120412037-120412038 on Build GRCh38
rs113062511 Chr.3: 120411840-120411840 on Build GRCh38
rs202002329 Chr.3: 120412002-120412002 on Build GRCh38
rs565792750 Chr.3: 120412187-120412187 on Build GRCh38
rs772305394 Chr.3: 120411915-120411915 on Build GRCh38
rs776988994 Chr.3: 120412152-120412152 on Build GRCh38
rs759276167 Chr.3: 120411886-120411886 on Build GRCh38
rs771681801 Chr.3: 120411868-120411868 on Build GRCh38
rs895922658 Chr.3: 120411890-120411890 on Build GRCh38
rs200274599 Chr.3: 120411838-120411838 on Build GRCh38
rs756234431 Chr.3: 120411933-120411933 on Build GRCh38
rs911676009 Chr.3: 120412127-120412127 on Build GRCh38
rs960385546 Chr.3: 120411876-120411876 on Build GRCh38
rs1259326 Chr.3: 120412052-120412052 on Build GRCh38
rs185906368 Chr.3: 120412027-120412027 on Build GRCh38
rs766123815 Chr.3: 120412197-120412197 on Build GRCh38
rs921597970 Chr.3: 120412058-120412059 on Build GRCh38
rs950143871 Chr.3: 120412209-120412209 on Build GRCh38
rs1021763086 Chr.3: 120411867-120411867 on Build GRCh38
rs201873665 Chr.3: 120412037-120412038 on Build GRCh38
rs756649712 Chr.3: 120411818-120411818 on Build GRCh38
rs55876532 Chr.3: 120412050-120412071 on Build GRCh38
rs780619417 Chr.3: 120411837-120411837 on Build GRCh38
rs769564796 Chr.3: 120412028-120412028 on Build GRCh38
rs751760368 Chr.3: 120411901-120411901 on Build GRCh38
rs992866173 Chr.3: 120411892-120411892 on Build GRCh38
rs930345750 Chr.3: 120412070-120412070 on Build GRCh38
rs540819614 Chr.3: 120412020-120412020 on Build GRCh38
rs984382445 Chr.3: 120412073-120412073 on Build GRCh38
rs983683454 Chr.3: 120412100-120412100 on Build GRCh38
rs764241520 Chr.3: 120411805-120411805 on Build GRCh38
rs776641591 Chr.3: 120411884-120411884 on Build GRCh38
rs766161987 Chr.3: 120411877-120411877 on Build GRCh38
rs561731537 Chr.3: 120411999-120411999 on Build GRCh38
rs879578060 Chr.3: 120412140-120412141 on Build GRCh38
rs929163327 Chr.3: 120412090-120412090 on Build GRCh38
rs1037215475 Chr.3: 120412142-120412142 on Build GRCh38
rs974929137 Chr.3: 120412057-120412058 on Build GRCh38
rs777485676 Chr.3: 120412006-120412006 on Build GRCh38
rs200742219 Chr.3: 120411866-120411866 on Build GRCh38
rs965783129 Chr.3: 120412051-120412052 on Build GRCh38
rs988609527 Chr.3: 120412044-120412057 on Build GRCh38
rs781298228 Chr.3: 120411815-120411815 on Build GRCh38
rs765058093 Chr.3: 120411900-120411900 on Build GRCh38
rs375088384 Chr.3: 120412105-120412105 on Build GRCh38
rs910143449 Chr.3: 120412129-120412129 on Build GRCh38
rs940147175 Chr.3: 120412135-120412136 on Build GRCh38
rs756353583 Chr.3: 120412036-120412037 on Build GRCh38
rs533123240 Chr.3: 120412049-120412049 on Build GRCh38
rs112712040 Chr.3: 120412056-120412056 on Build GRCh38

View Full Product Details