Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 131152298-131152803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK11
Entrez Gene ID
79858
Gene Name
NIMA related kinase 11
Gene Aliases
-
UniGene
Hs.657336
Gene Chromosome Location
Chr.3: 131026850-131350465 on Build GRCh38
Transcripts
NM_001146003.1 NM_001321220.1 NM_001321221.1 NM_001321222.1 NM_001321223.1 NM_001321224.1 NM_024800.4 NM_145910.3 XM_011513167.2 XM_011513173.1 XM_011513174.1 XM_011513175.1 XM_011513181.1 XM_017007205.1 XM_017007206.1 XM_017007207.1 XM_017007208.1 XM_017007209.1 XM_017007210.1 XM_017007211.1 XM_017007212.1 XM_017007213.1 XM_017007214.1 XM_017007215.1 XM_017007216.1 XM_017007217.1 XM_017007218.1 XM_017007219.1 XM_017007220.1 XM_017007221.1 XM_017007222.1 XM_017007223.1 XM_017007224.1 XM_017007225.1 XM_017007226.1 XM_017007227.1 XM_017007228.1 XM_017007229.1 XM_017007230.1 XM_017007231.1 XM_017007232.1 XM_017007233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3587485 c.688C>T p.H230Y Chr.3: 131152428-131152428 on Build GRCh38
7082104 c.764G>A p.R255K Chr.3: 131152504-131152504 on Build GRCh38
3587487 c.852C>T p.I284I Chr.3: 131152685-131152685 on Build GRCh38
6572318 c.760G>C p.E254Q Chr.3: 131152500-131152500 on Build GRCh38
419830 c.764G>A p.R255K Chr.3: 131152504-131152504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758562940 Chr.3: 131152502-131152502 on Build GRCh38
rs927238942 Chr.3: 131152743-131152743 on Build GRCh38
rs763736419 Chr.3: 131152396-131152396 on Build GRCh38
rs758099709 Chr.3: 131152395-131152395 on Build GRCh38
rs765783234 Chr.3: 131152591-131152591 on Build GRCh38
rs532250406 Chr.3: 131152430-131152430 on Build GRCh38
rs766974591 Chr.3: 131152546-131152546 on Build GRCh38
rs138886684 Chr.3: 131152709-131152709 on Build GRCh38
rs772315660 Chr.3: 131152420-131152420 on Build GRCh38
rs756894510 Chr.3: 131152465-131152465 on Build GRCh38
rs190113760 Chr.3: 131152365-131152365 on Build GRCh38
rs755357329 Chr.3: 131152408-131152408 on Build GRCh38
rs778100885 Chr.3: 131152609-131152609 on Build GRCh38
rs1000620375 Chr.3: 131152631-131152631 on Build GRCh38
rs781491564 Chr.3: 131152752-131152752 on Build GRCh38
rs547475674 Chr.3: 131152437-131152437 on Build GRCh38
rs763431929 Chr.3: 131152742-131152742 on Build GRCh38
rs758831166 Chr.3: 131152602-131152602 on Build GRCh38
rs753265082 Chr.3: 131152740-131152740 on Build GRCh38
rs760233095 Chr.3: 131152701-131152701 on Build GRCh38
rs374241502 Chr.3: 131152518-131152518 on Build GRCh38
rs779316802 Chr.3: 131152413-131152413 on Build GRCh38
rs905782421 Chr.3: 131152346-131152346 on Build GRCh38
rs150253539 Chr.3: 131152508-131152508 on Build GRCh38
rs781312028 Chr.3: 131152516-131152516 on Build GRCh38
rs371833879 Chr.3: 131152391-131152391 on Build GRCh38
rs753132110 Chr.3: 131152598-131152598 on Build GRCh38
rs962936171 Chr.3: 131152557-131152557 on Build GRCh38
rs776495361 Chr.3: 131152432-131152432 on Build GRCh38
rs773247820 Chr.3: 131152370-131152370 on Build GRCh38
rs374446054 Chr.3: 131152386-131152386 on Build GRCh38
rs201525279 Chr.3: 131152721-131152721 on Build GRCh38
rs771775294 Chr.3: 131152691-131152691 on Build GRCh38
rs770977418 Chr.3: 131152515-131152515 on Build GRCh38
rs776569792 Chr.3: 131152388-131152388 on Build GRCh38
rs1007050228 Chr.3: 131152505-131152505 on Build GRCh38
rs774204736 Chr.3: 131152541-131152541 on Build GRCh38
rs983567604 Chr.3: 131152653-131152653 on Build GRCh38
rs201954356 Chr.3: 131152530-131152530 on Build GRCh38
rs777979775 Chr.3: 131152512-131152512 on Build GRCh38
rs544037710 Chr.3: 131152324-131152324 on Build GRCh38
rs757515340 Chr.3: 131152746-131152746 on Build GRCh38
rs767041777 Chr.3: 131152469-131152469 on Build GRCh38
rs781232737 Chr.3: 131152640-131152640 on Build GRCh38
rs111740052 Chr.3: 131152558-131152558 on Build GRCh38
rs761658685 Chr.3: 131152678-131152678 on Build GRCh38
rs759501857 Chr.3: 131152436-131152436 on Build GRCh38
rs747231645 Chr.3: 131152514-131152514 on Build GRCh38
rs752348297 Chr.3: 131152392-131152392 on Build GRCh38
rs182509330 Chr.3: 131152671-131152671 on Build GRCh38
rs774505284 Chr.3: 131152359-131152359 on Build GRCh38
rs779244638 Chr.3: 131152498-131152498 on Build GRCh38
rs766330742 Chr.3: 131152380-131152380 on Build GRCh38
rs370179098 Chr.3: 131152372-131152372 on Build GRCh38
rs759250887 Chr.3: 131152390-131152390 on Build GRCh38
rs760052104 Chr.3: 131152590-131152590 on Build GRCh38
rs763861611 Chr.3: 131152441-131152441 on Build GRCh38
rs745984195 Chr.3: 131152643-131152643 on Build GRCh38
rs35567155 Chr.3: 131152527-131152527 on Build GRCh38
rs751642011 Chr.3: 131152479-131152479 on Build GRCh38
rs773377290 Chr.3: 131152423-131152423 on Build GRCh38
rs751831736 Chr.3: 131152624-131152624 on Build GRCh38
rs767561326 Chr.3: 131152404-131152404 on Build GRCh38
rs755476889 Chr.3: 131152488-131152488 on Build GRCh38
rs200414683 Chr.3: 131152726-131152726 on Build GRCh38
rs765771697 Chr.3: 131152406-131152406 on Build GRCh38
rs774254639 Chr.3: 131152675-131152675 on Build GRCh38
rs575424163 Chr.3: 131152360-131152360 on Build GRCh38
rs775243170 Chr.3: 131152523-131152523 on Build GRCh38
rs768998511 Chr.3: 131152337-131152337 on Build GRCh38
rs757472593 Chr.3: 131152630-131152630 on Build GRCh38
rs749977039 Chr.3: 131152582-131152582 on Build GRCh38
rs754350317 Chr.3: 131152480-131152480 on Build GRCh38
rs6765954 Chr.3: 131152499-131152499 on Build GRCh38
rs780042429 Chr.3: 131152349-131152349 on Build GRCh38
rs775322923 Chr.3: 131152440-131152440 on Build GRCh38
rs769704761 Chr.3: 131152670-131152670 on Build GRCh38
rs544061001 Chr.3: 131152775-131152775 on Build GRCh38
rs1029831619 Chr.3: 131152494-131152494 on Build GRCh38
rs772679025 Chr.3: 131152692-131152692 on Build GRCh38
rs751145856 Chr.3: 131152397-131152397 on Build GRCh38
rs761534097 Chr.3: 131152542-131152542 on Build GRCh38
rs111824170 Chr.3: 131152759-131152759 on Build GRCh38
rs756777968 Chr.3: 131152399-131152399 on Build GRCh38
rs1040285554 Chr.3: 131152509-131152509 on Build GRCh38
rs141613944 Chr.3: 131152744-131152744 on Build GRCh38
rs751190905 Chr.3: 131152459-131152459 on Build GRCh38
rs1031149457 Chr.3: 131152668-131152668 on Build GRCh38
rs745835190 Chr.3: 131152517-131152517 on Build GRCh38
rs748361250 Chr.3: 131152416-131152416 on Build GRCh38

View Full Product Details