Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138462002-138462500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESYT3
Entrez Gene ID
83850
Gene Name
extended synaptotagmin 3
Gene Aliases
CHR3SYT, E-Syt3, FAM62C
UniGene
Hs.477711
Gene Chromosome Location
Chr.3: 138434573-138479942 on Build GRCh38
Transcripts
NM_001322831.1 NM_001322834.1 NM_031913.4 NR_136409.1 XM_017007307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2155829 c.849C>G p.P283P Chr.3: 138462140-138462140 on Build GRCh38
2155793 c.845T>G p.L282R Chr.3: 138462136-138462136 on Build GRCh38
3587975 c.900C>T p.R300R Chr.3: 138462191-138462191 on Build GRCh38
5691445 c.888G>C p.L296L Chr.3: 138462179-138462179 on Build GRCh38
5708487 c.896T>A p.L299Q Chr.3: 138462187-138462187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879598705 Chr.3: 138462348-138462348 on Build GRCh38
rs996695986 Chr.3: 138462282-138462282 on Build GRCh38
rs370385604 Chr.3: 138462418-138462418 on Build GRCh38
rs762559847 Chr.3: 138462274-138462274 on Build GRCh38
rs528740470 Chr.3: 138462053-138462053 on Build GRCh38
rs769145835 Chr.3: 138462268-138462268 on Build GRCh38
rs756084496 Chr.3: 138462232-138462232 on Build GRCh38
rs955186835 Chr.3: 138462028-138462028 on Build GRCh38
rs772763628 Chr.3: 138462255-138462255 on Build GRCh38
rs543792035 Chr.3: 138462409-138462409 on Build GRCh38
rs555265855 Chr.3: 138462375-138462375 on Build GRCh38
rs557407338 Chr.3: 138462145-138462145 on Build GRCh38
rs955342621 Chr.3: 138462333-138462333 on Build GRCh38
rs756509546 Chr.3: 138462446-138462446 on Build GRCh38
rs764956189 Chr.3: 138462190-138462190 on Build GRCh38
rs749975880 Chr.3: 138462201-138462201 on Build GRCh38
rs899644332 Chr.3: 138462277-138462277 on Build GRCh38
rs1015295874 Chr.3: 138462342-138462342 on Build GRCh38
rs778091411 Chr.3: 138462239-138462239 on Build GRCh38
rs112771629 Chr.3: 138462123-138462123 on Build GRCh38
rs774898275 Chr.3: 138462251-138462251 on Build GRCh38
rs900088406 Chr.3: 138462365-138462365 on Build GRCh38
rs1057092960 Chr.3: 138462166-138462166 on Build GRCh38
rs577617034 Chr.3: 138462475-138462475 on Build GRCh38
rs761390122 Chr.3: 138462261-138462261 on Build GRCh38
rs773697324 Chr.3: 138462054-138462054 on Build GRCh38
rs73864579 Chr.3: 138462449-138462449 on Build GRCh38
rs774677139 Chr.3: 138462090-138462090 on Build GRCh38
rs753383716 Chr.3: 138462128-138462128 on Build GRCh38
rs955191795 Chr.3: 138462437-138462437 on Build GRCh38
rs771164179 Chr.3: 138462247-138462247 on Build GRCh38
rs989990110 Chr.3: 138462222-138462222 on Build GRCh38
rs36005313 Chr.3: 138462241-138462242 on Build GRCh38
rs750281464 Chr.3: 138462139-138462139 on Build GRCh38
rs941155548 Chr.3: 138462144-138462144 on Build GRCh38
rs575188463 Chr.3: 138462380-138462380 on Build GRCh38
rs746884598 Chr.3: 138462148-138462148 on Build GRCh38
rs755066353 Chr.3: 138462152-138462152 on Build GRCh38
rs749691078 Chr.3: 138462174-138462174 on Build GRCh38
rs747115250 Chr.3: 138462035-138462035 on Build GRCh38
rs75611734 Chr.3: 138462248-138462248 on Build GRCh38
rs1029265909 Chr.3: 138462398-138462398 on Build GRCh38
rs145754623 Chr.3: 138462186-138462186 on Build GRCh38
rs753834012 Chr.3: 138462094-138462097 on Build GRCh38
rs200430059 Chr.3: 138462328-138462328 on Build GRCh38
rs763200212 Chr.3: 138462062-138462062 on Build GRCh38
rs758243617 Chr.3: 138462203-138462203 on Build GRCh38
rs939431106 Chr.3: 138462299-138462299 on Build GRCh38
rs764765615 Chr.3: 138462138-138462138 on Build GRCh38
rs570915489 Chr.3: 138462241-138462241 on Build GRCh38
rs780072815 Chr.3: 138462147-138462147 on Build GRCh38
rs765127510 Chr.3: 138462181-138462181 on Build GRCh38
rs375955250 Chr.3: 138462071-138462071 on Build GRCh38
rs781310352 Chr.3: 138462153-138462153 on Build GRCh38
rs751189396 Chr.3: 138462311-138462311 on Build GRCh38
rs766037153 Chr.3: 138462205-138462205 on Build GRCh38
rs537021300 Chr.3: 138462171-138462171 on Build GRCh38
rs757035548 Chr.3: 138462137-138462137 on Build GRCh38
rs771130798 Chr.3: 138462177-138462177 on Build GRCh38
rs906553850 Chr.3: 138462284-138462284 on Build GRCh38
rs748280851 Chr.3: 138462226-138462226 on Build GRCh38
rs370771201 Chr.3: 138462304-138462304 on Build GRCh38
rs977127831 Chr.3: 138462438-138462438 on Build GRCh38
rs200708239 Chr.3: 138462032-138462032 on Build GRCh38
rs374301806 Chr.3: 138462216-138462216 on Build GRCh38
rs768683284 Chr.3: 138462043-138462043 on Build GRCh38
rs150982830 Chr.3: 138462157-138462157 on Build GRCh38
rs187584912 Chr.3: 138462252-138462252 on Build GRCh38
rs887209621 Chr.3: 138462327-138462327 on Build GRCh38
rs773254855 Chr.3: 138462170-138462170 on Build GRCh38
rs367957792 Chr.3: 138462393-138462393 on Build GRCh38
rs775008961 Chr.3: 138462181-138462181 on Build GRCh38
rs776082321 Chr.3: 138462258-138462258 on Build GRCh38
rs772938533 Chr.3: 138462273-138462273 on Build GRCh38
rs748225701 Chr.3: 138462156-138462156 on Build GRCh38
rs140959397 Chr.3: 138462189-138462189 on Build GRCh38
rs572962942 Chr.3: 138462341-138462341 on Build GRCh38
rs763638302 Chr.3: 138462124-138462124 on Build GRCh38
rs781100094 Chr.3: 138462221-138462221 on Build GRCh38
rs973752050 Chr.3: 138462381-138462381 on Build GRCh38
rs865837537 Chr.3: 138462080-138462080 on Build GRCh38
rs535707508 Chr.3: 138462347-138462347 on Build GRCh38

View Full Product Details