Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111691253-111691762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAP1A
Entrez Gene ID
5906
Gene Name
RAP1A, member of RAS oncogene family
Gene Aliases
C21KG, G-22K, KREV-1, KREV1, RAP1, SMGP21
UniGene
Hs.190334
Gene Chromosome Location
Chr.1: 111542223-111716695 on Build GRCh38
Transcripts
NM_001010935.2 NM_001291896.1 NM_002884.3 XM_017001960.1 XM_017001961.1 XM_017001962.1 XM_017001963.1 XM_017001964.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5093534 c.25C>A p.L9I Chr.1: 111691385-111691385 on Build GRCh38
6057534 c.32C>T p.S11L Chr.1: 111691392-111691392 on Build GRCh38
1686367 c.57+1G>A p.? Chr.1: 111691418-111691418 on Build GRCh38
3471270 c.30T>G p.G10G Chr.1: 111691390-111691390 on Build GRCh38
4778942 c.57G>C p.L19L Chr.1: 111691417-111691417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554680904 Chr.1: 111691448-111691448 on Build GRCh38
rs891484839 Chr.1: 111691587-111691587 on Build GRCh38
rs754387128 Chr.1: 111691350-111691350 on Build GRCh38
rs889494615 Chr.1: 111691622-111691622 on Build GRCh38
rs750254894 Chr.1: 111691451-111691451 on Build GRCh38
rs769182978 Chr.1: 111691313-111691314 on Build GRCh38
rs1007066552 Chr.1: 111691659-111691659 on Build GRCh38
rs35440055 Chr.1: 111691393-111691394 on Build GRCh38
rs371467548 Chr.1: 111691341-111691341 on Build GRCh38
rs973345897 Chr.1: 111691712-111691712 on Build GRCh38
rs1037387712 Chr.1: 111691427-111691427 on Build GRCh38
rs113859064 Chr.1: 111691331-111691331 on Build GRCh38
rs115419432 Chr.1: 111691498-111691498 on Build GRCh38
rs1052805638 Chr.1: 111691579-111691579 on Build GRCh38
rs748862864 Chr.1: 111691342-111691342 on Build GRCh38
rs375728545 Chr.1: 111691365-111691365 on Build GRCh38
rs750426286 Chr.1: 111691332-111691332 on Build GRCh38
rs758010054 Chr.1: 111691521-111691521 on Build GRCh38
rs506070 Chr.1: 111691446-111691446 on Build GRCh38
rs1017422908 Chr.1: 111691658-111691658 on Build GRCh38
rs772768735 Chr.1: 111691314-111691318 on Build GRCh38
rs762856693 Chr.1: 111691314-111691317 on Build GRCh38
rs778362438 Chr.1: 111691464-111691464 on Build GRCh38
rs74957931 Chr.1: 111691490-111691490 on Build GRCh38
rs994852090 Chr.1: 111691713-111691713 on Build GRCh38
rs907110070 Chr.1: 111691343-111691343 on Build GRCh38
rs931359315 Chr.1: 111691296-111691296 on Build GRCh38
rs541452310 Chr.1: 111691623-111691623 on Build GRCh38
rs763006476 Chr.1: 111691616-111691616 on Build GRCh38
rs527524248 Chr.1: 111691302-111691302 on Build GRCh38
rs770259165 Chr.1: 111691656-111691656 on Build GRCh38
rs980301474 Chr.1: 111691526-111691526 on Build GRCh38
rs534149452 Chr.1: 111691472-111691472 on Build GRCh38
rs766546960 Chr.1: 111691672-111691672 on Build GRCh38
rs561800944 Chr.1: 111691641-111691641 on Build GRCh38
rs1014181724 Chr.1: 111691477-111691477 on Build GRCh38
rs916658492 Chr.1: 111691517-111691517 on Build GRCh38
rs867923936 Chr.1: 111691382-111691382 on Build GRCh38
rs764258385 Chr.1: 111691437-111691437 on Build GRCh38
rs1042827462 Chr.1: 111691613-111691613 on Build GRCh38
rs1053165413 Chr.1: 111691408-111691408 on Build GRCh38
rs367963775 Chr.1: 111691337-111691337 on Build GRCh38
rs189841450 Chr.1: 111691336-111691336 on Build GRCh38
rs766346522 Chr.1: 111691458-111691458 on Build GRCh38
rs938569453 Chr.1: 111691577-111691577 on Build GRCh38
rs10489470 Chr.1: 111691593-111691593 on Build GRCh38
rs549024787 Chr.1: 111691669-111691669 on Build GRCh38
rs775259437 Chr.1: 111691689-111691689 on Build GRCh38
rs928575519 Chr.1: 111691570-111691570 on Build GRCh38
rs770003706 Chr.1: 111691558-111691558 on Build GRCh38
rs771324328 Chr.1: 111691358-111691358 on Build GRCh38
rs746166204 Chr.1: 111691421-111691421 on Build GRCh38
rs867550709 Chr.1: 111691364-111691364 on Build GRCh38
rs956739781 Chr.1: 111691537-111691537 on Build GRCh38
rs10489469 Chr.1: 111691462-111691462 on Build GRCh38
rs112667263 Chr.1: 111691419-111691419 on Build GRCh38
rs369905001 Chr.1: 111691431-111691431 on Build GRCh38
rs202011580 Chr.1: 111691311-111691311 on Build GRCh38
rs375591578 Chr.1: 111691456-111691456 on Build GRCh38
rs774094273 Chr.1: 111691340-111691341 on Build GRCh38
rs763110557 Chr.1: 111691432-111691432 on Build GRCh38
rs770899102 Chr.1: 111691318-111691321 on Build GRCh38
rs867883906 Chr.1: 111691361-111691362 on Build GRCh38
rs562995943 Chr.1: 111691556-111691556 on Build GRCh38
rs369687809 Chr.1: 111691460-111691460 on Build GRCh38
rs546487706 Chr.1: 111691523-111691523 on Build GRCh38
rs901446532 Chr.1: 111691684-111691684 on Build GRCh38
rs747793923 Chr.1: 111691354-111691354 on Build GRCh38
rs371598327 Chr.1: 111691426-111691426 on Build GRCh38
rs1009567682 Chr.1: 111691541-111691541 on Build GRCh38
rs753703252 Chr.1: 111691461-111691461 on Build GRCh38
rs537847126 Chr.1: 111691447-111691447 on Build GRCh38
rs144405448 Chr.1: 111691353-111691353 on Build GRCh38

View Full Product Details