Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111786753-111787161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCND3
Entrez Gene ID
3752
Gene Name
potassium voltage-gated channel subfamily D member 3
Gene Aliases
BRGDA9, KCND3L, KCND3S, KSHIVB, KV4.3, SCA19, SCA22
UniGene
Hs.666367
Gene Chromosome Location
Chr.1: 111770662-111989577 on Build GRCh38
Transcripts
NM_004980.4 NM_172198.2 XM_005270851.4 XM_006710629.3 XM_006710630.3 XM_006710631.3 XM_017001244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4460149 c.1154C>T p.S385F Chr.1: 111787059-111787059 on Build GRCh38
3862227 c.1218T>C p.F406F Chr.1: 111786995-111786995 on Build GRCh38
6690739 c.1224G>T p.R408R Chr.1: 111786989-111786989 on Build GRCh38
1472417 c.1242G>C p.Q414H Chr.1: 111786971-111786971 on Build GRCh38
81321 c.1209T>C p.V403V Chr.1: 111787004-111787004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3738298 Chr.1: 111786929-111786929 on Build GRCh38
rs369047161 Chr.1: 111786930-111786930 on Build GRCh38
rs768700500 Chr.1: 111786901-111786901 on Build GRCh38
rs185723423 Chr.1: 111787067-111787067 on Build GRCh38
rs533201615 Chr.1: 111787040-111787040 on Build GRCh38
rs755894090 Chr.1: 111786905-111786905 on Build GRCh38
rs761950312 Chr.1: 111787027-111787027 on Build GRCh38
rs746691414 Chr.1: 111787126-111787126 on Build GRCh38
rs779763029 Chr.1: 111786912-111786912 on Build GRCh38
rs754901454 Chr.1: 111786918-111786918 on Build GRCh38
rs1033158044 Chr.1: 111786835-111786835 on Build GRCh38
rs564816972 Chr.1: 111787016-111787016 on Build GRCh38
rs867259775 Chr.1: 111787101-111787102 on Build GRCh38
rs190089254 Chr.1: 111787103-111787103 on Build GRCh38
rs786205867 Chr.1: 111787039-111787039 on Build GRCh38
rs375562789 Chr.1: 111787052-111787052 on Build GRCh38
rs758980200 Chr.1: 111786780-111786780 on Build GRCh38
rs756921070 Chr.1: 111787131-111787131 on Build GRCh38
rs951672827 Chr.1: 111786870-111786870 on Build GRCh38
rs397515478 Chr.1: 111787044-111787044 on Build GRCh38
rs773440329 Chr.1: 111787049-111787049 on Build GRCh38
rs148428571 Chr.1: 111787082-111787082 on Build GRCh38
rs1057518007 Chr.1: 111787018-111787018 on Build GRCh38
rs778305654 Chr.1: 111786920-111786920 on Build GRCh38
rs113354737 Chr.1: 111786790-111786790 on Build GRCh38
rs774338559 Chr.1: 111786957-111786957 on Build GRCh38
rs865915853 Chr.1: 111787137-111787137 on Build GRCh38
rs760817061 Chr.1: 111787064-111787064 on Build GRCh38
rs72548726 Chr.1: 111786931-111786931 on Build GRCh38
rs753719400 Chr.1: 111786913-111786913 on Build GRCh38
rs750288762 Chr.1: 111786902-111786902 on Build GRCh38
rs764112439 Chr.1: 111786893-111786893 on Build GRCh38
rs1001321310 Chr.1: 111786821-111786821 on Build GRCh38
rs867777260 Chr.1: 111787033-111787033 on Build GRCh38
rs747464659 Chr.1: 111786822-111786822 on Build GRCh38
rs1057519453 Chr.1: 111787060-111787060 on Build GRCh38
rs397515477 Chr.1: 111787094-111787094 on Build GRCh38
rs941625672 Chr.1: 111786903-111786903 on Build GRCh38
rs958270299 Chr.1: 111786868-111786868 on Build GRCh38
rs373174767 Chr.1: 111787113-111787113 on Build GRCh38
rs180854655 Chr.1: 111786921-111786921 on Build GRCh38
rs751611455 Chr.1: 111786900-111786900 on Build GRCh38
rs886248434 Chr.1: 111787117-111787117 on Build GRCh38
rs372005674 Chr.1: 111786926-111786926 on Build GRCh38

View Full Product Details