Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113886858-113887403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4B1-AS1
Entrez Gene ID
100287722
Gene Name
AP4B1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.664669
Gene Chromosome Location
Chr.1: 113812612-113901237 on Build GRCh38
Transcripts
NR_037864.1 NR_125965.1 NM_001010922.2

Gene Symbol
BCL2L15
Entrez Gene ID
440603
Gene Name
BCL2 like 15
Gene Aliases
Bfk, C1orf178
UniGene
Hs.123106
Gene Chromosome Location
Chr.1: 113876814-113887547 on Build GRCh38
Transcripts
NR_037864.1 NR_125965.1 NM_001010922.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894088 c.32C>T p.T11M Chr.1: 113887344-113887344 on Build GRCh38
159352 c.121G>C p.D41H Chr.1: 113887255-113887255 on Build GRCh38
894087 c.66C>A p.F22L Chr.1: 113887310-113887310 on Build GRCh38
4020798 c.52C>T p.L18L Chr.1: 113887324-113887324 on Build GRCh38
5729956 c.99C>T p.N33N Chr.1: 113887277-113887277 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1046845874 Chr.1: 113887138-113887138 on Build GRCh38
rs772437113 Chr.1: 113886984-113886984 on Build GRCh38
rs758960282 Chr.1: 113887204-113887205 on Build GRCh38
rs759047386 Chr.1: 113887363-113887363 on Build GRCh38
rs780027706 Chr.1: 113887206-113887206 on Build GRCh38
rs774300907 Chr.1: 113887339-113887339 on Build GRCh38
rs889927996 Chr.1: 113886969-113886969 on Build GRCh38
rs879807094 Chr.1: 113887132-113887133 on Build GRCh38
rs144140962 Chr.1: 113887298-113887298 on Build GRCh38
rs768575243 Chr.1: 113887217-113887217 on Build GRCh38
rs534791954 Chr.1: 113887045-113887045 on Build GRCh38
rs757880844 Chr.1: 113887293-113887293 on Build GRCh38
rs752305101 Chr.1: 113887366-113887366 on Build GRCh38
rs958267516 Chr.1: 113886966-113886966 on Build GRCh38
rs879329111 Chr.1: 113887145-113887145 on Build GRCh38
rs972554756 Chr.1: 113887061-113887061 on Build GRCh38
rs974963099 Chr.1: 113886902-113886902 on Build GRCh38
rs959966066 Chr.1: 113887059-113887059 on Build GRCh38
rs765988230 Chr.1: 113887347-113887347 on Build GRCh38
rs986398980 Chr.1: 113887299-113887299 on Build GRCh38
rs939777721 Chr.1: 113886956-113886956 on Build GRCh38
rs535762318 Chr.1: 113886983-113886983 on Build GRCh38
rs375501688 Chr.1: 113887332-113887332 on Build GRCh38
rs934242023 Chr.1: 113887203-113887203 on Build GRCh38
rs375707613 Chr.1: 113887024-113887024 on Build GRCh38
rs565354388 Chr.1: 113886925-113886925 on Build GRCh38
rs1017000011 Chr.1: 113886991-113886991 on Build GRCh38
rs996330051 Chr.1: 113887250-113887250 on Build GRCh38
rs746295009 Chr.1: 113887296-113887296 on Build GRCh38
rs889980536 Chr.1: 113887261-113887261 on Build GRCh38
rs753383105 Chr.1: 113887273-113887273 on Build GRCh38
rs895088209 Chr.1: 113887134-113887134 on Build GRCh38
rs570301071 Chr.1: 113886953-113886953 on Build GRCh38
rs776161431 Chr.1: 113887348-113887348 on Build GRCh38
rs572310900 Chr.1: 113887015-113887015 on Build GRCh38
rs199687942 Chr.1: 113887343-113887343 on Build GRCh38
rs188979219 Chr.1: 113887110-113887110 on Build GRCh38
rs992676214 Chr.1: 113887096-113887096 on Build GRCh38
rs757827844 Chr.1: 113887367-113887367 on Build GRCh38
rs550186184 Chr.1: 113886944-113886944 on Build GRCh38
rs780147699 Chr.1: 113887300-113887300 on Build GRCh38
rs200731954 Chr.1: 113887342-113887342 on Build GRCh38
rs761542846 Chr.1: 113887228-113887228 on Build GRCh38
rs563551703 Chr.1: 113887111-113887111 on Build GRCh38
rs1049937665 Chr.1: 113886955-113886955 on Build GRCh38
rs994174144 Chr.1: 113887036-113887036 on Build GRCh38
rs953698784 Chr.1: 113886926-113886926 on Build GRCh38
rs551178343 Chr.1: 113887291-113887291 on Build GRCh38
rs879705209 Chr.1: 113887149-113887149 on Build GRCh38
rs775769245 Chr.1: 113887009-113887009 on Build GRCh38
rs528339863 Chr.1: 113887282-113887282 on Build GRCh38
rs1000019424 Chr.1: 113887188-113887188 on Build GRCh38
rs750255772 Chr.1: 113887254-113887254 on Build GRCh38
rs112780009 Chr.1: 113887189-113887189 on Build GRCh38
rs767327056 Chr.1: 113887229-113887229 on Build GRCh38
rs368423811 Chr.1: 113887281-113887281 on Build GRCh38
rs142883498 Chr.1: 113886907-113886907 on Build GRCh38
rs760296097 Chr.1: 113887259-113887259 on Build GRCh38
rs988494744 Chr.1: 113887192-113887192 on Build GRCh38
rs941534183 Chr.1: 113887290-113887290 on Build GRCh38
rs2358995 Chr.1: 113886893-113886893 on Build GRCh38
rs78095589 Chr.1: 113887364-113887364 on Build GRCh38
rs185505258 Chr.1: 113887310-113887310 on Build GRCh38
rs551503672 Chr.1: 113886918-113886918 on Build GRCh38
rs558274069 Chr.1: 113887100-113887100 on Build GRCh38
rs573945024 Chr.1: 113887135-113887135 on Build GRCh38
rs1022835495 Chr.1: 113887376-113887376 on Build GRCh38
rs767090830 Chr.1: 113887359-113887359 on Build GRCh38
rs934189501 Chr.1: 113886927-113886927 on Build GRCh38
rs202039250 Chr.1: 113887335-113887335 on Build GRCh38
rs538548057 Chr.1: 113887194-113887194 on Build GRCh38
rs542951887 Chr.1: 113887174-113887174 on Build GRCh38
rs528152624 Chr.1: 113886909-113886909 on Build GRCh38
rs184963665 Chr.1: 113887103-113887103 on Build GRCh38
rs146778119 Chr.1: 113887264-113887264 on Build GRCh38
rs761350107 Chr.1: 113887344-113887344 on Build GRCh38
rs1055036449 Chr.1: 113887113-113887113 on Build GRCh38
rs373828730 Chr.1: 113887008-113887008 on Build GRCh38
rs961111328 Chr.1: 113887312-113887312 on Build GRCh38
rs902563821 Chr.1: 113887139-113887139 on Build GRCh38
rs372411327 Chr.1: 113886936-113886936 on Build GRCh38
rs774173187 Chr.1: 113887224-113887224 on Build GRCh38
rs879942213 Chr.1: 113887141-113887141 on Build GRCh38
rs781674664 Chr.1: 113887294-113887294 on Build GRCh38

View Full Product Details