Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142157437-142157937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GK5
Entrez Gene ID
256356
Gene Name
glycerol kinase 5 (putative)
Gene Aliases
-
UniGene
Hs.135904
Gene Chromosome Location
Chr.3: 142157527-142225607 on Build GRCh38
Transcripts
NM_001039547.2 NR_033289.1 XM_011512624.2 XM_017006089.1 XM_017006090.1

SNP Detail

SNP ID Chromosome Location
rs1046281435 Chr.3: 142157495-142157495 on Build GRCh38
rs950332477 Chr.3: 142157491-142157491 on Build GRCh38
rs950339230 Chr.3: 142157806-142157806 on Build GRCh38
rs757337565 Chr.3: 142157788-142157788 on Build GRCh38
rs1038593532 Chr.3: 142157557-142157557 on Build GRCh38
rs949841229 Chr.3: 142157521-142157521 on Build GRCh38
rs1018113391 Chr.3: 142157690-142157690 on Build GRCh38
rs963913659 Chr.3: 142157704-142157704 on Build GRCh38
rs1032902147 Chr.3: 142157693-142157693 on Build GRCh38
rs936994368 Chr.3: 142157902-142157902 on Build GRCh38
rs867070962 Chr.3: 142157874-142157874 on Build GRCh38
rs565603761 Chr.3: 142157531-142157531 on Build GRCh38
rs927061493 Chr.3: 142157899-142157899 on Build GRCh38
rs1038472074 Chr.3: 142157910-142157910 on Build GRCh38
rs891214088 Chr.3: 142157580-142157580 on Build GRCh38
rs979861998 Chr.3: 142157663-142157663 on Build GRCh38
rs76244822 Chr.3: 142157515-142157515 on Build GRCh38
rs751628599 Chr.3: 142157550-142157550 on Build GRCh38
rs1025839716 Chr.3: 142157564-142157564 on Build GRCh38
rs983817765 Chr.3: 142157785-142157785 on Build GRCh38
rs532837433 Chr.3: 142157534-142157534 on Build GRCh38
rs369620736 Chr.3: 142157786-142157786 on Build GRCh38
rs550966744 Chr.3: 142157553-142157553 on Build GRCh38
rs573516834 Chr.3: 142157498-142157498 on Build GRCh38
rs530217904 Chr.3: 142157641-142157641 on Build GRCh38
rs913099632 Chr.3: 142157814-142157814 on Build GRCh38
rs1010290369 Chr.3: 142157552-142157552 on Build GRCh38
rs957547322 Chr.3: 142157764-142157764 on Build GRCh38
rs949893452 Chr.3: 142157841-142157841 on Build GRCh38
rs542330735 Chr.3: 142157728-142157731 on Build GRCh38
rs920648728 Chr.3: 142157729-142157729 on Build GRCh38
rs1018380502 Chr.3: 142157640-142157640 on Build GRCh38
rs548355086 Chr.3: 142157787-142157787 on Build GRCh38
rs1045363881 Chr.3: 142157529-142157529 on Build GRCh38
rs898756493 Chr.3: 142157558-142157558 on Build GRCh38
rs562713702 Chr.3: 142157619-142157619 on Build GRCh38
rs1008351073 Chr.3: 142157603-142157603 on Build GRCh38
rs994334090 Chr.3: 142157559-142157559 on Build GRCh38
rs981115111 Chr.3: 142157851-142157851 on Build GRCh38
rs908095552 Chr.3: 142157475-142157475 on Build GRCh38

View Full Product Details