Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 157381243-157381777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VEPH1
Entrez Gene ID
79674
Gene Name
ventricular zone expressed PH domain containing 1
Gene Aliases
MELT, VEPH
UniGene
Hs.658046
Gene Chromosome Location
Chr.3: 157259742-157503626 on Build GRCh38
Transcripts
NM_001167911.1 NM_001167912.1 NM_024621.2 XM_011513134.2 XM_011513135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5557152 c.991A>T p.I331F Chr.3: 157381292-157381292 on Build GRCh38
4514621 c.981C>T p.L327L Chr.3: 157381302-157381302 on Build GRCh38
6367363 c.953A>G p.N318S Chr.3: 157381330-157381330 on Build GRCh38
5653478 c.1020A>G p.S340S Chr.3: 157381263-157381263 on Build GRCh38
3005437 c.957G>A p.M319I Chr.3: 157381326-157381326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769601724 Chr.3: 157381322-157381322 on Build GRCh38
rs765357397 Chr.3: 157381382-157381382 on Build GRCh38
rs1037937249 Chr.3: 157381419-157381419 on Build GRCh38
rs764707629 Chr.3: 157381286-157381286 on Build GRCh38
rs1055029109 Chr.3: 157381713-157381713 on Build GRCh38
rs987128109 Chr.3: 157381630-157381630 on Build GRCh38
rs943082417 Chr.3: 157381648-157381648 on Build GRCh38
rs761229597 Chr.3: 157381284-157381284 on Build GRCh38
rs750545485 Chr.3: 157381384-157381384 on Build GRCh38
rs745509201 Chr.3: 157381350-157381350 on Build GRCh38
rs773791055 Chr.3: 157381341-157381341 on Build GRCh38
rs531973587 Chr.3: 157381753-157381753 on Build GRCh38
rs780936966 Chr.3: 157381318-157381318 on Build GRCh38
rs768386256 Chr.3: 157381273-157381273 on Build GRCh38
rs758689370 Chr.3: 157381317-157381317 on Build GRCh38
rs770954099 Chr.3: 157381502-157381502 on Build GRCh38
rs1022870518 Chr.3: 157381521-157381521 on Build GRCh38
rs978770594 Chr.3: 157381551-157381551 on Build GRCh38
rs773365211 Chr.3: 157381354-157381354 on Build GRCh38
rs757394631 Chr.3: 157381270-157381270 on Build GRCh38
rs993834905 Chr.3: 157381447-157381447 on Build GRCh38
rs768204493 Chr.3: 157381264-157381266 on Build GRCh38
rs759571268 Chr.3: 157381650-157381650 on Build GRCh38
rs770415901 Chr.3: 157381338-157381338 on Build GRCh38
rs543269498 Chr.3: 157381510-157381510 on Build GRCh38
rs79014676 Chr.3: 157381721-157381721 on Build GRCh38
rs371547756 Chr.3: 157381424-157381424 on Build GRCh38
rs199627611 Chr.3: 157381301-157381301 on Build GRCh38
rs906451534 Chr.3: 157381674-157381674 on Build GRCh38
rs12492929 Chr.3: 157381673-157381673 on Build GRCh38
rs1026206475 Chr.3: 157381456-157381456 on Build GRCh38
rs914831777 Chr.3: 157381626-157381626 on Build GRCh38
rs781024966 Chr.3: 157381276-157381276 on Build GRCh38
rs754319869 Chr.3: 157381292-157381292 on Build GRCh38
rs188430236 Chr.3: 157381575-157381575 on Build GRCh38
rs966877284 Chr.3: 157381548-157381548 on Build GRCh38
rs1036180339 Chr.3: 157381749-157381749 on Build GRCh38
rs922841110 Chr.3: 157381576-157381576 on Build GRCh38
rs9871100 Chr.3: 157381669-157381669 on Build GRCh38
rs561547891 Chr.3: 157381591-157381591 on Build GRCh38
rs761202849 Chr.3: 157381276-157381277 on Build GRCh38
rs773985894 Chr.3: 157381280-157381280 on Build GRCh38
rs778876064 Chr.3: 157381311-157381311 on Build GRCh38
rs1006183569 Chr.3: 157381721-157381721 on Build GRCh38
rs775971967 Chr.3: 157381356-157381356 on Build GRCh38
rs750649678 Chr.3: 157381312-157381312 on Build GRCh38
rs377372657 Chr.3: 157381325-157381325 on Build GRCh38
rs147644993 Chr.3: 157381277-157381277 on Build GRCh38
rs572671284 Chr.3: 157381454-157381454 on Build GRCh38
rs1003463911 Chr.3: 157381481-157381481 on Build GRCh38
rs779143424 Chr.3: 157381271-157381271 on Build GRCh38
rs770380500 Chr.3: 157381279-157381279 on Build GRCh38
rs372781207 Chr.3: 157381601-157381601 on Build GRCh38
rs887687706 Chr.3: 157381716-157381716 on Build GRCh38
rs906545010 Chr.3: 157381475-157381475 on Build GRCh38
rs1003537912 Chr.3: 157381708-157381708 on Build GRCh38
rs977556881 Chr.3: 157381645-157381645 on Build GRCh38
rs932982557 Chr.3: 157381611-157381611 on Build GRCh38
rs1014801862 Chr.3: 157381530-157381530 on Build GRCh38
rs759276325 Chr.3: 157381282-157381282 on Build GRCh38
rs1037569028 Chr.3: 157381654-157381654 on Build GRCh38
rs761070103 Chr.3: 157381365-157381365 on Build GRCh38
rs989628787 Chr.3: 157381516-157381516 on Build GRCh38
rs34823544 Chr.3: 157381298-157381298 on Build GRCh38
rs777141267 Chr.3: 157381374-157381374 on Build GRCh38
rs943218277 Chr.3: 157381412-157381412 on Build GRCh38
rs961790216 Chr.3: 157381545-157381545 on Build GRCh38
rs929115443 Chr.3: 157381671-157381671 on Build GRCh38
rs11923380 Chr.3: 157381328-157381328 on Build GRCh38
rs116855258 Chr.3: 157381320-157381320 on Build GRCh38
rs776125482 Chr.3: 157381270-157381271 on Build GRCh38
rs762289716 Chr.3: 157381380-157381380 on Build GRCh38
rs948748803 Chr.3: 157381712-157381712 on Build GRCh38
rs753177790 Chr.3: 157381283-157381283 on Build GRCh38
rs915872633 Chr.3: 157381679-157381679 on Build GRCh38
rs560421888 Chr.3: 157381370-157381370 on Build GRCh38
rs746086109 Chr.3: 157381272-157381272 on Build GRCh38
rs749404991 Chr.3: 157381432-157381432 on Build GRCh38
rs894505360 Chr.3: 157381349-157381349 on Build GRCh38
rs758597656 Chr.3: 157381389-157381389 on Build GRCh38
rs781661788 Chr.3: 157381278-157381278 on Build GRCh38
rs924286889 Chr.3: 157381309-157381309 on Build GRCh38
rs143118936 Chr.3: 157381641-157381641 on Build GRCh38

View Full Product Details