Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186619776-186620268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSG
Entrez Gene ID
197
Gene Name
alpha 2-HS glycoprotein
Gene Aliases
A2HS, AHS, FETUA, HSGA
UniGene
Hs.324746
Gene Chromosome Location
Chr.3: 186612928-186621318 on Build GRCh38
Transcripts
NM_001622.2 XM_017005840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5615245 c.728T>A p.V243D Chr.3: 186619909-186619909 on Build GRCh38
1308923 c.750C>G p.F250L Chr.3: 186619931-186619931 on Build GRCh38
4002545 c.743T>C p.M248T Chr.3: 186619924-186619924 on Build GRCh38
6597078 c.727_728insT p.A244fs*23 Chr.3: 186619908-186619909 on Build GRCh38
4660628 c.676-1G>T p.? Chr.3: 186619856-186619856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755020851 Chr.3: 186619989-186619989 on Build GRCh38
rs371986917 Chr.3: 186619952-186619952 on Build GRCh38
rs556876120 Chr.3: 186620133-186620133 on Build GRCh38
rs773723863 Chr.3: 186620233-186620233 on Build GRCh38
rs111891162 Chr.3: 186620081-186620081 on Build GRCh38
rs1021740928 Chr.3: 186620164-186620164 on Build GRCh38
rs199746616 Chr.3: 186619980-186619980 on Build GRCh38
rs772342633 Chr.3: 186620130-186620130 on Build GRCh38
rs553019044 Chr.3: 186620092-186620092 on Build GRCh38
rs956371288 Chr.3: 186619804-186619804 on Build GRCh38
rs751252833 Chr.3: 186619825-186619825 on Build GRCh38
rs4917 Chr.3: 186619924-186619924 on Build GRCh38
rs1049461930 Chr.3: 186620080-186620080 on Build GRCh38
rs751721045 Chr.3: 186619982-186619982 on Build GRCh38
rs996320342 Chr.3: 186620071-186620071 on Build GRCh38
rs548479632 Chr.3: 186620010-186620010 on Build GRCh38
rs753869839 Chr.3: 186619972-186619972 on Build GRCh38
rs932351912 Chr.3: 186620075-186620075 on Build GRCh38
rs1053998057 Chr.3: 186620021-186620021 on Build GRCh38
rs184392275 Chr.3: 186620082-186620082 on Build GRCh38
rs761184894 Chr.3: 186619986-186619987 on Build GRCh38
rs1032312931 Chr.3: 186620086-186620086 on Build GRCh38
rs202020744 Chr.3: 186619841-186619841 on Build GRCh38
rs777587342 Chr.3: 186619863-186619863 on Build GRCh38
rs917785676 Chr.3: 186620197-186620197 on Build GRCh38
rs773644727 Chr.3: 186619809-186619809 on Build GRCh38
rs541740551 Chr.3: 186619813-186619813 on Build GRCh38
rs200581915 Chr.3: 186619960-186619960 on Build GRCh38
rs146838610 Chr.3: 186620091-186620091 on Build GRCh38
rs776837665 Chr.3: 186619807-186619807 on Build GRCh38
rs772446285 Chr.3: 186619865-186619865 on Build GRCh38
rs923966652 Chr.3: 186619964-186619964 on Build GRCh38
rs1022105841 Chr.3: 186619828-186619828 on Build GRCh38
rs149819140 Chr.3: 186619957-186619957 on Build GRCh38
rs961066390 Chr.3: 186620104-186620104 on Build GRCh38
rs752462963 Chr.3: 186619845-186619845 on Build GRCh38
rs762188966 Chr.3: 186619808-186619808 on Build GRCh38
rs759043456 Chr.3: 186619945-186619945 on Build GRCh38
rs755179125 Chr.3: 186619893-186619893 on Build GRCh38
rs369319525 Chr.3: 186619969-186619969 on Build GRCh38
rs368520628 Chr.3: 186619872-186619872 on Build GRCh38
rs747284297 Chr.3: 186619879-186619879 on Build GRCh38
rs139766517 Chr.3: 186620122-186620122 on Build GRCh38
rs749520427 Chr.3: 186619927-186619927 on Build GRCh38
rs753816561 Chr.3: 186619869-186619869 on Build GRCh38
rs748467513 Chr.3: 186619913-186619913 on Build GRCh38
rs562630713 Chr.3: 186620124-186620124 on Build GRCh38
rs775935712 Chr.3: 186619962-186619962 on Build GRCh38
rs570354531 Chr.3: 186620036-186620036 on Build GRCh38
rs991071602 Chr.3: 186620191-186620191 on Build GRCh38
rs767145807 Chr.3: 186619836-186619836 on Build GRCh38
rs113984135 Chr.3: 186619854-186619854 on Build GRCh38
rs759161765 Chr.3: 186619829-186619829 on Build GRCh38
rs770477954 Chr.3: 186619925-186619925 on Build GRCh38
rs767078835 Chr.3: 186619951-186619951 on Build GRCh38
rs765868370 Chr.3: 186619819-186619819 on Build GRCh38
rs574877295 Chr.3: 186620222-186620222 on Build GRCh38
rs372945050 Chr.3: 186620131-186620131 on Build GRCh38
rs780254597 Chr.3: 186619876-186619876 on Build GRCh38
rs201550870 Chr.3: 186619899-186619899 on Build GRCh38
rs770178913 Chr.3: 186619922-186619922 on Build GRCh38
rs773773652 Chr.3: 186619923-186619923 on Build GRCh38
rs528205795 Chr.3: 186619928-186619928 on Build GRCh38
rs2518136 Chr.3: 186620038-186620038 on Build GRCh38

View Full Product Details