Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 191218659-191219154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSTN
Entrez Gene ID
344901
Gene Name
osteocrin
Gene Aliases
MUSCLIN
UniGene
Hs.526794
Gene Chromosome Location
Chr.3: 191199060-191265601 on Build GRCh38
Transcripts
NM_198184.1 XM_005247428.4 XM_017006303.1 NR_133663.1

Gene Symbol
OSTN-AS1
Entrez Gene ID
106480738
Gene Name
OSTN antisense RNA 1
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.3: 191213291-191234605 on Build GRCh38
Transcripts
NM_198184.1 XM_005247428.4 XM_017006303.1 NR_133663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3333355 c.253T>C p.S85P Chr.3: 191218897-191218897 on Build GRCh38
317773 c.282C>T p.L94L Chr.3: 191218926-191218926 on Build GRCh38
6273613 c.124G>T p.D42Y Chr.3: 191218768-191218768 on Build GRCh38
276564 c.275A>G p.D92G Chr.3: 191218919-191218919 on Build GRCh38
6272119 c.231G>T p.E77D Chr.3: 191218875-191218875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34687554 Chr.3: 191218788-191218788 on Build GRCh38
rs533510465 Chr.3: 191219026-191219026 on Build GRCh38
rs778562671 Chr.3: 191218748-191218748 on Build GRCh38
rs761722305 Chr.3: 191218800-191218800 on Build GRCh38
rs781309287 Chr.3: 191218897-191218897 on Build GRCh38
rs932591911 Chr.3: 191218907-191218907 on Build GRCh38
rs547008935 Chr.3: 191219102-191219102 on Build GRCh38
rs957502750 Chr.3: 191218781-191218781 on Build GRCh38
rs764548243 Chr.3: 191218927-191218927 on Build GRCh38
rs560545017 Chr.3: 191218828-191218828 on Build GRCh38
rs1027831831 Chr.3: 191219117-191219117 on Build GRCh38
rs998894673 Chr.3: 191219129-191219129 on Build GRCh38
rs751040629 Chr.3: 191218968-191218968 on Build GRCh38
rs184878937 Chr.3: 191218727-191218727 on Build GRCh38
rs199689299 Chr.3: 191218963-191218963 on Build GRCh38
rs760770385 Chr.3: 191218922-191218922 on Build GRCh38
rs769920798 Chr.3: 191219013-191219013 on Build GRCh38
rs144554844 Chr.3: 191218812-191218812 on Build GRCh38
rs1032082725 Chr.3: 191219130-191219130 on Build GRCh38
rs756850110 Chr.3: 191218744-191218744 on Build GRCh38
rs1006216902 Chr.3: 191219003-191219003 on Build GRCh38
rs948474207 Chr.3: 191218827-191218827 on Build GRCh38
rs549582746 Chr.3: 191218852-191218852 on Build GRCh38
rs138575978 Chr.3: 191218768-191218768 on Build GRCh38
rs200624720 Chr.3: 191218967-191218967 on Build GRCh38
rs768717375 Chr.3: 191218779-191218779 on Build GRCh38
rs748085535 Chr.3: 191218982-191218982 on Build GRCh38
rs375405607 Chr.3: 191218919-191218919 on Build GRCh38
rs757583938 Chr.3: 191218866-191218866 on Build GRCh38
rs887402498 Chr.3: 191218933-191218933 on Build GRCh38
rs1045422679 Chr.3: 191218879-191218881 on Build GRCh38
rs994884079 Chr.3: 191219103-191219103 on Build GRCh38
rs756391990 Chr.3: 191218683-191218683 on Build GRCh38
rs758051030 Chr.3: 191218871-191218871 on Build GRCh38
rs899967804 Chr.3: 191219081-191219081 on Build GRCh38
rs779294960 Chr.3: 191218867-191218867 on Build GRCh38
rs148417658 Chr.3: 191218899-191218899 on Build GRCh38
rs897909335 Chr.3: 191219082-191219082 on Build GRCh38
rs555038682 Chr.3: 191218819-191218819 on Build GRCh38
rs924574166 Chr.3: 191218752-191218752 on Build GRCh38
rs375573178 Chr.3: 191218714-191218714 on Build GRCh38
rs749659244 Chr.3: 191218730-191218730 on Build GRCh38
rs771584947 Chr.3: 191218766-191218766 on Build GRCh38
rs769690649 Chr.3: 191218901-191218901 on Build GRCh38
rs1038552256 Chr.3: 191219057-191219057 on Build GRCh38
rs765579699 Chr.3: 191218953-191218953 on Build GRCh38
rs759801670 Chr.3: 191218915-191218915 on Build GRCh38
rs866474828 Chr.3: 191218736-191218736 on Build GRCh38
rs928314962 Chr.3: 191218893-191218893 on Build GRCh38
rs756580144 Chr.3: 191218830-191218830 on Build GRCh38
rs749879736 Chr.3: 191218759-191218759 on Build GRCh38
rs780898720 Chr.3: 191218972-191218972 on Build GRCh38
rs1036477873 Chr.3: 191219062-191219062 on Build GRCh38
rs9290979 Chr.3: 191218693-191218693 on Build GRCh38
rs751640938 Chr.3: 191218713-191218713 on Build GRCh38
rs556347841 Chr.3: 191218721-191218721 on Build GRCh38
rs766721385 Chr.3: 191218712-191218712 on Build GRCh38
rs779548793 Chr.3: 191218877-191218877 on Build GRCh38
rs531909625 Chr.3: 191218930-191218930 on Build GRCh38
rs202216550 Chr.3: 191218722-191218722 on Build GRCh38
rs147289386 Chr.3: 191218913-191218913 on Build GRCh38
rs181134884 Chr.3: 191218849-191218849 on Build GRCh38
rs772940268 Chr.3: 191218905-191218905 on Build GRCh38
rs373617423 Chr.3: 191218776-191218776 on Build GRCh38
rs868419514 Chr.3: 191218945-191218945 on Build GRCh38
rs145218293 Chr.3: 191218961-191218961 on Build GRCh38
rs759726596 Chr.3: 191218815-191218815 on Build GRCh38
rs746513224 Chr.3: 191218883-191218883 on Build GRCh38
rs769575990 Chr.3: 191218776-191218776 on Build GRCh38
rs754318603 Chr.3: 191218928-191218928 on Build GRCh38
rs778109141 Chr.3: 191218728-191218728 on Build GRCh38
rs1029729938 Chr.3: 191219101-191219101 on Build GRCh38
rs563138982 Chr.3: 191218868-191218868 on Build GRCh38
rs183867158 Chr.3: 191218970-191218970 on Build GRCh38
rs766668093 Chr.3: 191218802-191218802 on Build GRCh38
rs768666020 Chr.3: 191218895-191218895 on Build GRCh38

View Full Product Details