Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 4240189-4240692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM128
Entrez Gene ID
85013
Gene Name
transmembrane protein 128
Gene Aliases
-
UniGene
Hs.12845
Gene Chromosome Location
Chr.4: 4235542-4248265 on Build GRCh38
Transcripts
NM_001297551.1 NM_001297552.1 NM_032927.3 XM_005248034.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4443482 c.244T>C p.Y82H Chr.4: 4240403-4240403 on Build GRCh38
5931560 c.305C>T p.S102F Chr.4: 4240342-4240342 on Build GRCh38
5978808 c.241T>C p.W81R Chr.4: 4240406-4240406 on Build GRCh38
481277 c.258A>G p.G86G Chr.4: 4240389-4240389 on Build GRCh38
6239911 c.255C>A p.G85G Chr.4: 4240464-4240464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs62286916 Chr.4: 4240637-4240637 on Build GRCh38
rs762954025 Chr.4: 4240356-4240356 on Build GRCh38
rs1023354238 Chr.4: 4240344-4240344 on Build GRCh38
rs1033370438 Chr.4: 4240461-4240461 on Build GRCh38
rs746215375 Chr.4: 4240531-4240532 on Build GRCh38
rs373749767 Chr.4: 4240283-4240283 on Build GRCh38
rs796268704 Chr.4: 4240253-4240253 on Build GRCh38
rs183662146 Chr.4: 4240596-4240596 on Build GRCh38
rs980642548 Chr.4: 4240657-4240657 on Build GRCh38
rs756631858 Chr.4: 4240280-4240280 on Build GRCh38
rs769711689 Chr.4: 4240519-4240519 on Build GRCh38
rs956816937 Chr.4: 4240379-4240379 on Build GRCh38
rs139174323 Chr.4: 4240269-4240269 on Build GRCh38
rs750827231 Chr.4: 4240404-4240404 on Build GRCh38
rs897395350 Chr.4: 4240233-4240233 on Build GRCh38
rs770871960 Chr.4: 4240298-4240298 on Build GRCh38
rs201453309 Chr.4: 4240405-4240405 on Build GRCh38
rs752195763 Chr.4: 4240435-4240435 on Build GRCh38
rs368502996 Chr.4: 4240497-4240498 on Build GRCh38
rs956579889 Chr.4: 4240486-4240486 on Build GRCh38
rs769683962 Chr.4: 4240350-4240350 on Build GRCh38
rs747310088 Chr.4: 4240255-4240255 on Build GRCh38
rs757875461 Chr.4: 4240436-4240436 on Build GRCh38
rs187417277 Chr.4: 4240370-4240370 on Build GRCh38
rs759483260 Chr.4: 4240343-4240343 on Build GRCh38
rs777594245 Chr.4: 4240531-4240531 on Build GRCh38
rs778457193 Chr.4: 4240281-4240281 on Build GRCh38
rs777570065 Chr.4: 4240293-4240293 on Build GRCh38
rs768748517 Chr.4: 4240526-4240526 on Build GRCh38
rs748557720 Chr.4: 4240295-4240295 on Build GRCh38
rs757010540 Chr.4: 4240480-4240480 on Build GRCh38
rs915091139 Chr.4: 4240406-4240406 on Build GRCh38
rs943999171 Chr.4: 4240225-4240225 on Build GRCh38
rs1021912165 Chr.4: 4240542-4240542 on Build GRCh38
rs780019366 Chr.4: 4240279-4240280 on Build GRCh38
rs932117496 Chr.4: 4240382-4240383 on Build GRCh38
rs1048646390 Chr.4: 4240657-4240658 on Build GRCh38
rs994364489 Chr.4: 4240235-4240235 on Build GRCh38
rs771744508 Chr.4: 4240286-4240286 on Build GRCh38
rs751490924 Chr.4: 4240322-4240322 on Build GRCh38
rs370168661 Chr.4: 4240521-4240521 on Build GRCh38
rs74484663 Chr.4: 4240279-4240279 on Build GRCh38
rs144061281 Chr.4: 4240401-4240401 on Build GRCh38
rs375812180 Chr.4: 4240391-4240391 on Build GRCh38
rs781125457 Chr.4: 4240489-4240489 on Build GRCh38
rs145069843 Chr.4: 4240510-4240510 on Build GRCh38
rs373499108 Chr.4: 4240437-4240437 on Build GRCh38
rs749350757 Chr.4: 4240525-4240525 on Build GRCh38
rs776617288 Chr.4: 4240342-4240342 on Build GRCh38
rs200910248 Chr.4: 4240269-4240270 on Build GRCh38
rs780620933 Chr.4: 4240404-4240404 on Build GRCh38
rs567729155 Chr.4: 4240531-4240531 on Build GRCh38
rs190648276 Chr.4: 4240534-4240534 on Build GRCh38
rs762119908 Chr.4: 4240400-4240400 on Build GRCh38
rs146482952 Chr.4: 4240634-4240634 on Build GRCh38
rs375611658 Chr.4: 4240507-4240507 on Build GRCh38
rs370219053 Chr.4: 4240479-4240479 on Build GRCh38
rs754503165 Chr.4: 4240362-4240363 on Build GRCh38
rs571035872 Chr.4: 4240613-4240613 on Build GRCh38
rs778056715 Chr.4: 4240542-4240543 on Build GRCh38
rs927814329 Chr.4: 4240664-4240664 on Build GRCh38
rs562285701 Chr.4: 4240355-4240355 on Build GRCh38
rs3817719 Chr.4: 4240226-4240226 on Build GRCh38
rs750178033 Chr.4: 4240270-4240270 on Build GRCh38
rs751860577 Chr.4: 4240275-4240275 on Build GRCh38
rs774590273 Chr.4: 4240374-4240374 on Build GRCh38
rs200562278 Chr.4: 4240294-4240294 on Build GRCh38
rs111900550 Chr.4: 4240338-4240338 on Build GRCh38
rs746636568 Chr.4: 4240464-4240464 on Build GRCh38
rs757626874 Chr.4: 4240278-4240278 on Build GRCh38
rs747672700 Chr.4: 4240469-4240470 on Build GRCh38

View Full Product Details