Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115642476-115643011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VANGL1
Entrez Gene ID
81839
Gene Name
VANGL planar cell polarity protein 1
Gene Aliases
KITENIN, LPP2, STB2, STBM2
UniGene
Hs.515130
Gene Chromosome Location
Chr.1: 115641953-115698224 on Build GRCh38
Transcripts
NM_001172411.1 NM_001172412.1 NM_138959.2

SNP Detail

SNP ID Chromosome Location
rs967286840 Chr.1: 115642940-115642940 on Build GRCh38
rs956709771 Chr.1: 115642918-115642918 on Build GRCh38
rs3010378 Chr.1: 115642983-115642983 on Build GRCh38
rs1029572753 Chr.1: 115642515-115642515 on Build GRCh38
rs943609766 Chr.1: 115642849-115642850 on Build GRCh38
rs763127427 Chr.1: 115642802-115642802 on Build GRCh38
rs1033710579 Chr.1: 115642879-115642879 on Build GRCh38
rs894278241 Chr.1: 115642557-115642557 on Build GRCh38
rs1021092718 Chr.1: 115642925-115642925 on Build GRCh38
rs930316301 Chr.1: 115642750-115642750 on Build GRCh38
rs766334582 Chr.1: 115642568-115642568 on Build GRCh38
rs528070290 Chr.1: 115642730-115642730 on Build GRCh38
rs1032020599 Chr.1: 115642891-115642892 on Build GRCh38
rs576863234 Chr.1: 115642593-115642593 on Build GRCh38
rs899627914 Chr.1: 115642504-115642504 on Build GRCh38
rs935668405 Chr.1: 115642683-115642683 on Build GRCh38
rs186489559 Chr.1: 115642617-115642617 on Build GRCh38
rs745943874 Chr.1: 115642922-115642922 on Build GRCh38
rs77156804 Chr.1: 115642891-115642891 on Build GRCh38
rs754744338 Chr.1: 115642580-115642580 on Build GRCh38
rs3010377 Chr.1: 115642979-115642979 on Build GRCh38
rs918893363 Chr.1: 115642748-115642748 on Build GRCh38
rs944982520 Chr.1: 115642713-115642713 on Build GRCh38
rs945624205 Chr.1: 115642590-115642590 on Build GRCh38
rs1033766449 Chr.1: 115642555-115642555 on Build GRCh38
rs574452695 Chr.1: 115642696-115642696 on Build GRCh38
rs989551144 Chr.1: 115642666-115642666 on Build GRCh38
rs772750859 Chr.1: 115642968-115642968 on Build GRCh38
rs1004556661 Chr.1: 115642540-115642540 on Build GRCh38
rs956923866 Chr.1: 115642632-115642632 on Build GRCh38
rs777991738 Chr.1: 115642609-115642609 on Build GRCh38
rs78205573 Chr.1: 115642545-115642545 on Build GRCh38
rs1004101818 Chr.1: 115642743-115642743 on Build GRCh38
rs1013640045 Chr.1: 115642903-115642903 on Build GRCh38
rs933793853 Chr.1: 115642704-115642704 on Build GRCh38
rs992313239 Chr.1: 115642578-115642578 on Build GRCh38
rs117124095 Chr.1: 115642816-115642816 on Build GRCh38
rs1024156488 Chr.1: 115642579-115642579 on Build GRCh38
rs541870608 Chr.1: 115642728-115642728 on Build GRCh38
rs530845782 Chr.1: 115642792-115642792 on Build GRCh38
rs980105037 Chr.1: 115642643-115642643 on Build GRCh38
rs114793902 Chr.1: 115642945-115642945 on Build GRCh38
rs377154941 Chr.1: 115642695-115642695 on Build GRCh38
rs375978264 Chr.1: 115642539-115642539 on Build GRCh38
rs1031000608 Chr.1: 115642625-115642625 on Build GRCh38
rs980786373 Chr.1: 115642552-115642552 on Build GRCh38
rs1003490283 Chr.1: 115642608-115642608 on Build GRCh38
rs1054477862 Chr.1: 115642553-115642553 on Build GRCh38
rs866486955 Chr.1: 115642769-115642769 on Build GRCh38
rs1016908662 Chr.1: 115642542-115642542 on Build GRCh38
rs912173404 Chr.1: 115642532-115642532 on Build GRCh38
rs1050893319 Chr.1: 115642711-115642711 on Build GRCh38
rs1045573920 Chr.1: 115642869-115642869 on Build GRCh38
rs1012719402 Chr.1: 115642574-115642574 on Build GRCh38
rs368126073 Chr.1: 115642822-115642822 on Build GRCh38
rs899613165 Chr.1: 115642701-115642701 on Build GRCh38
rs953554667 Chr.1: 115642693-115642693 on Build GRCh38
rs887055237 Chr.1: 115642516-115642516 on Build GRCh38
rs866727381 Chr.1: 115642931-115642931 on Build GRCh38
rs960851696 Chr.1: 115642559-115642559 on Build GRCh38
rs998053324 Chr.1: 115642505-115642505 on Build GRCh38
rs1048778656 Chr.1: 115642772-115642773 on Build GRCh38
rs12029798 Chr.1: 115642675-115642675 on Build GRCh38

View Full Product Details