Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 38680553-38681100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLF3
Entrez Gene ID
51274
Gene Name
Kruppel like factor 3
Gene Aliases
BKLF
UniGene
Hs.298658
Gene Chromosome Location
Chr.4: 38664050-38701508 on Build GRCh38
Transcripts
NM_016531.5 XM_017008279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7096047 c.6C>T p.L2L Chr.4: 38680631-38680631 on Build GRCh38
1055093 c.51C>T p.V17V Chr.4: 38680676-38680676 on Build GRCh38
4124516 c.4C>A p.L2I Chr.4: 38680629-38680629 on Build GRCh38
3603495 c.38C>T p.A13V Chr.4: 38680663-38680663 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974632761 Chr.4: 38680958-38680958 on Build GRCh38
rs1018977432 Chr.4: 38681009-38681009 on Build GRCh38
rs745639335 Chr.4: 38680620-38680620 on Build GRCh38
rs758590333 Chr.4: 38680598-38680598 on Build GRCh38
rs759970773 Chr.4: 38680709-38680709 on Build GRCh38
rs376574389 Chr.4: 38680772-38680772 on Build GRCh38
rs767146040 Chr.4: 38680678-38680678 on Build GRCh38
rs113777729 Chr.4: 38681013-38681013 on Build GRCh38
rs775480358 Chr.4: 38680644-38680644 on Build GRCh38
rs902928945 Chr.4: 38680880-38680880 on Build GRCh38
rs962933104 Chr.4: 38680922-38680922 on Build GRCh38
rs368232443 Chr.4: 38680789-38680789 on Build GRCh38
rs912236905 Chr.4: 38680984-38680984 on Build GRCh38
rs748231742 Chr.4: 38680597-38680597 on Build GRCh38
rs1029263523 Chr.4: 38680965-38680965 on Build GRCh38
rs369046290 Chr.4: 38680594-38680594 on Build GRCh38
rs542048069 Chr.4: 38680642-38680642 on Build GRCh38
rs947207635 Chr.4: 38680845-38680845 on Build GRCh38
rs59800324 Chr.4: 38680770-38680770 on Build GRCh38
rs911974688 Chr.4: 38680792-38680792 on Build GRCh38
rs929963850 Chr.4: 38680974-38680974 on Build GRCh38
rs538412961 Chr.4: 38680816-38680816 on Build GRCh38
rs766702022 Chr.4: 38680698-38680698 on Build GRCh38
rs754329020 Chr.4: 38680708-38680708 on Build GRCh38
rs899081992 Chr.4: 38680618-38680618 on Build GRCh38
rs777881664 Chr.4: 38680604-38680604 on Build GRCh38
rs1015774555 Chr.4: 38680756-38680756 on Build GRCh38
rs921911116 Chr.4: 38680777-38680777 on Build GRCh38
rs983009887 Chr.4: 38680744-38680744 on Build GRCh38
rs920456174 Chr.4: 38680800-38680800 on Build GRCh38
rs7686818 Chr.4: 38680991-38680991 on Build GRCh38
rs930541891 Chr.4: 38680824-38680824 on Build GRCh38
rs867895440 Chr.4: 38680661-38680661 on Build GRCh38
rs7686990 Chr.4: 38681074-38681074 on Build GRCh38
rs998649320 Chr.4: 38680895-38680895 on Build GRCh38
rs765747220 Chr.4: 38680712-38680712 on Build GRCh38
rs530847522 Chr.4: 38680676-38680676 on Build GRCh38
rs539185731 Chr.4: 38680582-38680582 on Build GRCh38
rs758440170 Chr.4: 38680726-38680726 on Build GRCh38
rs1055655319 Chr.4: 38680841-38680841 on Build GRCh38
rs761067325 Chr.4: 38680692-38680692 on Build GRCh38
rs544382357 Chr.4: 38681033-38681033 on Build GRCh38
rs564387505 Chr.4: 38680743-38680743 on Build GRCh38
rs545371823 Chr.4: 38681059-38681060 on Build GRCh38
rs571898977 Chr.4: 38680876-38680876 on Build GRCh38
rs188574512 Chr.4: 38680903-38680903 on Build GRCh38
rs779032725 Chr.4: 38680585-38680585 on Build GRCh38
rs755027700 Chr.4: 38680583-38680583 on Build GRCh38
rs894002609 Chr.4: 38680911-38680911 on Build GRCh38
rs754382317 Chr.4: 38680581-38680581 on Build GRCh38
rs574188968 Chr.4: 38680964-38680964 on Build GRCh38
rs1043840537 Chr.4: 38680849-38680849 on Build GRCh38
rs972297924 Chr.4: 38680798-38680798 on Build GRCh38
rs558200054 Chr.4: 38680822-38680822 on Build GRCh38
rs894115781 Chr.4: 38680854-38680854 on Build GRCh38
rs114311199 Chr.4: 38680679-38680679 on Build GRCh38
rs868550494 Chr.4: 38680924-38680924 on Build GRCh38
rs146585530 Chr.4: 38680773-38680773 on Build GRCh38
rs139101829 Chr.4: 38680788-38680788 on Build GRCh38
rs894168109 Chr.4: 38680705-38680705 on Build GRCh38
rs940822790 Chr.4: 38680797-38680797 on Build GRCh38
rs751238714 Chr.4: 38680605-38680605 on Build GRCh38
rs1000105949 Chr.4: 38680750-38680750 on Build GRCh38
rs949759473 Chr.4: 38680860-38680860 on Build GRCh38
rs773927544 Chr.4: 38680668-38680668 on Build GRCh38
rs184595489 Chr.4: 38680775-38680775 on Build GRCh38
rs7686626 Chr.4: 38680919-38680919 on Build GRCh38
rs76718870 Chr.4: 38681060-38681060 on Build GRCh38
rs1011140269 Chr.4: 38680942-38680942 on Build GRCh38
rs752727483 Chr.4: 38680725-38680725 on Build GRCh38
rs879499436 Chr.4: 38680785-38680785 on Build GRCh38
rs564265829 Chr.4: 38681046-38681046 on Build GRCh38
rs755927689 Chr.4: 38680731-38680731 on Build GRCh38
rs1002267181 Chr.4: 38680865-38680865 on Build GRCh38
rs13115081 Chr.4: 38680980-38680980 on Build GRCh38
rs768274115 Chr.4: 38680659-38680659 on Build GRCh38
rs893005216 Chr.4: 38680821-38680821 on Build GRCh38
rs561821179 Chr.4: 38680643-38680643 on Build GRCh38
rs970885499 Chr.4: 38680742-38680742 on Build GRCh38
rs778028275 Chr.4: 38680730-38680730 on Build GRCh38

View Full Product Details