Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 53390390-53390901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIP1L1
Entrez Gene ID
81608
Gene Name
factor interacting with PAPOLA and CPSF1
Gene Aliases
FIP1, Rhe, hFip1
UniGene
Hs.555109
Gene Chromosome Location
Chr.4: 53377572-53462611 on Build GRCh38
Transcripts
NM_001134937.1 NM_001134938.1 NM_030917.3 XM_005265769.4 XM_005265773.4 XM_005265774.4 XM_005265778.4 XM_005265779.4 XM_005265781.4 XM_005265782.4 XM_017008662.1 XM_017008663.1 XM_017008664.1 XM_017008665.1 XM_017008666.1 XM_017008667.1 XM_017008668.1 XM_017008669.1 XM_017008670.1 XM_017008671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3301140 c.398G>A p.G133E Chr.4: 53390521-53390521 on Build GRCh38
3380945 c.452G>A p.R151H Chr.4: 53390620-53390620 on Build GRCh38
3946430 c.415G>C p.E139Q Chr.4: 53390583-53390583 on Build GRCh38
1328494 c.454C>G p.L152V Chr.4: 53390577-53390577 on Build GRCh38
1055982 c.476C>A p.S159Y Chr.4: 53390599-53390599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200532181 Chr.4: 53390643-53390643 on Build GRCh38
rs903578300 Chr.4: 53390759-53390759 on Build GRCh38
rs962829726 Chr.4: 53390869-53390869 on Build GRCh38
rs776910628 Chr.4: 53390501-53390501 on Build GRCh38
rs747516253 Chr.4: 53390585-53390585 on Build GRCh38
rs533015237 Chr.4: 53390557-53390557 on Build GRCh38
rs900153117 Chr.4: 53390810-53390810 on Build GRCh38
rs756117540 Chr.4: 53390531-53390531 on Build GRCh38
rs576169299 Chr.4: 53390870-53390870 on Build GRCh38
rs553168623 Chr.4: 53390441-53390441 on Build GRCh38
rs962714089 Chr.4: 53390602-53390602 on Build GRCh38
rs6847795 Chr.4: 53390867-53390867 on Build GRCh38
rs771449831 Chr.4: 53390482-53390482 on Build GRCh38
rs1000457953 Chr.4: 53390454-53390454 on Build GRCh38
rs761071476 Chr.4: 53390491-53390491 on Build GRCh38
rs950561985 Chr.4: 53390480-53390480 on Build GRCh38
rs536447705 Chr.4: 53390551-53390551 on Build GRCh38
rs201898974 Chr.4: 53390504-53390504 on Build GRCh38
rs764060640 Chr.4: 53390521-53390521 on Build GRCh38
rs376633164 Chr.4: 53390439-53390439 on Build GRCh38
rs772589019 Chr.4: 53390578-53390578 on Build GRCh38
rs564742007 Chr.4: 53390421-53390421 on Build GRCh38
rs746015493 Chr.4: 53390670-53390670 on Build GRCh38
rs372261150 Chr.4: 53390505-53390505 on Build GRCh38
rs376761566 Chr.4: 53390474-53390474 on Build GRCh38
rs1030225660 Chr.4: 53390815-53390815 on Build GRCh38
rs750432865 Chr.4: 53390522-53390522 on Build GRCh38
rs757828548 Chr.4: 53390670-53390670 on Build GRCh38
rs763075105 Chr.4: 53390674-53390674 on Build GRCh38
rs147147160 Chr.4: 53390611-53390611 on Build GRCh38
rs564318110 Chr.4: 53390499-53390499 on Build GRCh38
rs1042132128 Chr.4: 53390748-53390750 on Build GRCh38
rs775831867 Chr.4: 53390652-53390652 on Build GRCh38
rs773815782 Chr.4: 53390483-53390483 on Build GRCh38
rs754706833 Chr.4: 53390559-53390559 on Build GRCh38
rs749901001 Chr.4: 53390490-53390490 on Build GRCh38
rs779800742 Chr.4: 53390681-53390682 on Build GRCh38
rs28575132 Chr.4: 53390473-53390473 on Build GRCh38
rs771668615 Chr.4: 53390575-53390575 on Build GRCh38
rs903547111 Chr.4: 53390420-53390420 on Build GRCh38
rs180796398 Chr.4: 53390470-53390470 on Build GRCh38
rs1030609504 Chr.4: 53390476-53390476 on Build GRCh38
rs751527746 Chr.4: 53390679-53390679 on Build GRCh38
rs771373532 Chr.4: 53390608-53390608 on Build GRCh38
rs938294834 Chr.4: 53390636-53390636 on Build GRCh38
rs377306119 Chr.4: 53390462-53390462 on Build GRCh38
rs528844036 Chr.4: 53390782-53390782 on Build GRCh38
rs1039144593 Chr.4: 53390792-53390792 on Build GRCh38
rs113733465 Chr.4: 53390859-53390859 on Build GRCh38
rs77155086 Chr.4: 53390677-53390677 on Build GRCh38
rs765397396 Chr.4: 53390512-53390512 on Build GRCh38
rs548991942 Chr.4: 53390826-53390826 on Build GRCh38
rs926643535 Chr.4: 53390872-53390872 on Build GRCh38
rs778666066 Chr.4: 53390560-53390560 on Build GRCh38
rs565635564 Chr.4: 53390836-53390836 on Build GRCh38
rs747730970 Chr.4: 53390562-53390562 on Build GRCh38
rs759856536 Chr.4: 53390635-53390635 on Build GRCh38
rs758546983 Chr.4: 53390517-53390517 on Build GRCh38
rs945523366 Chr.4: 53390725-53390725 on Build GRCh38
rs780007050 Chr.4: 53390546-53390546 on Build GRCh38
rs926639704 Chr.4: 53390620-53390620 on Build GRCh38
rs773486825 Chr.4: 53390657-53390657 on Build GRCh38
rs867597145 Chr.4: 53390469-53390469 on Build GRCh38
rs752754997 Chr.4: 53390514-53390514 on Build GRCh38

View Full Product Details