Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70522620-70523166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMTN
Entrez Gene ID
401138
Gene Name
amelotin
Gene Aliases
UNQ689
UniGene
Hs.453069
Gene Chromosome Location
Chr.4: 70518572-70532743 on Build GRCh38
Transcripts
NM_001286731.1 NM_212557.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3918084 c.79C>T p.P27S Chr.4: 70522779-70522779 on Build GRCh38
2830206 c.79C>A p.P27T Chr.4: 70522779-70522779 on Build GRCh38
1694491 c.73_74GG>AA p.G25K Chr.4: 70522773-70522774 on Build GRCh38
238776 c.125A>C p.Q42P Chr.4: 70522825-70522825 on Build GRCh38
3760718 c.134A>G p.N45S Chr.4: 70522834-70522834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111449428 Chr.4: 70522672-70522672 on Build GRCh38
rs763467017 Chr.4: 70522696-70522696 on Build GRCh38
rs113159590 Chr.4: 70522887-70522887 on Build GRCh38
rs372431208 Chr.4: 70522717-70522717 on Build GRCh38
rs146511674 Chr.4: 70522960-70522960 on Build GRCh38
rs1003723588 Chr.4: 70523110-70523110 on Build GRCh38
rs906672235 Chr.4: 70523083-70523083 on Build GRCh38
rs1013281349 Chr.4: 70523085-70523085 on Build GRCh38
rs755665274 Chr.4: 70522738-70522738 on Build GRCh38
rs577753771 Chr.4: 70522902-70522902 on Build GRCh38
rs756527265 Chr.4: 70522848-70522848 on Build GRCh38
rs920123071 Chr.4: 70522995-70522995 on Build GRCh38
rs904378746 Chr.4: 70523131-70523132 on Build GRCh38
rs576236311 Chr.4: 70522861-70522861 on Build GRCh38
rs892705959 Chr.4: 70523140-70523140 on Build GRCh38
rs763285086 Chr.4: 70522825-70522825 on Build GRCh38
rs746970202 Chr.4: 70523011-70523011 on Build GRCh38
rs867307831 Chr.4: 70522801-70522801 on Build GRCh38
rs777319072 Chr.4: 70522799-70522799 on Build GRCh38
rs987320269 Chr.4: 70523010-70523010 on Build GRCh38
rs989887637 Chr.4: 70522940-70522940 on Build GRCh38
rs17149005 Chr.4: 70522949-70522949 on Build GRCh38
rs764545054 Chr.4: 70522709-70522709 on Build GRCh38
rs4123809 Chr.4: 70522677-70522677 on Build GRCh38
rs1012033653 Chr.4: 70522877-70522877 on Build GRCh38
rs1044856437 Chr.4: 70523066-70523066 on Build GRCh38
rs768170923 Chr.4: 70522757-70522757 on Build GRCh38
rs781763600 Chr.4: 70522732-70522732 on Build GRCh38
rs1037766671 Chr.4: 70523016-70523016 on Build GRCh38
rs539569834 Chr.4: 70523121-70523121 on Build GRCh38
rs921801005 Chr.4: 70522729-70522729 on Build GRCh38
rs986811493 Chr.4: 70522962-70522962 on Build GRCh38
rs116013987 Chr.4: 70522872-70522872 on Build GRCh38
rs1056726471 Chr.4: 70523025-70523026 on Build GRCh38
rs563726713 Chr.4: 70522924-70522924 on Build GRCh38
rs1030989720 Chr.4: 70522951-70522951 on Build GRCh38
rs898206114 Chr.4: 70522788-70522788 on Build GRCh38
rs752145690 Chr.4: 70522769-70522769 on Build GRCh38
rs1003776088 Chr.4: 70522896-70522896 on Build GRCh38
rs375591289 Chr.4: 70522932-70522932 on Build GRCh38
rs565640919 Chr.4: 70522728-70522728 on Build GRCh38
rs1042890811 Chr.4: 70523093-70523093 on Build GRCh38
rs767574361 Chr.4: 70522857-70522886 on Build GRCh38
rs898860320 Chr.4: 70523052-70523052 on Build GRCh38
rs528708150 Chr.4: 70522969-70522969 on Build GRCh38
rs751953663 Chr.4: 70522715-70522715 on Build GRCh38
rs923970167 Chr.4: 70522653-70522653 on Build GRCh38
rs17676469 Chr.4: 70523132-70523132 on Build GRCh38
rs368417913 Chr.4: 70522833-70522833 on Build GRCh38
rs779660073 Chr.4: 70522854-70522854 on Build GRCh38
rs199658784 Chr.4: 70522798-70522798 on Build GRCh38
rs894356577 Chr.4: 70523061-70523061 on Build GRCh38
rs933172138 Chr.4: 70523000-70523000 on Build GRCh38
rs544996199 Chr.4: 70522917-70522917 on Build GRCh38
rs1040295997 Chr.4: 70522773-70522773 on Build GRCh38
rs747929560 Chr.4: 70522768-70522768 on Build GRCh38
rs550241813 Chr.4: 70523005-70523005 on Build GRCh38
rs559429206 Chr.4: 70522901-70522901 on Build GRCh38
rs73826536 Chr.4: 70522989-70522989 on Build GRCh38
rs573800800 Chr.4: 70522884-70522884 on Build GRCh38
rs28754427 Chr.4: 70522662-70522662 on Build GRCh38
rs773143693 Chr.4: 70522784-70522784 on Build GRCh38
rs750973162 Chr.4: 70522847-70522847 on Build GRCh38
rs139060712 Chr.4: 70522869-70522869 on Build GRCh38
rs1012710819 Chr.4: 70522873-70522873 on Build GRCh38
rs7660807 Chr.4: 70522834-70522834 on Build GRCh38
rs908206008 Chr.4: 70523013-70523013 on Build GRCh38
rs779389513 Chr.4: 70522753-70522753 on Build GRCh38
rs745430063 Chr.4: 70522734-70522734 on Build GRCh38
rs1023797373 Chr.4: 70522886-70522886 on Build GRCh38
rs947768723 Chr.4: 70523054-70523054 on Build GRCh38
rs767938897 Chr.4: 70522839-70522839 on Build GRCh38
rs139939210 Chr.4: 70523012-70523012 on Build GRCh38
rs774171618 Chr.4: 70522766-70522766 on Build GRCh38
rs915321311 Chr.4: 70522676-70522676 on Build GRCh38
rs748991501 Chr.4: 70522754-70522754 on Build GRCh38
rs979420240 Chr.4: 70522646-70522646 on Build GRCh38
rs772051982 Chr.4: 70522778-70522778 on Build GRCh38
rs371617949 Chr.4: 70522863-70522863 on Build GRCh38
rs768685017 Chr.4: 70522681-70522681 on Build GRCh38
rs145073103 Chr.4: 70522796-70522796 on Build GRCh38

View Full Product Details