Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70818008-70818553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRSF1
Entrez Gene ID
2926
Gene Name
G-rich RNA sequence binding factor 1
Gene Aliases
-
UniGene
Hs.309763
Gene Chromosome Location
Chr.4: 70815782-70843274 on Build GRCh38
Transcripts
NM_001098477.1 NM_002092.3

SNP Detail

SNP ID Chromosome Location
rs937101552 Chr.4: 70818447-70818447 on Build GRCh38
rs192400687 Chr.4: 70818264-70818264 on Build GRCh38
rs775416312 Chr.4: 70818179-70818179 on Build GRCh38
rs911070306 Chr.4: 70818346-70818346 on Build GRCh38
rs530995081 Chr.4: 70818171-70818171 on Build GRCh38
rs915464260 Chr.4: 70818461-70818461 on Build GRCh38
rs1045491146 Chr.4: 70818129-70818129 on Build GRCh38
rs922715417 Chr.4: 70818507-70818507 on Build GRCh38
rs982263364 Chr.4: 70818134-70818134 on Build GRCh38
rs771838002 Chr.4: 70818133-70818133 on Build GRCh38
rs563764173 Chr.4: 70818152-70818152 on Build GRCh38
rs189247677 Chr.4: 70818092-70818092 on Build GRCh38
rs546376966 Chr.4: 70818469-70818469 on Build GRCh38
rs764153099 Chr.4: 70818349-70818349 on Build GRCh38
rs942701368 Chr.4: 70818466-70818466 on Build GRCh38
rs997759932 Chr.4: 70818255-70818255 on Build GRCh38
rs1039667697 Chr.4: 70818472-70818472 on Build GRCh38
rs768787481 Chr.4: 70818329-70818329 on Build GRCh38
rs756357301 Chr.4: 70818062-70818063 on Build GRCh38
rs5020093 Chr.4: 70818220-70818220 on Build GRCh38
rs893789270 Chr.4: 70818188-70818188 on Build GRCh38
rs372364262 Chr.4: 70818421-70818422 on Build GRCh38
rs761895286 Chr.4: 70818365-70818365 on Build GRCh38
rs183977761 Chr.4: 70818072-70818072 on Build GRCh38
rs983152305 Chr.4: 70818090-70818090 on Build GRCh38
rs963389677 Chr.4: 70818323-70818323 on Build GRCh38
rs895387593 Chr.4: 70818154-70818154 on Build GRCh38
rs1022538532 Chr.4: 70818509-70818509 on Build GRCh38
rs765531123 Chr.4: 70818492-70818492 on Build GRCh38
rs929648883 Chr.4: 70818372-70818372 on Build GRCh38
rs901222401 Chr.4: 70818225-70818225 on Build GRCh38
rs778522331 Chr.4: 70818113-70818113 on Build GRCh38
rs886387367 Chr.4: 70818272-70818272 on Build GRCh38
rs895333874 Chr.4: 70818474-70818474 on Build GRCh38
rs1004788113 Chr.4: 70818275-70818275 on Build GRCh38
rs942692667 Chr.4: 70818210-70818210 on Build GRCh38
rs77486851 Chr.4: 70818118-70818118 on Build GRCh38
rs376004155 Chr.4: 70818078-70818078 on Build GRCh38
rs1016167126 Chr.4: 70818319-70818319 on Build GRCh38
rs924097691 Chr.4: 70818355-70818355 on Build GRCh38
rs538846494 Chr.4: 70818276-70818276 on Build GRCh38
rs989708690 Chr.4: 70818407-70818407 on Build GRCh38
rs941312596 Chr.4: 70818063-70818063 on Build GRCh38
rs567624255 Chr.4: 70818195-70818195 on Build GRCh38
rs934079715 Chr.4: 70818508-70818508 on Build GRCh38
rs906932117 Chr.4: 70818446-70818446 on Build GRCh38
rs770734950 Chr.4: 70818097-70818097 on Build GRCh38
rs181975116 Chr.4: 70818335-70818335 on Build GRCh38
rs955668327 Chr.4: 70818255-70818256 on Build GRCh38
rs749645819 Chr.4: 70818302-70818305 on Build GRCh38
rs571638775 Chr.4: 70818273-70818273 on Build GRCh38
rs990996362 Chr.4: 70818112-70818112 on Build GRCh38
rs767740051 Chr.4: 70818053-70818053 on Build GRCh38
rs112514106 Chr.4: 70818142-70818142 on Build GRCh38
rs186529981 Chr.4: 70818184-70818184 on Build GRCh38
rs776801562 Chr.4: 70818351-70818351 on Build GRCh38
rs918210841 Chr.4: 70818352-70818352 on Build GRCh38
rs1037974391 Chr.4: 70818091-70818091 on Build GRCh38

View Full Product Details