Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 102576824-102577361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFKB1
Entrez Gene ID
4790
Gene Name
nuclear factor kappa B subunit 1
Gene Aliases
CVID12, EBP-1, KBF1, NF-kB1, NF-kappa-B, NF-kappaB, NFKB-p105, NFKB-p50, NFkappaB, p105, p50
UniGene
Hs.618430
Gene Chromosome Location
Chr.4: 102501329-102617302 on Build GRCh38
Transcripts
NM_001165412.1 NM_001319226.1 NM_003998.3 XM_011532006.2 XM_011532008.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3660815 c.424A>G p.I142V Chr.4: 102576892-102576892 on Build GRCh38
6098804 c.541G>C p.A181P Chr.4: 102577009-102577009 on Build GRCh38
6441510 c.547G>T p.G183C Chr.4: 102577015-102577015 on Build GRCh38
325945 c.524A>T p.D175V Chr.4: 102576992-102576992 on Build GRCh38
4552755 c.569G>A p.G190E Chr.4: 102577037-102577037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752604471 Chr.4: 102577088-102577088 on Build GRCh38
rs938675952 Chr.4: 102577293-102577293 on Build GRCh38
rs142991798 Chr.4: 102576996-102576996 on Build GRCh38
rs989172843 Chr.4: 102576959-102576959 on Build GRCh38
rs111881231 Chr.4: 102577044-102577044 on Build GRCh38
rs1055399432 Chr.4: 102577314-102577314 on Build GRCh38
rs373392563 Chr.4: 102577091-102577091 on Build GRCh38
rs772170990 Chr.4: 102576957-102576957 on Build GRCh38
rs569471547 Chr.4: 102577254-102577254 on Build GRCh38
rs35791621 Chr.4: 102577337-102577338 on Build GRCh38
rs749720974 Chr.4: 102576895-102576895 on Build GRCh38
rs765087404 Chr.4: 102577001-102577001 on Build GRCh38
rs776627927 Chr.4: 102577074-102577074 on Build GRCh38
rs974909934 Chr.4: 102576863-102576863 on Build GRCh38
rs769167080 Chr.4: 102576915-102576915 on Build GRCh38
rs369582218 Chr.4: 102576879-102576879 on Build GRCh38
rs770490082 Chr.4: 102576869-102576869 on Build GRCh38
rs544055935 Chr.4: 102577299-102577299 on Build GRCh38
rs538979542 Chr.4: 102577328-102577328 on Build GRCh38
rs777359468 Chr.4: 102576865-102576865 on Build GRCh38
rs989224990 Chr.4: 102577219-102577219 on Build GRCh38
rs230542 Chr.4: 102577298-102577298 on Build GRCh38
rs768984306 Chr.4: 102577046-102577046 on Build GRCh38
rs779868472 Chr.4: 102577035-102577035 on Build GRCh38
rs777291215 Chr.4: 102577024-102577024 on Build GRCh38
rs373249652 Chr.4: 102576951-102576951 on Build GRCh38
rs772378391 Chr.4: 102577060-102577060 on Build GRCh38
rs779388557 Chr.4: 102577048-102577048 on Build GRCh38
rs1004846467 Chr.4: 102577083-102577083 on Build GRCh38
rs869320754 Chr.4: 102576933-102576934 on Build GRCh38
rs760714767 Chr.4: 102576963-102576963 on Build GRCh38
rs953757585 Chr.4: 102576882-102576882 on Build GRCh38
rs376486117 Chr.4: 102577067-102577067 on Build GRCh38
rs945312107 Chr.4: 102576976-102576976 on Build GRCh38
rs150692807 Chr.4: 102577182-102577182 on Build GRCh38
rs927891718 Chr.4: 102577295-102577295 on Build GRCh38
rs752377230 Chr.4: 102577008-102577008 on Build GRCh38
rs765072888 Chr.4: 102577077-102577077 on Build GRCh38
rs760835829 Chr.4: 102577064-102577064 on Build GRCh38
rs996772190 Chr.4: 102577141-102577141 on Build GRCh38
rs541231003 Chr.4: 102577076-102577076 on Build GRCh38
rs201874528 Chr.4: 102577028-102577028 on Build GRCh38
rs751158419 Chr.4: 102577026-102577026 on Build GRCh38
rs778505137 Chr.4: 102577229-102577230 on Build GRCh38
rs927338898 Chr.4: 102577286-102577286 on Build GRCh38
rs980284843 Chr.4: 102577281-102577281 on Build GRCh38
rs544807864 Chr.4: 102577184-102577184 on Build GRCh38
rs762107099 Chr.4: 102576956-102576956 on Build GRCh38
rs909357536 Chr.4: 102577180-102577180 on Build GRCh38
rs753794335 Chr.4: 102576985-102576985 on Build GRCh38
rs563271057 Chr.4: 102577236-102577236 on Build GRCh38
rs569980066 Chr.4: 102576975-102576975 on Build GRCh38
rs370312017 Chr.4: 102577018-102577018 on Build GRCh38
rs780802592 Chr.4: 102577038-102577038 on Build GRCh38
rs869225322 Chr.4: 102576874-102576874 on Build GRCh38
rs766407539 Chr.4: 102577187-102577187 on Build GRCh38
rs746509528 Chr.4: 102576866-102576866 on Build GRCh38
rs891658460 Chr.4: 102576863-102576864 on Build GRCh38
rs771028054 Chr.4: 102577069-102577069 on Build GRCh38
rs914953389 Chr.4: 102577222-102577222 on Build GRCh38
rs966296147 Chr.4: 102577137-102577137 on Build GRCh38
rs376562815 Chr.4: 102576961-102576961 on Build GRCh38
rs571841268 Chr.4: 102577328-102577328 on Build GRCh38
rs553248692 Chr.4: 102577146-102577146 on Build GRCh38
rs371141633 Chr.4: 102577063-102577063 on Build GRCh38
rs977759503 Chr.4: 102577264-102577264 on Build GRCh38
rs748452969 Chr.4: 102577057-102577057 on Build GRCh38

View Full Product Details