Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107858392-107858887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGMS2
Entrez Gene ID
166929
Gene Name
sphingomyelin synthase 2
Gene Aliases
SMS2
UniGene
Hs.595423
Gene Chromosome Location
Chr.4: 107823645-107915047 on Build GRCh38
Transcripts
NM_001136258.1 XM_011531698.2 XM_011531699.2 XM_011531700.2 XM_011531701.2 XM_011531702.2 XM_017007839.1 XM_017007840.1 XM_017007841.1

SNP Detail

SNP ID Chromosome Location
rs535797912 Chr.4: 107858686-107858686 on Build GRCh38
rs912546716 Chr.4: 107858796-107858796 on Build GRCh38
rs910783158 Chr.4: 107858847-107858847 on Build GRCh38
rs551654504 Chr.4: 107858749-107858749 on Build GRCh38
rs60086311 Chr.4: 107858850-107858850 on Build GRCh38
rs962862533 Chr.4: 107858715-107858715 on Build GRCh38
rs987235204 Chr.4: 107858779-107858779 on Build GRCh38
rs976047954 Chr.4: 107858851-107858851 on Build GRCh38
rs1010334468 Chr.4: 107858555-107858555 on Build GRCh38
rs762580462 Chr.4: 107858843-107858843 on Build GRCh38
rs986412236 Chr.4: 107858830-107858830 on Build GRCh38
rs199832850 Chr.4: 107858677-107858678 on Build GRCh38
rs776579797 Chr.4: 107858683-107858683 on Build GRCh38
rs962205893 Chr.4: 107858427-107858427 on Build GRCh38
rs974230309 Chr.4: 107858727-107858727 on Build GRCh38
rs903229623 Chr.4: 107858631-107858631 on Build GRCh38
rs570465982 Chr.4: 107858417-107858417 on Build GRCh38
rs764942621 Chr.4: 107858792-107858792 on Build GRCh38
rs930947645 Chr.4: 107858546-107858546 on Build GRCh38
rs1002472264 Chr.4: 107858793-107858793 on Build GRCh38
rs934489910 Chr.4: 107858646-107858646 on Build GRCh38
rs182519364 Chr.4: 107858773-107858773 on Build GRCh38
rs900395507 Chr.4: 107858831-107858831 on Build GRCh38
rs986767746 Chr.4: 107858564-107858564 on Build GRCh38
rs918686030 Chr.4: 107858739-107858739 on Build GRCh38
rs1033182629 Chr.4: 107858475-107858475 on Build GRCh38
rs891504575 Chr.4: 107858519-107858519 on Build GRCh38
rs114703098 Chr.4: 107858659-107858659 on Build GRCh38
rs910581075 Chr.4: 107858569-107858569 on Build GRCh38
rs112395721 Chr.4: 107858459-107858459 on Build GRCh38
rs191996416 Chr.4: 107858807-107858807 on Build GRCh38
rs80058543 Chr.4: 107858474-107858474 on Build GRCh38
rs1012861885 Chr.4: 107858716-107858716 on Build GRCh38
rs1040499668 Chr.4: 107858618-107858618 on Build GRCh38
rs1044693779 Chr.4: 107858740-107858740 on Build GRCh38
rs1015303650 Chr.4: 107858801-107858801 on Build GRCh38
rs1023944411 Chr.4: 107858633-107858633 on Build GRCh38
rs924599906 Chr.4: 107858855-107858855 on Build GRCh38
rs756949867 Chr.4: 107858861-107858862 on Build GRCh38
rs1027843407 Chr.4: 107858818-107858818 on Build GRCh38
rs944661148 Chr.4: 107858588-107858588 on Build GRCh38
rs1038867703 Chr.4: 107858420-107858420 on Build GRCh38
rs961143294 Chr.4: 107858804-107858804 on Build GRCh38
rs142194051 Chr.4: 107858670-107858672 on Build GRCh38
rs556321743 Chr.4: 107858583-107858583 on Build GRCh38
rs1038921849 Chr.4: 107858824-107858824 on Build GRCh38
rs754381734 Chr.4: 107858536-107858537 on Build GRCh38
rs1015269488 Chr.4: 107858426-107858426 on Build GRCh38
rs944613236 Chr.4: 107858418-107858418 on Build GRCh38
rs60536621 Chr.4: 107858677-107858679 on Build GRCh38
rs187143225 Chr.4: 107858775-107858775 on Build GRCh38
rs750139499 Chr.4: 107858828-107858828 on Build GRCh38
rs1034555037 Chr.4: 107858694-107858694 on Build GRCh38
rs539311752 Chr.4: 107858675-107858675 on Build GRCh38

View Full Product Details