Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121332740-121333241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QRFPR
Entrez Gene ID
84109
Gene Name
pyroglutamylated RFamide peptide receptor
Gene Aliases
AQ27, GPR103, SP9155
UniGene
Hs.368977
Gene Chromosome Location
Chr.4: 121328642-121381029 on Build GRCh38
Transcripts
NM_198179.2 XM_017008693.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
378780 c.582T>A p.Y194* Chr.4: 121333036-121333036 on Build GRCh38
3767672 c.794C>T p.A265V Chr.4: 121332824-121332824 on Build GRCh38
171373 c.761G>A p.R254Q Chr.4: 121332857-121332857 on Build GRCh38
1753515 c.760C>T p.R254* Chr.4: 121332858-121332858 on Build GRCh38
2149989 c.622C>T p.P208S Chr.4: 121332996-121332996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050741030 Chr.4: 121332828-121332828 on Build GRCh38
rs755461217 Chr.4: 121332782-121332782 on Build GRCh38
rs762933333 Chr.4: 121332942-121332943 on Build GRCh38
rs754271299 Chr.4: 121333098-121333098 on Build GRCh38
rs1050249246 Chr.4: 121333089-121333089 on Build GRCh38
rs1046962572 Chr.4: 121333026-121333026 on Build GRCh38
rs995397062 Chr.4: 121333003-121333003 on Build GRCh38
rs550527843 Chr.4: 121333041-121333041 on Build GRCh38
rs778680995 Chr.4: 121332917-121332917 on Build GRCh38
rs946116238 Chr.4: 121333006-121333006 on Build GRCh38
rs763101064 Chr.4: 121332989-121332989 on Build GRCh38
rs896541011 Chr.4: 121333139-121333139 on Build GRCh38
rs369660516 Chr.4: 121332797-121332797 on Build GRCh38
rs746457807 Chr.4: 121333174-121333174 on Build GRCh38
rs984408619 Chr.4: 121332947-121332947 on Build GRCh38
rs142777322 Chr.4: 121332991-121332991 on Build GRCh38
rs751054833 Chr.4: 121333106-121333106 on Build GRCh38
rs866096999 Chr.4: 121333210-121333210 on Build GRCh38
rs944616615 Chr.4: 121333125-121333125 on Build GRCh38
rs76291924 Chr.4: 121333107-121333107 on Build GRCh38
rs1023599033 Chr.4: 121333148-121333148 on Build GRCh38
rs968453156 Chr.4: 121333151-121333151 on Build GRCh38
rs535593070 Chr.4: 121333198-121333198 on Build GRCh38
rs781563504 Chr.4: 121332783-121332783 on Build GRCh38
rs755448253 Chr.4: 121332873-121332873 on Build GRCh38
rs372254772 Chr.4: 121333055-121333055 on Build GRCh38
rs762721901 Chr.4: 121333116-121333116 on Build GRCh38
rs33977434 Chr.4: 121332964-121332964 on Build GRCh38
rs55975435 Chr.4: 121332859-121332859 on Build GRCh38
rs768479826 Chr.4: 121332852-121332852 on Build GRCh38
rs543792498 Chr.4: 121332857-121332857 on Build GRCh38
rs776477870 Chr.4: 121332948-121332948 on Build GRCh38
rs147498995 Chr.4: 121332868-121332868 on Build GRCh38
rs528669363 Chr.4: 121332946-121332946 on Build GRCh38
rs771751283 Chr.4: 121332839-121332839 on Build GRCh38
rs773463011 Chr.4: 121332862-121332862 on Build GRCh38
rs935480016 Chr.4: 121332900-121332900 on Build GRCh38
rs747943209 Chr.4: 121332963-121332963 on Build GRCh38
rs769585879 Chr.4: 121333086-121333086 on Build GRCh38
rs758150585 Chr.4: 121332933-121332933 on Build GRCh38
rs745501370 Chr.4: 121332875-121332875 on Build GRCh38
rs753093066 Chr.4: 121332915-121332915 on Build GRCh38
rs376848061 Chr.4: 121333093-121333093 on Build GRCh38
rs759592563 Chr.4: 121333021-121333021 on Build GRCh38
rs139457842 Chr.4: 121332923-121332923 on Build GRCh38
rs1006162971 Chr.4: 121333199-121333199 on Build GRCh38
rs756930579 Chr.4: 121332813-121332813 on Build GRCh38
rs766292815 Chr.4: 121333101-121333102 on Build GRCh38
rs139519471 Chr.4: 121333173-121333173 on Build GRCh38
rs746564389 Chr.4: 121332945-121332945 on Build GRCh38
rs539983896 Chr.4: 121333143-121333143 on Build GRCh38
rs756680535 Chr.4: 121333064-121333064 on Build GRCh38
rs766396199 Chr.4: 121332869-121332869 on Build GRCh38
rs977469806 Chr.4: 121333013-121333013 on Build GRCh38
rs897463201 Chr.4: 121333141-121333141 on Build GRCh38
rs267600005 Chr.4: 121332845-121332845 on Build GRCh38
rs779908589 Chr.4: 121333102-121333102 on Build GRCh38
rs745354141 Chr.4: 121332837-121332837 on Build GRCh38
rs536718920 Chr.4: 121332801-121332801 on Build GRCh38
rs942376930 Chr.4: 121332936-121332936 on Build GRCh38
rs140968526 Chr.4: 121332858-121332858 on Build GRCh38
rs758116121 Chr.4: 121333100-121333100 on Build GRCh38
rs193075383 Chr.4: 121332916-121332916 on Build GRCh38
rs925822475 Chr.4: 121333043-121333043 on Build GRCh38
rs935807134 Chr.4: 121333071-121333071 on Build GRCh38
rs1041611110 Chr.4: 121332767-121332767 on Build GRCh38
rs555418974 Chr.4: 121333209-121333209 on Build GRCh38
rs144384829 Chr.4: 121332848-121332848 on Build GRCh38
rs1000125285 Chr.4: 121332798-121332798 on Build GRCh38
rs139685422 Chr.4: 121332987-121332987 on Build GRCh38
rs924147002 Chr.4: 121332877-121332877 on Build GRCh38
rs55776058 Chr.4: 121333129-121333129 on Build GRCh38
rs958932028 Chr.4: 121332971-121332971 on Build GRCh38
rs778766546 Chr.4: 121332833-121332833 on Build GRCh38
rs774410992 Chr.4: 121332992-121332992 on Build GRCh38
rs773074046 Chr.4: 121332880-121332880 on Build GRCh38
rs912326460 Chr.4: 121333206-121333206 on Build GRCh38
rs116679707 Chr.4: 121332870-121332870 on Build GRCh38
rs765333959 Chr.4: 121332774-121332784 on Build GRCh38
rs779655405 Chr.4: 121332944-121332944 on Build GRCh38
rs776518040 Chr.4: 121332931-121332931 on Build GRCh38
rs775174264 Chr.4: 121332846-121332846 on Build GRCh38
rs201762984 Chr.4: 121333061-121333061 on Build GRCh38

View Full Product Details