Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122187765-122188269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1109
Entrez Gene ID
84162
Gene Name
KIAA1109
Gene Aliases
FSA, Tweek
UniGene
Hs.408142
Gene Chromosome Location
Chr.4: 122152333-122362759 on Build GRCh38
Transcripts
NM_015312.3 XM_005263282.1 XM_005263287.1 XM_006714344.1 XM_011532319.1 XM_011532320.2 XM_011532321.2 XM_011532322.1 XM_011532323.1 XM_011532324.1 XM_011532325.1 XM_011532326.1 XM_011532327.1 XM_017008695.1 XM_017008696.1 XM_017008697.1 XM_017008699.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5208668 c.791G>T p.S264I Chr.4: 122188058-122188058 on Build GRCh38
1212064 c.632G>A p.R211H Chr.4: 122187899-122187899 on Build GRCh38
4121741 c.812A>G p.Q271R Chr.4: 122188079-122188079 on Build GRCh38
4414656 c.766A>T p.N256Y Chr.4: 122188033-122188033 on Build GRCh38
6357877 c.789T>C p.P263P Chr.4: 122188056-122188056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201183349 Chr.4: 122187964-122187964 on Build GRCh38
rs75241146 Chr.4: 122187892-122187892 on Build GRCh38
rs777456549 Chr.4: 122187940-122187940 on Build GRCh38
rs749466883 Chr.4: 122187952-122187952 on Build GRCh38
rs1003075996 Chr.4: 122187803-122187803 on Build GRCh38
rs923084336 Chr.4: 122188194-122188194 on Build GRCh38
rs754213562 Chr.4: 122187880-122187880 on Build GRCh38
rs549555909 Chr.4: 122188129-122188129 on Build GRCh38
rs766410435 Chr.4: 122187882-122187882 on Build GRCh38
rs779716498 Chr.4: 122187851-122187851 on Build GRCh38
rs773421614 Chr.4: 122187877-122187877 on Build GRCh38
rs764532863 Chr.4: 122187880-122187881 on Build GRCh38
rs745359276 Chr.4: 122187847-122187847 on Build GRCh38
rs768113084 Chr.4: 122187893-122187893 on Build GRCh38
rs77045869 Chr.4: 122187875-122187875 on Build GRCh38
rs911435456 Chr.4: 122188170-122188170 on Build GRCh38
rs1057498830 Chr.4: 122188002-122188002 on Build GRCh38
rs941842841 Chr.4: 122188224-122188224 on Build GRCh38
rs533492901 Chr.4: 122187963-122187963 on Build GRCh38
rs776257970 Chr.4: 122187871-122187871 on Build GRCh38
rs748039202 Chr.4: 122187872-122187872 on Build GRCh38
rs1031675963 Chr.4: 122187879-122187879 on Build GRCh38
rs754477649 Chr.4: 122188060-122188060 on Build GRCh38
rs904649154 Chr.4: 122188068-122188068 on Build GRCh38
rs771140500 Chr.4: 122188105-122188105 on Build GRCh38
rs895018619 Chr.4: 122188230-122188230 on Build GRCh38
rs866799293 Chr.4: 122188188-122188188 on Build GRCh38
rs1047551618 Chr.4: 122187885-122187885 on Build GRCh38
rs750381940 Chr.4: 122188128-122188128 on Build GRCh38
rs887592355 Chr.4: 122188218-122188218 on Build GRCh38
rs1055061790 Chr.4: 122188225-122188225 on Build GRCh38
rs751097213 Chr.4: 122188046-122188046 on Build GRCh38
rs776009746 Chr.4: 122187975-122187975 on Build GRCh38
rs772446521 Chr.4: 122187974-122187974 on Build GRCh38
rs371242033 Chr.4: 122188103-122188103 on Build GRCh38
rs954662218 Chr.4: 122188113-122188113 on Build GRCh38
rs10009888 Chr.4: 122187864-122187864 on Build GRCh38
rs950494057 Chr.4: 122188227-122188227 on Build GRCh38
rs777606154 Chr.4: 122188104-122188104 on Build GRCh38
rs762124617 Chr.4: 122187879-122187880 on Build GRCh38
rs780567816 Chr.4: 122188084-122188084 on Build GRCh38
rs76417860 Chr.4: 122187911-122187911 on Build GRCh38
rs560741861 Chr.4: 122188015-122188015 on Build GRCh38
rs149259417 Chr.4: 122188014-122188014 on Build GRCh38
rs80041392 Chr.4: 122187883-122187883 on Build GRCh38
rs779166057 Chr.4: 122188122-122188122 on Build GRCh38
rs761092038 Chr.4: 122187984-122187984 on Build GRCh38
rs771195744 Chr.4: 122187956-122187956 on Build GRCh38
rs929049378 Chr.4: 122188198-122188198 on Build GRCh38
rs373288568 Chr.4: 122187926-122187926 on Build GRCh38
rs758575559 Chr.4: 122188093-122188094 on Build GRCh38
rs976115359 Chr.4: 122188181-122188181 on Build GRCh38
rs746105011 Chr.4: 122188125-122188125 on Build GRCh38
rs769562271 Chr.4: 122187936-122187936 on Build GRCh38
rs753000567 Chr.4: 122187904-122187904 on Build GRCh38
rs887393464 Chr.4: 122187793-122187793 on Build GRCh38
rs750025993 Chr.4: 122187923-122187923 on Build GRCh38
rs1022089563 Chr.4: 122187857-122187857 on Build GRCh38
rs757923971 Chr.4: 122187850-122187850 on Build GRCh38
rs761161148 Chr.4: 122187920-122187920 on Build GRCh38
rs764479755 Chr.4: 122187992-122187992 on Build GRCh38
rs758144941 Chr.4: 122187924-122187924 on Build GRCh38
rs530404096 Chr.4: 122188101-122188101 on Build GRCh38
rs894607299 Chr.4: 122187835-122187835 on Build GRCh38
rs920437020 Chr.4: 122188226-122188226 on Build GRCh38
rs114241742 Chr.4: 122188200-122188200 on Build GRCh38
rs529510065 Chr.4: 122188064-122188064 on Build GRCh38
rs766199212 Chr.4: 122188041-122188041 on Build GRCh38
rs1035886220 Chr.4: 122187834-122187834 on Build GRCh38
rs78156050 Chr.4: 122187879-122187879 on Build GRCh38
rs964582677 Chr.4: 122188193-122188193 on Build GRCh38
rs760016138 Chr.4: 122187884-122187884 on Build GRCh38
rs746893883 Chr.4: 122187852-122187852 on Build GRCh38
rs748046439 Chr.4: 122187930-122187930 on Build GRCh38
rs138600216 Chr.4: 122187927-122187927 on Build GRCh38
rs377687856 Chr.4: 122188098-122188098 on Build GRCh38
rs750511847 Chr.4: 122187935-122187936 on Build GRCh38
rs558906578 Chr.4: 122187969-122187969 on Build GRCh38
rs373274740 Chr.4: 122187876-122187876 on Build GRCh38
rs67625562 Chr.4: 122188180-122188180 on Build GRCh38
rs1008832349 Chr.4: 122188164-122188164 on Build GRCh38

View Full Product Details