Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 151688551-151689043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATB
Entrez Gene ID
5188
Gene Name
glutamyl-tRNA amidotransferase subunit B
Gene Aliases
HSPC199, PET112, PET112L
UniGene
Hs.119316
Gene Chromosome Location
Chr.4: 151669773-151761023 on Build GRCh38
Transcripts
NM_004564.2 XM_005263063.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5865878 c.1201G>A p.E401K Chr.4: 151688760-151688760 on Build GRCh38
1052074 c.1291C>A p.L431M Chr.4: 151688670-151688670 on Build GRCh38
6744051 c.1200C>T p.N400N Chr.4: 151688761-151688761 on Build GRCh38
291166 c.1262_1263insA p.V422fs*3 Chr.4: 151688698-151688699 on Build GRCh38
365292 c.1229A>T p.N410I Chr.4: 151688732-151688732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027888065 Chr.4: 151688663-151688663 on Build GRCh38
rs372072363 Chr.4: 151688809-151688809 on Build GRCh38
rs750592921 Chr.4: 151688796-151688796 on Build GRCh38
rs773816347 Chr.4: 151688679-151688679 on Build GRCh38
rs74890783 Chr.4: 151688780-151688780 on Build GRCh38
rs776583990 Chr.4: 151688629-151688629 on Build GRCh38
rs923987621 Chr.4: 151688934-151688934 on Build GRCh38
rs367744397 Chr.4: 151688776-151688777 on Build GRCh38
rs62327344 Chr.4: 151688674-151688674 on Build GRCh38
rs946607778 Chr.4: 151688691-151688691 on Build GRCh38
rs199613429 Chr.4: 151688847-151688847 on Build GRCh38
rs1002002047 Chr.4: 151688865-151688865 on Build GRCh38
rs75029272 Chr.4: 151688632-151688632 on Build GRCh38
rs779160408 Chr.4: 151688790-151688790 on Build GRCh38
rs371343864 Chr.4: 151688678-151688678 on Build GRCh38
rs536730940 Chr.4: 151689022-151689022 on Build GRCh38
rs201498468 Chr.4: 151688845-151688846 on Build GRCh38
rs954072085 Chr.4: 151688787-151688787 on Build GRCh38
rs553772039 Chr.4: 151688770-151688770 on Build GRCh38
rs766116675 Chr.4: 151688736-151688736 on Build GRCh38
rs970625367 Chr.4: 151688829-151688829 on Build GRCh38
rs778290594 Chr.4: 151688586-151688586 on Build GRCh38
rs765991308 Chr.4: 151688644-151688644 on Build GRCh38
rs749510749 Chr.4: 151688769-151688771 on Build GRCh38
rs754883655 Chr.4: 151688957-151688957 on Build GRCh38
rs11390610 Chr.4: 151688846-151688847 on Build GRCh38
rs756493784 Chr.4: 151688582-151688582 on Build GRCh38
rs943494089 Chr.4: 151688717-151688717 on Build GRCh38
rs747068068 Chr.4: 151688804-151688804 on Build GRCh38
rs745931789 Chr.4: 151688604-151688604 on Build GRCh38
rs773182148 Chr.4: 151688733-151688733 on Build GRCh38
rs777645735 Chr.4: 151688810-151688810 on Build GRCh38
rs184095954 Chr.4: 151688727-151688727 on Build GRCh38
rs762793295 Chr.4: 151688638-151688638 on Build GRCh38
rs3749561 Chr.4: 151688965-151688965 on Build GRCh38
rs778169685 Chr.4: 151688768-151688769 on Build GRCh38
rs939465400 Chr.4: 151688959-151688959 on Build GRCh38
rs752245667 Chr.4: 151688761-151688761 on Build GRCh38
rs771268620 Chr.4: 151688593-151688593 on Build GRCh38
rs1056643820 Chr.4: 151688583-151688583 on Build GRCh38
rs1012861004 Chr.4: 151689011-151689011 on Build GRCh38
rs761753310 Chr.4: 151688630-151688630 on Build GRCh38
rs72960993 Chr.4: 151688876-151688876 on Build GRCh38
rs1015296995 Chr.4: 151688713-151688713 on Build GRCh38
rs773220169 Chr.4: 151688672-151688683 on Build GRCh38
rs200642263 Chr.4: 151688776-151688776 on Build GRCh38
rs749741185 Chr.4: 151688587-151688587 on Build GRCh38
rs895298892 Chr.4: 151689007-151689007 on Build GRCh38
rs192265062 Chr.4: 151688651-151688651 on Build GRCh38
rs376818767 Chr.4: 151688755-151688755 on Build GRCh38
rs774003095 Chr.4: 151688753-151688753 on Build GRCh38
rs772133016 Chr.4: 151688697-151688697 on Build GRCh38
rs781690104 Chr.4: 151688732-151688732 on Build GRCh38
rs546867224 Chr.4: 151688710-151688710 on Build GRCh38
rs375829780 Chr.4: 151689016-151689016 on Build GRCh38
rs1023109028 Chr.4: 151688609-151688609 on Build GRCh38
rs760380028 Chr.4: 151688745-151688745 on Build GRCh38
rs759119923 Chr.4: 151688759-151688759 on Build GRCh38
rs7376710 Chr.4: 151688777-151688777 on Build GRCh38
rs987728041 Chr.4: 151688662-151688662 on Build GRCh38
rs757648076 Chr.4: 151688779-151688779 on Build GRCh38
rs746102249 Chr.4: 151688689-151688689 on Build GRCh38
rs915190951 Chr.4: 151688815-151688815 on Build GRCh38
rs538542170 Chr.4: 151688769-151688770 on Build GRCh38
rs772077154 Chr.4: 151688617-151688617 on Build GRCh38
rs774488966 Chr.4: 151688597-151688597 on Build GRCh38
rs901882038 Chr.4: 151688754-151688754 on Build GRCh38
rs139737204 Chr.4: 151688686-151688686 on Build GRCh38
rs775437747 Chr.4: 151688699-151688699 on Build GRCh38
rs200981641 Chr.4: 151688763-151688763 on Build GRCh38
rs33954718 Chr.4: 151688847-151688848 on Build GRCh38
rs946690756 Chr.4: 151688817-151688817 on Build GRCh38
rs767270767 Chr.4: 151688760-151688760 on Build GRCh38
rs11292952 Chr.4: 151688769-151688769 on Build GRCh38
rs796620397 Chr.4: 151688781-151688781 on Build GRCh38
rs764667192 Chr.4: 151688768-151688768 on Build GRCh38
rs779157602 Chr.4: 151688688-151688688 on Build GRCh38
rs780380365 Chr.4: 151688803-151688803 on Build GRCh38
rs3210929 Chr.4: 151688705-151688705 on Build GRCh38

View Full Product Details