Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 161385336-161385846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL5
Entrez Gene ID
56884
Gene Name
follistatin like 5
Gene Aliases
-
UniGene
Hs.32452
Gene Chromosome Location
Chr.4: 161383892-162164034 on Build GRCh38
Transcripts
NM_001128427.2 NM_001128428.2 NM_020116.4 XM_011532126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5556090 c.2534G>C p.G845A Chr.4: 161385757-161385757 on Build GRCh38
3601509 c.2512G>A p.G838R Chr.4: 161385779-161385779 on Build GRCh38
4538195 c.2513G>A p.G838E Chr.4: 161385778-161385778 on Build GRCh38
7260168 c.2517T>A p.N839K Chr.4: 161385774-161385774 on Build GRCh38
3601510 c.2492A>C p.E831A Chr.4: 161385799-161385799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021789229 Chr.4: 161385401-161385401 on Build GRCh38
rs866954589 Chr.4: 161385532-161385532 on Build GRCh38
rs558519180 Chr.4: 161385420-161385420 on Build GRCh38
rs34535035 Chr.4: 161385377-161385378 on Build GRCh38
rs3210497 Chr.4: 161385377-161385377 on Build GRCh38
rs759803829 Chr.4: 161385411-161385411 on Build GRCh38
rs879225654 Chr.4: 161385547-161385550 on Build GRCh38
rs777240415 Chr.4: 161385789-161385789 on Build GRCh38
rs917065188 Chr.4: 161385407-161385407 on Build GRCh38
rs772552857 Chr.4: 161385728-161385728 on Build GRCh38
rs770280281 Chr.4: 161385803-161385803 on Build GRCh38
rs997924070 Chr.4: 161385522-161385522 on Build GRCh38
rs199897436 Chr.4: 161385783-161385783 on Build GRCh38
rs181618554 Chr.4: 161385673-161385673 on Build GRCh38
rs959316670 Chr.4: 161385421-161385421 on Build GRCh38
rs35914540 Chr.4: 161385459-161385460 on Build GRCh38
rs377376012 Chr.4: 161385503-161385503 on Build GRCh38
rs983196630 Chr.4: 161385492-161385492 on Build GRCh38
rs1022512254 Chr.4: 161385539-161385539 on Build GRCh38
rs185856951 Chr.4: 161385412-161385412 on Build GRCh38
rs748706336 Chr.4: 161385712-161385712 on Build GRCh38
rs904702888 Chr.4: 161385632-161385632 on Build GRCh38
rs751443756 Chr.4: 161385768-161385768 on Build GRCh38
rs563187263 Chr.4: 161385820-161385820 on Build GRCh38
rs372559754 Chr.4: 161385748-161385748 on Build GRCh38
rs752384275 Chr.4: 161385784-161385784 on Build GRCh38
rs776031059 Chr.4: 161385821-161385821 on Build GRCh38
rs980772089 Chr.4: 161385591-161385591 on Build GRCh38
rs1051813787 Chr.4: 161385374-161385374 on Build GRCh38
rs148257997 Chr.4: 161385757-161385757 on Build GRCh38
rs373898553 Chr.4: 161385716-161385716 on Build GRCh38
rs991862378 Chr.4: 161385440-161385440 on Build GRCh38
rs983110936 Chr.4: 161385739-161385739 on Build GRCh38
rs763158960 Chr.4: 161385701-161385701 on Build GRCh38
rs369496219 Chr.4: 161385612-161385612 on Build GRCh38
rs752011014 Chr.4: 161385819-161385819 on Build GRCh38
rs370089550 Chr.4: 161385738-161385738 on Build GRCh38
rs960806419 Chr.4: 161385679-161385679 on Build GRCh38
rs775904717 Chr.4: 161385734-161385734 on Build GRCh38
rs1043089306 Chr.4: 161385622-161385622 on Build GRCh38
rs748842658 Chr.4: 161385791-161385791 on Build GRCh38
rs3210496 Chr.4: 161385378-161385378 on Build GRCh38
rs755793404 Chr.4: 161385788-161385788 on Build GRCh38
rs933223336 Chr.4: 161385523-161385523 on Build GRCh38
rs79881082 Chr.4: 161385403-161385403 on Build GRCh38
rs760950961 Chr.4: 161385737-161385737 on Build GRCh38
rs1001688051 Chr.4: 161385641-161385641 on Build GRCh38
rs1014506166 Chr.4: 161385682-161385682 on Build GRCh38
rs113295707 Chr.4: 161385531-161385531 on Build GRCh38
rs909046482 Chr.4: 161385500-161385500 on Build GRCh38
rs760012562 Chr.4: 161385718-161385718 on Build GRCh38
rs753977385 Chr.4: 161385744-161385744 on Build GRCh38
rs188920078 Chr.4: 161385659-161385659 on Build GRCh38
rs981270926 Chr.4: 161385381-161385381 on Build GRCh38
rs1024633163 Chr.4: 161385456-161385456 on Build GRCh38
rs921845949 Chr.4: 161385513-161385513 on Build GRCh38
rs926698814 Chr.4: 161385387-161385387 on Build GRCh38
rs909778559 Chr.4: 161385771-161385771 on Build GRCh38
rs3210498 Chr.4: 161385362-161385362 on Build GRCh38
rs779532456 Chr.4: 161385804-161385804 on Build GRCh38
rs577611440 Chr.4: 161385675-161385675 on Build GRCh38
rs72987242 Chr.4: 161385486-161385486 on Build GRCh38
rs746356276 Chr.4: 161385815-161385815 on Build GRCh38
rs555576226 Chr.4: 161385467-161385467 on Build GRCh38
rs555020870 Chr.4: 161385544-161385544 on Build GRCh38
rs754822086 Chr.4: 161385772-161385772 on Build GRCh38
rs892636027 Chr.4: 161385524-161385524 on Build GRCh38
rs116508077 Chr.4: 161385510-161385510 on Build GRCh38
rs376492064 Chr.4: 161385717-161385717 on Build GRCh38

View Full Product Details