Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184694790-184695315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPU
Entrez Gene ID
79682
Gene Name
centromere protein U
Gene Aliases
CENP50, CENPU50, KLIP1, MLF1IP, PBIP1
UniGene
Hs.575032
Gene Chromosome Location
Chr.4: 184694065-184734132 on Build GRCh38
Transcripts
NM_024629.3 NR_104593.1 XM_005263218.4 NM_001300767.1 NM_001300768.1 NM_152683.3 XM_011531719.1 XM_011531720.1 XM_011531721.2 XM_011531723.1 XM_011531724.2 XM_011531725.1 XM_011531726.2 XM_011531729.1 XM_011531730.2 XM_017007864.1 XM_017007865.1 XM_017007866.1 XM_017007867.1 XM_017007868.1 XM_017007869.1 XM_017007870.1 XM_017007871.1 XM_017007872.1 XM_017007873.1 XM_017007874.1 XM_017007875.1

Gene Symbol
PRIMPOL
Entrez Gene ID
201973
Gene Name
primase and DNA directed polymerase
Gene Aliases
CCDC111, MYP22
UniGene
Hs.481307
Gene Chromosome Location
Chr.4: 184649613-184694959 on Build GRCh38
Transcripts
NM_024629.3 NR_104593.1 XM_005263218.4 NM_001300767.1 NM_001300768.1 NM_152683.3 XM_011531719.1 XM_011531720.1 XM_011531721.2 XM_011531723.1 XM_011531724.2 XM_011531725.1 XM_011531726.2 XM_011531729.1 XM_011531730.2 XM_017007864.1 XM_017007865.1 XM_017007866.1 XM_017007867.1 XM_017007868.1 XM_017007869.1 XM_017007870.1 XM_017007871.1 XM_017007872.1 XM_017007873.1 XM_017007874.1 XM_017007875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3602447 c.1251G>A p.Q417Q Chr.4: 184695294-184695294 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112946554 Chr.4: 184694825-184694825 on Build GRCh38
rs115257498 Chr.4: 184694938-184694938 on Build GRCh38
rs548503845 Chr.4: 184695135-184695135 on Build GRCh38
rs535081246 Chr.4: 184695136-184695136 on Build GRCh38
rs546766922 Chr.4: 184694905-184694905 on Build GRCh38
rs201805354 Chr.4: 184694969-184694971 on Build GRCh38
rs775203598 Chr.4: 184694823-184694823 on Build GRCh38
rs923203544 Chr.4: 184695162-184695162 on Build GRCh38
rs954604137 Chr.4: 184695193-184695193 on Build GRCh38
rs934700831 Chr.4: 184694965-184694965 on Build GRCh38
rs145587090 Chr.4: 184695066-184695069 on Build GRCh38
rs753840311 Chr.4: 184695259-184695259 on Build GRCh38
rs914767728 Chr.4: 184695272-184695272 on Build GRCh38
rs943309086 Chr.4: 184694847-184694847 on Build GRCh38
rs879899009 Chr.4: 184695207-184695207 on Build GRCh38
rs747549012 Chr.4: 184695279-184695279 on Build GRCh38
rs1016133809 Chr.4: 184695111-184695111 on Build GRCh38
rs182729371 Chr.4: 184695155-184695155 on Build GRCh38
rs150279210 Chr.4: 184694971-184694974 on Build GRCh38
rs1013205261 Chr.4: 184694991-184694991 on Build GRCh38
rs368201719 Chr.4: 184694977-184694977 on Build GRCh38
rs777929955 Chr.4: 184695247-184695247 on Build GRCh38
rs780331725 Chr.4: 184695270-184695270 on Build GRCh38
rs751544653 Chr.4: 184695238-184695238 on Build GRCh38
rs568878807 Chr.4: 184695031-184695031 on Build GRCh38
rs754828733 Chr.4: 184695243-184695243 on Build GRCh38
rs78781309 Chr.4: 184695146-184695146 on Build GRCh38
rs1053017503 Chr.4: 184695105-184695105 on Build GRCh38
rs113547036 Chr.4: 184695093-184695093 on Build GRCh38
rs1006351271 Chr.4: 184695048-184695048 on Build GRCh38
rs994531984 Chr.4: 184694932-184694932 on Build GRCh38
rs978798727 Chr.4: 184695022-184695022 on Build GRCh38
rs539917926 Chr.4: 184695039-184695039 on Build GRCh38
rs924717350 Chr.4: 184694958-184694958 on Build GRCh38
rs975058653 Chr.4: 184694875-184694875 on Build GRCh38
rs1004453102 Chr.4: 184695267-184695267 on Build GRCh38
rs934604195 Chr.4: 184695087-184695087 on Build GRCh38
rs746289988 Chr.4: 184695263-184695263 on Build GRCh38
rs905551745 Chr.4: 184694910-184694911 on Build GRCh38
rs529952585 Chr.4: 184694842-184694842 on Build GRCh38
rs112552929 Chr.4: 184695017-184695017 on Build GRCh38
rs772441825 Chr.4: 184695266-184695266 on Build GRCh38
rs895649470 Chr.4: 184694830-184694830 on Build GRCh38
rs986765211 Chr.4: 184695237-184695237 on Build GRCh38
rs952975149 Chr.4: 184694950-184694950 on Build GRCh38
rs933975899 Chr.4: 184694913-184694913 on Build GRCh38
rs537825967 Chr.4: 184695040-184695040 on Build GRCh38
rs375081091 Chr.4: 184695260-184695260 on Build GRCh38
rs191677497 Chr.4: 184694966-184694966 on Build GRCh38
rs768540804 Chr.4: 184695001-184695001 on Build GRCh38
rs372080848 Chr.4: 184694816-184694816 on Build GRCh38
rs917116732 Chr.4: 184695108-184695108 on Build GRCh38
rs371773929 Chr.4: 184694980-184694980 on Build GRCh38
rs6826883 Chr.4: 184695013-184695013 on Build GRCh38
rs13478 Chr.4: 184695284-184695284 on Build GRCh38
rs551184112 Chr.4: 184694900-184694903 on Build GRCh38
rs893284449 Chr.4: 184694989-184694989 on Build GRCh38
rs1002554741 Chr.4: 184695019-184695019 on Build GRCh38
rs796798501 Chr.4: 184694903-184694903 on Build GRCh38
rs905626809 Chr.4: 184695220-184695220 on Build GRCh38
rs768998598 Chr.4: 184695281-184695281 on Build GRCh38

View Full Product Details