Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186069066-186069565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR3
Entrez Gene ID
7098
Gene Name
toll like receptor 3
Gene Aliases
CD283, IIAE2
UniGene
Hs.657724
Gene Chromosome Location
Chr.4: 186069155-186086724 on Build GRCh38
Transcripts
NM_003265.2

SNP Detail

SNP ID Chromosome Location
rs765240183 Chr.4: 186069254-186069254 on Build GRCh38
rs990745818 Chr.4: 186069121-186069121 on Build GRCh38
rs556041380 Chr.4: 186069303-186069303 on Build GRCh38
rs1045781049 Chr.4: 186069427-186069427 on Build GRCh38
rs933801247 Chr.4: 186069288-186069288 on Build GRCh38
rs181240423 Chr.4: 186069363-186069363 on Build GRCh38
rs1016541165 Chr.4: 186069270-186069270 on Build GRCh38
rs975243939 Chr.4: 186069131-186069131 on Build GRCh38
rs1054454732 Chr.4: 186069193-186069193 on Build GRCh38
rs888058447 Chr.4: 186069507-186069507 on Build GRCh38
rs5743308 Chr.4: 186069138-186069138 on Build GRCh38
rs1007069093 Chr.4: 186069198-186069198 on Build GRCh38
rs1054486123 Chr.4: 186069360-186069360 on Build GRCh38
rs548726936 Chr.4: 186069162-186069162 on Build GRCh38
rs369227060 Chr.4: 186069141-186069142 on Build GRCh38
rs958721805 Chr.4: 186069379-186069379 on Build GRCh38
rs377321123 Chr.4: 186069324-186069324 on Build GRCh38
rs970958273 Chr.4: 186069435-186069435 on Build GRCh38
rs752829886 Chr.4: 186069423-186069423 on Build GRCh38
rs983784673 Chr.4: 186069269-186069269 on Build GRCh38
rs972557876 Chr.4: 186069436-186069436 on Build GRCh38
rs572349948 Chr.4: 186069105-186069105 on Build GRCh38
rs942444985 Chr.4: 186069117-186069117 on Build GRCh38
rs907365237 Chr.4: 186069428-186069428 on Build GRCh38
rs917727850 Chr.4: 186069133-186069134 on Build GRCh38
rs994160580 Chr.4: 186069242-186069242 on Build GRCh38
rs1024910007 Chr.4: 186069213-186069213 on Build GRCh38
rs918418515 Chr.4: 186069535-186069535 on Build GRCh38
rs116729895 Chr.4: 186069190-186069190 on Build GRCh38
rs993718265 Chr.4: 186069431-186069431 on Build GRCh38
rs5743310 Chr.4: 186069239-186069239 on Build GRCh38
rs555039956 Chr.4: 186069442-186069442 on Build GRCh38
rs1012989183 Chr.4: 186069211-186069211 on Build GRCh38
rs975104016 Chr.4: 186069284-186069284 on Build GRCh38
rs563826923 Chr.4: 186069096-186069096 on Build GRCh38
rs922437267 Chr.4: 186069287-186069287 on Build GRCh38
rs1006408929 Chr.4: 186069518-186069518 on Build GRCh38
rs1016572452 Chr.4: 186069533-186069533 on Build GRCh38
rs1048354027 Chr.4: 186069474-186069474 on Build GRCh38
rs963647505 Chr.4: 186069541-186069541 on Build GRCh38
rs1041602179 Chr.4: 186069176-186069176 on Build GRCh38
rs894458865 Chr.4: 186069375-186069375 on Build GRCh38
rs5743309 Chr.4: 186069191-186069191 on Build GRCh38
rs546403644 Chr.4: 186069231-186069231 on Build GRCh38
rs1026525491 Chr.4: 186069243-186069243 on Build GRCh38
rs539644579 Chr.4: 186069290-186069290 on Build GRCh38
rs529322725 Chr.4: 186069098-186069098 on Build GRCh38
rs138085273 Chr.4: 186069195-186069195 on Build GRCh38
rs367653100 Chr.4: 186069413-186069413 on Build GRCh38
rs373809349 Chr.4: 186069281-186069281 on Build GRCh38

View Full Product Details