Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186082993-186083504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR3
Entrez Gene ID
7098
Gene Name
toll like receptor 3
Gene Aliases
CD283, IIAE2
UniGene
Hs.657724
Gene Chromosome Location
Chr.4: 186069155-186086724 on Build GRCh38
Transcripts
NM_003265.2 XM_017008577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1753673 c.1755A>G p.L585L Chr.4: 186083441-186083441 on Build GRCh38
4529011 c.1579G>T p.E527* Chr.4: 186083265-186083265 on Build GRCh38
1229396 c.1378G>A p.E460K Chr.4: 186083064-186083064 on Build GRCh38
6802507 c.1462C>T p.R488* Chr.4: 186083148-186083148 on Build GRCh38
1753672 c.1738G>C p.E580Q Chr.4: 186083424-186083424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747344213 Chr.4: 186083399-186083399 on Build GRCh38
rs750322659 Chr.4: 186083332-186083332 on Build GRCh38
rs751421740 Chr.4: 186083353-186083353 on Build GRCh38
rs140712645 Chr.4: 186083402-186083402 on Build GRCh38
rs1053020064 Chr.4: 186083046-186083046 on Build GRCh38
rs767185774 Chr.4: 186083147-186083147 on Build GRCh38
rs867445555 Chr.4: 186083164-186083164 on Build GRCh38
rs759176345 Chr.4: 186083137-186083137 on Build GRCh38
rs767914092 Chr.4: 186083212-186083212 on Build GRCh38
rs774023990 Chr.4: 186083026-186083026 on Build GRCh38
rs751968287 Chr.4: 186083145-186083145 on Build GRCh38
rs368882892 Chr.4: 186083433-186083433 on Build GRCh38
rs192543136 Chr.4: 186083456-186083456 on Build GRCh38
rs376735334 Chr.4: 186083411-186083411 on Build GRCh38
rs772594650 Chr.4: 186083385-186083385 on Build GRCh38
rs532943362 Chr.4: 186083264-186083264 on Build GRCh38
rs767401326 Chr.4: 186083448-186083448 on Build GRCh38
rs149184129 Chr.4: 186083370-186083370 on Build GRCh38
rs769114608 Chr.4: 186083400-186083400 on Build GRCh38
rs772360575 Chr.4: 186083291-186083291 on Build GRCh38
rs144550375 Chr.4: 186083064-186083064 on Build GRCh38
rs764701077 Chr.4: 186083124-186083124 on Build GRCh38
rs35617964 Chr.4: 186083363-186083363 on Build GRCh38
rs757517639 Chr.4: 186083459-186083459 on Build GRCh38
rs746428963 Chr.4: 186083286-186083286 on Build GRCh38
rs765738799 Chr.4: 186083027-186083028 on Build GRCh38
rs753746965 Chr.4: 186083153-186083153 on Build GRCh38
rs763770806 Chr.4: 186083300-186083300 on Build GRCh38
rs757180314 Chr.4: 186083231-186083231 on Build GRCh38
rs768784321 Chr.4: 186083040-186083040 on Build GRCh38
rs764584083 Chr.4: 186083403-186083403 on Build GRCh38
rs184426724 Chr.4: 186083460-186083460 on Build GRCh38
rs774735663 Chr.4: 186083202-186083202 on Build GRCh38
rs751110231 Chr.4: 186083228-186083230 on Build GRCh38
rs113275119 Chr.4: 186083089-186083089 on Build GRCh38
rs766450307 Chr.4: 186083417-186083417 on Build GRCh38
rs748439037 Chr.4: 186083177-186083177 on Build GRCh38
rs779058242 Chr.4: 186083379-186083379 on Build GRCh38
rs200785357 Chr.4: 186083465-186083465 on Build GRCh38
rs202066722 Chr.4: 186083214-186083214 on Build GRCh38
rs121434431 Chr.4: 186083346-186083346 on Build GRCh38
rs775850017 Chr.4: 186083292-186083292 on Build GRCh38
rs143330461 Chr.4: 186083384-186083384 on Build GRCh38
rs750424441 Chr.4: 186083247-186083247 on Build GRCh38
rs762995868 Chr.4: 186083110-186083110 on Build GRCh38
rs751583950 Chr.4: 186083421-186083421 on Build GRCh38
rs571300094 Chr.4: 186083369-186083369 on Build GRCh38
rs780060149 Chr.4: 186083256-186083256 on Build GRCh38
rs369886993 Chr.4: 186083175-186083175 on Build GRCh38
rs200494519 Chr.4: 186083038-186083038 on Build GRCh38
rs893177672 Chr.4: 186083312-186083312 on Build GRCh38
rs764860383 Chr.4: 186083331-186083331 on Build GRCh38
rs372569709 Chr.4: 186083441-186083441 on Build GRCh38
rs758255171 Chr.4: 186083251-186083251 on Build GRCh38
rs953898407 Chr.4: 186083426-186083426 on Build GRCh38
rs1051348300 Chr.4: 186083245-186083245 on Build GRCh38
rs763457697 Chr.4: 186083184-186083184 on Build GRCh38
rs112666655 Chr.4: 186083320-186083320 on Build GRCh38
rs760275329 Chr.4: 186083299-186083299 on Build GRCh38
rs199658443 Chr.4: 186083203-186083203 on Build GRCh38
rs776136889 Chr.4: 186083401-186083401 on Build GRCh38
rs145252794 Chr.4: 186083367-186083367 on Build GRCh38
rs772992927 Chr.4: 186083148-186083148 on Build GRCh38
rs763540090 Chr.4: 186083223-186083223 on Build GRCh38
rs751882299 Chr.4: 186083129-186083129 on Build GRCh38
rs757158320 Chr.4: 186083154-186083154 on Build GRCh38
rs745682135 Chr.4: 186083157-186083157 on Build GRCh38
rs776387492 Chr.4: 186083302-186083302 on Build GRCh38
rs35111588 Chr.4: 186083390-186083390 on Build GRCh38
rs1011526105 Chr.4: 186083329-186083329 on Build GRCh38
rs771361588 Chr.4: 186083193-186083193 on Build GRCh38
rs1011578363 Chr.4: 186083088-186083088 on Build GRCh38
rs34597061 Chr.4: 186083479-186083480 on Build GRCh38
rs373400665 Chr.4: 186083257-186083257 on Build GRCh38
rs773841499 Chr.4: 186083104-186083104 on Build GRCh38
rs778686045 Chr.4: 186083232-186083232 on Build GRCh38
rs932524368 Chr.4: 186083238-186083238 on Build GRCh38
rs758414785 Chr.4: 186083342-186083342 on Build GRCh38
rs758516737 Chr.4: 186083469-186083469 on Build GRCh38
rs201222071 Chr.4: 186083271-186083271 on Build GRCh38
rs770759437 Chr.4: 186083025-186083025 on Build GRCh38
rs762934381 Chr.4: 186083412-186083412 on Build GRCh38
rs3775290 Chr.4: 186083063-186083063 on Build GRCh38
rs771080193 Chr.4: 186083274-186083274 on Build GRCh38
rs772033168 Chr.4: 186083168-186083168 on Build GRCh38
rs749246118 Chr.4: 186083020-186083020 on Build GRCh38
rs769990022 Chr.4: 186083181-186083181 on Build GRCh38
rs753313305 Chr.4: 186083230-186083230 on Build GRCh38
rs760328773 Chr.4: 186083220-186083220 on Build GRCh38
rs777335615 Chr.4: 186083149-186083149 on Build GRCh38
rs773167266 Chr.4: 186083183-186083183 on Build GRCh38
rs749355507 Chr.4: 186083272-186083272 on Build GRCh38
rs147873871 Chr.4: 186083330-186083330 on Build GRCh38
rs201792200 Chr.4: 186083173-186083173 on Build GRCh38
rs770043370 Chr.4: 186083067-186083067 on Build GRCh38
rs780517657 Chr.4: 186083471-186083471 on Build GRCh38
rs866891121 Chr.4: 186083075-186083076 on Build GRCh38
rs774695261 Chr.4: 186083281-186083281 on Build GRCh38
rs977425444 Chr.4: 186083136-186083136 on Build GRCh38

View Full Product Details