Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186597888-186598388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT1
Entrez Gene ID
2195
Gene Name
FAT atypical cadherin 1
Gene Aliases
CDHF7, CDHR8, FAT, ME5, hFat1
UniGene
Hs.481371
Gene Chromosome Location
Chr.4: 186587783-186726915 on Build GRCh38
Transcripts
NM_005245.3 XM_005262834.2 XM_005262835.2 XM_006714139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4123861 c.12245A>G p.Y4082C Chr.4: 186597984-186597984 on Build GRCh38
6099753 c.12215G>A p.G4072E Chr.4: 186598014-186598014 on Build GRCh38
6099752 c.12215G>A p.G4072E Chr.4: 186598014-186598014 on Build GRCh38
6922210 c.12253C>G p.Q4085E Chr.4: 186597976-186597976 on Build GRCh38
3940840 c.12128T>A p.L4043* Chr.4: 186598101-186598101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768136150 Chr.4: 186598041-186598041 on Build GRCh38
rs545481607 Chr.4: 186598053-186598053 on Build GRCh38
rs754875557 Chr.4: 186598146-186598146 on Build GRCh38
rs748431505 Chr.4: 186598133-186598133 on Build GRCh38
rs894089321 Chr.4: 186598167-186598167 on Build GRCh38
rs946765093 Chr.4: 186598321-186598321 on Build GRCh38
rs746415591 Chr.4: 186598025-186598025 on Build GRCh38
rs374696255 Chr.4: 186598110-186598110 on Build GRCh38
rs371227366 Chr.4: 186597984-186597984 on Build GRCh38
rs368487784 Chr.4: 186598232-186598232 on Build GRCh38
rs576041146 Chr.4: 186598068-186598068 on Build GRCh38
rs115127841 Chr.4: 186598223-186598223 on Build GRCh38
rs779558478 Chr.4: 186598024-186598024 on Build GRCh38
rs755438142 Chr.4: 186598100-186598100 on Build GRCh38
rs867710406 Chr.4: 186598092-186598093 on Build GRCh38
rs973067762 Chr.4: 186598181-186598181 on Build GRCh38
rs371989627 Chr.4: 186598016-186598016 on Build GRCh38
rs780625419 Chr.4: 186598085-186598085 on Build GRCh38
rs367812758 Chr.4: 186598014-186598014 on Build GRCh38
rs986439129 Chr.4: 186598215-186598215 on Build GRCh38
rs139477231 Chr.4: 186598134-186598134 on Build GRCh38
rs780242823 Chr.4: 186598032-186598032 on Build GRCh38
rs1015086793 Chr.4: 186598163-186598163 on Build GRCh38
rs371249526 Chr.4: 186597948-186597948 on Build GRCh38
rs921145801 Chr.4: 186598320-186598320 on Build GRCh38
rs751396320 Chr.4: 186598015-186598015 on Build GRCh38
rs561566004 Chr.4: 186598291-186598291 on Build GRCh38
rs762708042 Chr.4: 186598327-186598327 on Build GRCh38
rs372319969 Chr.4: 186598233-186598233 on Build GRCh38
rs1004488220 Chr.4: 186598243-186598243 on Build GRCh38
rs369005392 Chr.4: 186598132-186598132 on Build GRCh38
rs185555465 Chr.4: 186598105-186598105 on Build GRCh38
rs757467460 Chr.4: 186598022-186598022 on Build GRCh38
rs1035560651 Chr.4: 186598011-186598011 on Build GRCh38
rs1016711774 Chr.4: 186598266-186598266 on Build GRCh38
rs149665748 Chr.4: 186598236-186598236 on Build GRCh38
rs77814178 Chr.4: 186597962-186597962 on Build GRCh38
rs778058808 Chr.4: 186598017-186598017 on Build GRCh38
rs761400229 Chr.4: 186597990-186597990 on Build GRCh38
rs750552688 Chr.4: 186598073-186598073 on Build GRCh38
rs1035185907 Chr.4: 186598231-186598231 on Build GRCh38
rs181531937 Chr.4: 186598265-186598265 on Build GRCh38
rs999664921 Chr.4: 186597923-186597923 on Build GRCh38
rs375987791 Chr.4: 186598161-186598161 on Build GRCh38
rs772540738 Chr.4: 186598031-186598031 on Build GRCh38
rs762709565 Chr.4: 186598012-186598012 on Build GRCh38
rs564219664 Chr.4: 186598155-186598155 on Build GRCh38
rs1043728008 Chr.4: 186598357-186598357 on Build GRCh38
rs961687769 Chr.4: 186598020-186598020 on Build GRCh38
rs561788984 Chr.4: 186598076-186598076 on Build GRCh38
rs904874640 Chr.4: 186598217-186598217 on Build GRCh38
rs953505903 Chr.4: 186598345-186598345 on Build GRCh38
rs200661419 Chr.4: 186597968-186597968 on Build GRCh38
rs760293297 Chr.4: 186597964-186597964 on Build GRCh38
rs780055518 Chr.4: 186597932-186597934 on Build GRCh38
rs138084499 Chr.4: 186598021-186598021 on Build GRCh38
rs779197760 Chr.4: 186598112-186598112 on Build GRCh38
rs771493652 Chr.4: 186597932-186597932 on Build GRCh38
rs557619128 Chr.4: 186598253-186598253 on Build GRCh38
rs201130322 Chr.4: 186598044-186598044 on Build GRCh38
rs772225327 Chr.4: 186598224-186598224 on Build GRCh38
rs374812339 Chr.4: 186597992-186597992 on Build GRCh38
rs921954002 Chr.4: 186598088-186598088 on Build GRCh38
rs376155526 Chr.4: 186597936-186597936 on Build GRCh38
rs150584542 Chr.4: 186598064-186598064 on Build GRCh38
rs143744017 Chr.4: 186598043-186598043 on Build GRCh38
rs773113119 Chr.4: 186598047-186598047 on Build GRCh38
rs935535855 Chr.4: 186598162-186598162 on Build GRCh38
rs200611830 Chr.4: 186598072-186598072 on Build GRCh38
rs905458117 Chr.4: 186598091-186598091 on Build GRCh38
rs1280097 Chr.4: 186598052-186598052 on Build GRCh38
rs188747847 Chr.4: 186598178-186598178 on Build GRCh38
rs749414274 Chr.4: 186597929-186597929 on Build GRCh38
rs747406113 Chr.4: 186598037-186598037 on Build GRCh38
rs767384858 Chr.4: 186598164-186598164 on Build GRCh38
rs775938826 Chr.4: 186597975-186597975 on Build GRCh38
rs180899930 Chr.4: 186597926-186597926 on Build GRCh38
rs751682765 Chr.4: 186598094-186598095 on Build GRCh38
rs748797862 Chr.4: 186598131-186598131 on Build GRCh38
rs769337342 Chr.4: 186598045-186598045 on Build GRCh38
rs890632292 Chr.4: 186597919-186597919 on Build GRCh38
rs776839670 Chr.4: 186598298-186598298 on Build GRCh38
rs1027300703 Chr.4: 186598193-186598193 on Build GRCh38
rs187068229 Chr.4: 186598284-186598284 on Build GRCh38
rs774367320 Chr.4: 186598065-186598065 on Build GRCh38
rs953287921 Chr.4: 186598214-186598214 on Build GRCh38
rs765147751 Chr.4: 186597991-186597991 on Build GRCh38
rs757379580 Chr.4: 186598071-186598071 on Build GRCh38
rs905176609 Chr.4: 186598361-186598361 on Build GRCh38
rs780680388 Chr.4: 186598103-186598103 on Build GRCh38
rs988275444 Chr.4: 186598262-186598262 on Build GRCh38

View Full Product Details