Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 14304360-14304857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIO
Entrez Gene ID
7204
Gene Name
trio Rho guanine nucleotide exchange factor
Gene Aliases
ARHGEF23, tgat
UniGene
Hs.130031
Gene Chromosome Location
Chr.5: 14143350-14510204 on Build GRCh38
Transcripts
NM_007118.3 NR_134469.1 XM_011514107.2 XM_011514108.1 XM_011514109.2 XM_011514110.2 XM_017009801.1 XM_017009802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3661557 c.1482C>T p.I494I Chr.5: 14304574-14304574 on Build GRCh38
7124413 c.1486C>G p.L496V Chr.5: 14304578-14304578 on Build GRCh38
3661556 c.1418T>C p.L473P Chr.5: 14304510-14304510 on Build GRCh38
5732338 c.1448C>A p.A483D Chr.5: 14304540-14304540 on Build GRCh38
4961857 c.1482C>A p.I494I Chr.5: 14304574-14304574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779169021 Chr.5: 14304442-14304443 on Build GRCh38
rs910199562 Chr.5: 14304726-14304726 on Build GRCh38
rs572350585 Chr.5: 14304689-14304689 on Build GRCh38
rs548481562 Chr.5: 14304808-14304808 on Build GRCh38
rs1031734850 Chr.5: 14304499-14304499 on Build GRCh38
rs776654412 Chr.5: 14304456-14304456 on Build GRCh38
rs746220040 Chr.5: 14304411-14304411 on Build GRCh38
rs954143041 Chr.5: 14304826-14304826 on Build GRCh38
rs961815437 Chr.5: 14304651-14304651 on Build GRCh38
rs915415546 Chr.5: 14304647-14304647 on Build GRCh38
rs551894535 Chr.5: 14304803-14304803 on Build GRCh38
rs758886933 Chr.5: 14304415-14304415 on Build GRCh38
rs984762016 Chr.5: 14304615-14304615 on Build GRCh38
rs910165776 Chr.5: 14304620-14304620 on Build GRCh38
rs139674608 Chr.5: 14304512-14304512 on Build GRCh38
rs115116719 Chr.5: 14304704-14304704 on Build GRCh38
rs758660684 Chr.5: 14304646-14304647 on Build GRCh38
rs780607003 Chr.5: 14304430-14304430 on Build GRCh38
rs999058855 Chr.5: 14304768-14304768 on Build GRCh38
rs763790629 Chr.5: 14304582-14304582 on Build GRCh38
rs556514343 Chr.5: 14304661-14304661 on Build GRCh38
rs745897325 Chr.5: 14304598-14304599 on Build GRCh38
rs985913124 Chr.5: 14304643-14304643 on Build GRCh38
rs768746522 Chr.5: 14304441-14304441 on Build GRCh38
rs771815030 Chr.5: 14304670-14304671 on Build GRCh38
rs562814256 Chr.5: 14304743-14304743 on Build GRCh38
rs34705730 Chr.5: 14304587-14304588 on Build GRCh38
rs567502037 Chr.5: 14304707-14304707 on Build GRCh38
rs758803105 Chr.5: 14304599-14304599 on Build GRCh38
rs541624073 Chr.5: 14304667-14304667 on Build GRCh38
rs780232464 Chr.5: 14304610-14304610 on Build GRCh38
rs560783610 Chr.5: 14304809-14304809 on Build GRCh38
rs762800062 Chr.5: 14304497-14304497 on Build GRCh38
rs998709797 Chr.5: 14304465-14304465 on Build GRCh38
rs527968742 Chr.5: 14304821-14304821 on Build GRCh38
rs202241476 Chr.5: 14304614-14304614 on Build GRCh38
rs749344768 Chr.5: 14304408-14304408 on Build GRCh38
rs769978944 Chr.5: 14304473-14304473 on Build GRCh38
rs773484658 Chr.5: 14304636-14304636 on Build GRCh38
rs755881570 Chr.5: 14304576-14304576 on Build GRCh38
rs759501399 Chr.5: 14304559-14304559 on Build GRCh38
rs750820152 Chr.5: 14304587-14304587 on Build GRCh38
rs372683 Chr.5: 14304757-14304757 on Build GRCh38
rs960116615 Chr.5: 14304529-14304529 on Build GRCh38
rs749416980 Chr.5: 14304642-14304642 on Build GRCh38
rs1014767267 Chr.5: 14304496-14304496 on Build GRCh38
rs1004657893 Chr.5: 14304745-14304745 on Build GRCh38
rs766016794 Chr.5: 14304513-14304513 on Build GRCh38
rs772934278 Chr.5: 14304486-14304486 on Build GRCh38
rs747416894 Chr.5: 14304433-14304433 on Build GRCh38
rs777551741 Chr.5: 14304569-14304569 on Build GRCh38
rs757422994 Chr.5: 14304462-14304462 on Build GRCh38
rs1022965266 Chr.5: 14304608-14304608 on Build GRCh38
rs544597276 Chr.5: 14304715-14304715 on Build GRCh38
rs1052950191 Chr.5: 14304793-14304793 on Build GRCh38
rs748211102 Chr.5: 14304618-14304618 on Build GRCh38
rs894771360 Chr.5: 14304674-14304674 on Build GRCh38
rs901764703 Chr.5: 14304760-14304760 on Build GRCh38
rs770065221 Chr.5: 14304624-14304624 on Build GRCh38
rs532511526 Chr.5: 14304712-14304712 on Build GRCh38
rs553965345 Chr.5: 14304678-14304678 on Build GRCh38
rs939040185 Chr.5: 14304661-14304661 on Build GRCh38
rs767169936 Chr.5: 14304560-14304560 on Build GRCh38
rs1043334624 Chr.5: 14304742-14304742 on Build GRCh38
rs5866099 Chr.5: 14304647-14304648 on Build GRCh38
rs961385688 Chr.5: 14304541-14304541 on Build GRCh38
rs781295961 Chr.5: 14304616-14304616 on Build GRCh38
rs752225450 Chr.5: 14304573-14304573 on Build GRCh38
rs943076724 Chr.5: 14304741-14304741 on Build GRCh38
rs774362448 Chr.5: 14304537-14304537 on Build GRCh38
rs574493985 Chr.5: 14304646-14304646 on Build GRCh38
rs146347032 Chr.5: 14304498-14304498 on Build GRCh38
rs748407607 Chr.5: 14304468-14304468 on Build GRCh38

View Full Product Details