Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33453097-33453616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TARS
Entrez Gene ID
6897
Gene Name
threonyl-tRNA synthetase
Gene Aliases
ThrRS
UniGene
Hs.481860
Gene Chromosome Location
Chr.5: 33440696-33468091 on Build GRCh38
Transcripts
NM_001258437.1 NM_001258438.1 NM_152295.4 NR_047676.1 NR_047677.1 NR_047678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6300534 c.442G>T p.E148* Chr.5: 33453401-33453401 on Build GRCh38
1067342 c.383A>G p.D128G Chr.5: 33453342-33453342 on Build GRCh38
5996124 c.370A>G p.N124D Chr.5: 33453329-33453329 on Build GRCh38
5344927 c.371A>T p.N124I Chr.5: 33453330-33453330 on Build GRCh38
185937 c.392G>A p.R131H Chr.5: 33453351-33453351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767290886 Chr.5: 33453351-33453351 on Build GRCh38
rs573320172 Chr.5: 33453183-33453183 on Build GRCh38
rs373876089 Chr.5: 33453251-33453251 on Build GRCh38
rs142292628 Chr.5: 33453193-33453193 on Build GRCh38
rs750064653 Chr.5: 33453286-33453286 on Build GRCh38
rs368011320 Chr.5: 33453252-33453252 on Build GRCh38
rs989032752 Chr.5: 33453285-33453285 on Build GRCh38
rs898309371 Chr.5: 33453550-33453550 on Build GRCh38
rs570788981 Chr.5: 33453170-33453170 on Build GRCh38
rs762573943 Chr.5: 33453285-33453286 on Build GRCh38
rs758973733 Chr.5: 33453300-33453300 on Build GRCh38
rs367864007 Chr.5: 33453295-33453295 on Build GRCh38
rs868402981 Chr.5: 33453589-33453589 on Build GRCh38
rs775441141 Chr.5: 33453264-33453266 on Build GRCh38
rs145788981 Chr.5: 33453302-33453302 on Build GRCh38
rs746332564 Chr.5: 33453240-33453240 on Build GRCh38
rs760541363 Chr.5: 33453264-33453265 on Build GRCh38
rs970153040 Chr.5: 33453576-33453576 on Build GRCh38
rs749380627 Chr.5: 33453430-33453430 on Build GRCh38
rs569001039 Chr.5: 33453395-33453395 on Build GRCh38
rs759344160 Chr.5: 33453267-33453267 on Build GRCh38
rs200074421 Chr.5: 33453262-33453263 on Build GRCh38
rs759139113 Chr.5: 33453402-33453402 on Build GRCh38
rs942431335 Chr.5: 33453549-33453549 on Build GRCh38
rs202220873 Chr.5: 33453436-33453436 on Build GRCh38
rs773407350 Chr.5: 33453316-33453316 on Build GRCh38
rs755358781 Chr.5: 33453410-33453410 on Build GRCh38
rs780528077 Chr.5: 33453301-33453301 on Build GRCh38
rs752351029 Chr.5: 33453287-33453287 on Build GRCh38
rs372038429 Chr.5: 33453260-33453260 on Build GRCh38
rs776344762 Chr.5: 33453263-33453264 on Build GRCh38
rs763653176 Chr.5: 33453366-33453366 on Build GRCh38
rs1023039637 Chr.5: 33453565-33453565 on Build GRCh38
rs763111369 Chr.5: 33453319-33453319 on Build GRCh38
rs35931457 Chr.5: 33453285-33453285 on Build GRCh38
rs112663402 Chr.5: 33453531-33453531 on Build GRCh38
rs748775731 Chr.5: 33453167-33453167 on Build GRCh38
rs766413784 Chr.5: 33453265-33453265 on Build GRCh38
rs890803196 Chr.5: 33453493-33453493 on Build GRCh38
rs772768525 Chr.5: 33453149-33453149 on Build GRCh38
rs752407851 Chr.5: 33453353-33453353 on Build GRCh38
rs536871787 Chr.5: 33453125-33453125 on Build GRCh38
rs1006419294 Chr.5: 33453253-33453253 on Build GRCh38
rs781302514 Chr.5: 33453310-33453310 on Build GRCh38
rs773046311 Chr.5: 33453262-33453262 on Build GRCh38
rs995773834 Chr.5: 33453564-33453564 on Build GRCh38
rs761661848 Chr.5: 33453264-33453267 on Build GRCh38
rs774448715 Chr.5: 33453450-33453450 on Build GRCh38
rs1016459223 Chr.5: 33453255-33453255 on Build GRCh38
rs936488118 Chr.5: 33453497-33453497 on Build GRCh38
rs182746823 Chr.5: 33453528-33453528 on Build GRCh38
rs989247291 Chr.5: 33453527-33453527 on Build GRCh38
rs771876897 Chr.5: 33453461-33453461 on Build GRCh38
rs769876534 Chr.5: 33453315-33453315 on Build GRCh38
rs566392744 Chr.5: 33453292-33453292 on Build GRCh38
rs371779976 Chr.5: 33453425-33453425 on Build GRCh38
rs556193539 Chr.5: 33453183-33453184 on Build GRCh38
rs375341932 Chr.5: 33453438-33453438 on Build GRCh38
rs745846263 Chr.5: 33453451-33453451 on Build GRCh38
rs755597091 Chr.5: 33453288-33453288 on Build GRCh38
rs780546165 Chr.5: 33453401-33453401 on Build GRCh38
rs190048471 Chr.5: 33453137-33453137 on Build GRCh38
rs777799549 Chr.5: 33453429-33453429 on Build GRCh38
rs538061732 Chr.5: 33453490-33453490 on Build GRCh38
rs772222439 Chr.5: 33453136-33453138 on Build GRCh38
rs577990514 Chr.5: 33453559-33453559 on Build GRCh38
rs1040683558 Chr.5: 33453192-33453193 on Build GRCh38
rs753448647 Chr.5: 33453371-33453371 on Build GRCh38
rs772549806 Chr.5: 33453242-33453242 on Build GRCh38
rs767280308 Chr.5: 33453286-33453286 on Build GRCh38
rs962875535 Chr.5: 33453182-33453182 on Build GRCh38
rs759432043 Chr.5: 33453340-33453340 on Build GRCh38
rs954761409 Chr.5: 33453290-33453290 on Build GRCh38
rs756701557 Chr.5: 33453391-33453391 on Build GRCh38
rs771003139 Chr.5: 33453320-33453320 on Build GRCh38
rs779315898 Chr.5: 33453239-33453239 on Build GRCh38
rs774360891 Chr.5: 33453336-33453336 on Build GRCh38
rs746197594 Chr.5: 33453264-33453264 on Build GRCh38
rs11541416 Chr.5: 33453311-33453311 on Build GRCh38
rs146234110 Chr.5: 33453197-33453197 on Build GRCh38
rs900833522 Chr.5: 33453583-33453583 on Build GRCh38
rs760419283 Chr.5: 33453354-33453354 on Build GRCh38
rs751456672 Chr.5: 33453266-33453266 on Build GRCh38
rs780440188 Chr.5: 33453250-33453250 on Build GRCh38
rs769878313 Chr.5: 33453261-33453261 on Build GRCh38
rs111754118 Chr.5: 33453264-33453264 on Build GRCh38
rs970016863 Chr.5: 33453231-33453231 on Build GRCh38
rs540226740 Chr.5: 33453191-33453191 on Build GRCh38
rs534126094 Chr.5: 33453580-33453580 on Build GRCh38
rs1015943512 Chr.5: 33453541-33453541 on Build GRCh38
rs776856951 Chr.5: 33453256-33453256 on Build GRCh38
rs768944528 Chr.5: 33453308-33453308 on Build GRCh38

View Full Product Details