Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38475523-38476066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIFR
Entrez Gene ID
3977
Gene Name
leukemia inhibitory factor receptor alpha
Gene Aliases
CD118, LIF-R, SJS2, STWS, SWS
UniGene
Hs.133421
Gene Chromosome Location
Chr.5: 38474963-38606290 on Build GRCh38
Transcripts
NM_001127671.1 NM_002310.5 XM_017009462.1

SNP Detail

SNP ID Chromosome Location
rs191019863 Chr.5: 38475864-38475864 on Build GRCh38
rs571064663 Chr.5: 38475670-38475670 on Build GRCh38
rs914580137 Chr.5: 38475720-38475720 on Build GRCh38
rs768004410 Chr.5: 38475896-38475896 on Build GRCh38
rs566540398 Chr.5: 38475861-38475861 on Build GRCh38
rs967119249 Chr.5: 38475609-38475609 on Build GRCh38
rs942793434 Chr.5: 38475688-38475688 on Build GRCh38
rs542021288 Chr.5: 38475553-38475554 on Build GRCh38
rs886060605 Chr.5: 38475799-38475799 on Build GRCh38
rs34965361 Chr.5: 38475585-38475586 on Build GRCh38
rs962404965 Chr.5: 38475584-38475588 on Build GRCh38
rs550842280 Chr.5: 38475728-38475728 on Build GRCh38
rs886060604 Chr.5: 38475609-38475609 on Build GRCh38
rs538503092 Chr.5: 38475658-38475658 on Build GRCh38
rs556240742 Chr.5: 38475802-38475802 on Build GRCh38
rs991831481 Chr.5: 38475761-38475761 on Build GRCh38
rs536207682 Chr.5: 38475854-38475854 on Build GRCh38
rs558399671 Chr.5: 38475914-38475914 on Build GRCh38
rs1038352546 Chr.5: 38475799-38475799 on Build GRCh38
rs1008367730 Chr.5: 38475883-38475883 on Build GRCh38
rs569182863 Chr.5: 38475773-38475773 on Build GRCh38
rs989820767 Chr.5: 38475577-38475577 on Build GRCh38
rs551184868 Chr.5: 38475651-38475652 on Build GRCh38
rs930052685 Chr.5: 38475827-38475827 on Build GRCh38
rs376744831 Chr.5: 38475618-38475621 on Build GRCh38
rs1025884057 Chr.5: 38475600-38475600 on Build GRCh38
rs1011698764 Chr.5: 38475943-38475943 on Build GRCh38
rs554626836 Chr.5: 38475856-38475856 on Build GRCh38
rs912532145 Chr.5: 38476010-38476010 on Build GRCh38
rs149932478 Chr.5: 38475961-38475961 on Build GRCh38
rs760051833 Chr.5: 38475859-38475859 on Build GRCh38
rs886060606 Chr.5: 38475827-38475827 on Build GRCh38
rs955614984 Chr.5: 38475932-38475935 on Build GRCh38
rs948076814 Chr.5: 38475786-38475786 on Build GRCh38
rs964189582 Chr.5: 38476001-38476001 on Build GRCh38
rs977288362 Chr.5: 38475611-38475611 on Build GRCh38
rs998610352 Chr.5: 38476041-38476041 on Build GRCh38
rs1021409519 Chr.5: 38475608-38475608 on Build GRCh38
rs3822425 Chr.5: 38475925-38475925 on Build GRCh38
rs1035416865 Chr.5: 38476042-38476042 on Build GRCh38
rs375363202 Chr.5: 38475979-38475979 on Build GRCh38
rs1016623024 Chr.5: 38475881-38475881 on Build GRCh38
rs973420869 Chr.5: 38475585-38475585 on Build GRCh38
rs1011876276 Chr.5: 38475571-38475571 on Build GRCh38
rs986926057 Chr.5: 38475655-38475656 on Build GRCh38
rs939185069 Chr.5: 38475820-38475820 on Build GRCh38
rs1026318279 Chr.5: 38475928-38475928 on Build GRCh38
rs369004224 Chr.5: 38475734-38475734 on Build GRCh38
rs886060607 Chr.5: 38475928-38475928 on Build GRCh38
rs1011992574 Chr.5: 38475862-38475862 on Build GRCh38
rs1042709820 Chr.5: 38475746-38475746 on Build GRCh38
rs928463083 Chr.5: 38475612-38475612 on Build GRCh38
rs946055402 Chr.5: 38475741-38475741 on Build GRCh38
rs990230466 Chr.5: 38475694-38475694 on Build GRCh38

View Full Product Details