Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38480699-38481198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIFR
Entrez Gene ID
3977
Gene Name
leukemia inhibitory factor receptor alpha
Gene Aliases
CD118, LIF-R, SJS2, STWS, SWS
UniGene
Hs.133421
Gene Chromosome Location
Chr.5: 38474963-38606290 on Build GRCh38
Transcripts
NM_001127671.1 NM_002310.5 XM_011514040.2 XM_011514042.2 XM_017009462.1 XM_017009463.1

SNP Detail

SNP ID Chromosome Location
rs186664100 Chr.5: 38480858-38480858 on Build GRCh38
rs1017327410 Chr.5: 38481151-38481151 on Build GRCh38
rs116625329 Chr.5: 38481169-38481169 on Build GRCh38
rs751619532 Chr.5: 38480783-38480783 on Build GRCh38
rs528435493 Chr.5: 38481090-38481090 on Build GRCh38
rs544807588 Chr.5: 38480985-38480985 on Build GRCh38
rs770527327 Chr.5: 38481041-38481041 on Build GRCh38
rs1016159112 Chr.5: 38480731-38480731 on Build GRCh38
rs546989449 Chr.5: 38481099-38481099 on Build GRCh38
rs755894698 Chr.5: 38480865-38480865 on Build GRCh38
rs754981305 Chr.5: 38480811-38480811 on Build GRCh38
rs774127787 Chr.5: 38480868-38480871 on Build GRCh38
rs1021549861 Chr.5: 38480917-38480917 on Build GRCh38
rs543297073 Chr.5: 38480782-38480782 on Build GRCh38
rs972469393 Chr.5: 38481015-38481015 on Build GRCh38
rs549697077 Chr.5: 38481040-38481040 on Build GRCh38
rs781748205 Chr.5: 38480873-38480873 on Build GRCh38
rs909668342 Chr.5: 38480997-38480997 on Build GRCh38
rs1007554825 Chr.5: 38480831-38480831 on Build GRCh38
rs961890704 Chr.5: 38480747-38480747 on Build GRCh38
rs541023079 Chr.5: 38480899-38480899 on Build GRCh38
rs982790262 Chr.5: 38480947-38480947 on Build GRCh38
rs1021014288 Chr.5: 38481157-38481157 on Build GRCh38
rs993407707 Chr.5: 38481045-38481045 on Build GRCh38
rs2561819 Chr.5: 38481009-38481009 on Build GRCh38
rs893778990 Chr.5: 38481109-38481109 on Build GRCh38
rs376245959 Chr.5: 38480894-38480894 on Build GRCh38
rs1007751946 Chr.5: 38481116-38481116 on Build GRCh38
rs897711684 Chr.5: 38481042-38481042 on Build GRCh38
rs889691332 Chr.5: 38481078-38481078 on Build GRCh38
rs754726326 Chr.5: 38480921-38480921 on Build GRCh38
rs943391630 Chr.5: 38481095-38481095 on Build GRCh38
rs184101529 Chr.5: 38481156-38481156 on Build GRCh38
rs3822427 Chr.5: 38481049-38481049 on Build GRCh38
rs116040046 Chr.5: 38480982-38480982 on Build GRCh38
rs1039047142 Chr.5: 38481104-38481104 on Build GRCh38
rs938116389 Chr.5: 38480938-38480938 on Build GRCh38
rs756472947 Chr.5: 38480986-38480986 on Build GRCh38
rs193019517 Chr.5: 38480994-38480994 on Build GRCh38
rs555282081 Chr.5: 38480828-38480828 on Build GRCh38
rs941235840 Chr.5: 38480743-38480743 on Build GRCh38
rs778051771 Chr.5: 38480990-38480990 on Build GRCh38
rs993291701 Chr.5: 38480762-38480762 on Build GRCh38
rs933809480 Chr.5: 38481048-38481048 on Build GRCh38
rs1011411405 Chr.5: 38481152-38481152 on Build GRCh38
rs1020566455 Chr.5: 38481173-38481173 on Build GRCh38
rs886060624 Chr.5: 38480947-38480947 on Build GRCh38
rs966864203 Chr.5: 38480920-38480920 on Build GRCh38
rs748590225 Chr.5: 38480941-38480941 on Build GRCh38
rs1037264075 Chr.5: 38480744-38480744 on Build GRCh38
rs896951008 Chr.5: 38480759-38480759 on Build GRCh38
rs570512971 Chr.5: 38480936-38480936 on Build GRCh38
rs1050997871 Chr.5: 38480815-38480815 on Build GRCh38

View Full Product Details