Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38846124-38846621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSMR
Entrez Gene ID
9180
Gene Name
oncostatin M receptor
Gene Aliases
IL-31R-beta, IL-31RB, OSMRB, PLCA1
UniGene
Hs.120658
Gene Chromosome Location
Chr.5: 38845858-38945579 on Build GRCh38
Transcripts
NM_001168355.2 NM_001323504.1 NM_001323505.1 NM_001323506.1 NM_001323507.1 NM_003999.2 XM_005248384.1 XM_005248386.2 XM_005248387.2 XM_011514161.2 XM_017010019.1

SNP Detail

SNP ID Chromosome Location
rs1032123262 Chr.5: 38846160-38846160 on Build GRCh38
rs574339421 Chr.5: 38846254-38846254 on Build GRCh38
rs970252816 Chr.5: 38846188-38846188 on Build GRCh38
rs935231198 Chr.5: 38846246-38846246 on Build GRCh38
rs1004577317 Chr.5: 38846147-38846147 on Build GRCh38
rs537699813 Chr.5: 38846206-38846206 on Build GRCh38
rs572163338 Chr.5: 38846377-38846377 on Build GRCh38
rs781286372 Chr.5: 38846503-38846504 on Build GRCh38
rs1053077650 Chr.5: 38846350-38846350 on Build GRCh38
rs558318060 Chr.5: 38846165-38846165 on Build GRCh38
rs1047467242 Chr.5: 38846233-38846233 on Build GRCh38
rs996072130 Chr.5: 38846364-38846364 on Build GRCh38
rs77590691 Chr.5: 38846489-38846489 on Build GRCh38
rs953152845 Chr.5: 38846518-38846518 on Build GRCh38
rs975792435 Chr.5: 38846591-38846591 on Build GRCh38
rs983632648 Chr.5: 38846525-38846525 on Build GRCh38
rs900434011 Chr.5: 38846353-38846353 on Build GRCh38
rs184280395 Chr.5: 38846443-38846443 on Build GRCh38
rs560635484 Chr.5: 38846505-38846507 on Build GRCh38
rs970285347 Chr.5: 38846481-38846481 on Build GRCh38
rs956634852 Chr.5: 38846593-38846593 on Build GRCh38
rs980255403 Chr.5: 38846204-38846204 on Build GRCh38
rs192375340 Chr.5: 38846373-38846373 on Build GRCh38
rs917492268 Chr.5: 38846181-38846181 on Build GRCh38
rs546429542 Chr.5: 38846383-38846383 on Build GRCh38
rs753299650 Chr.5: 38846418-38846418 on Build GRCh38
rs925105123 Chr.5: 38846581-38846581 on Build GRCh38
rs952212709 Chr.5: 38846503-38846503 on Build GRCh38
rs944426240 Chr.5: 38846545-38846545 on Build GRCh38
rs979697041 Chr.5: 38846560-38846560 on Build GRCh38
rs1014587775 Chr.5: 38846148-38846148 on Build GRCh38
rs907574100 Chr.5: 38846260-38846260 on Build GRCh38
rs529314245 Chr.5: 38846533-38846533 on Build GRCh38
rs979281543 Chr.5: 38846212-38846212 on Build GRCh38
rs77150157 Chr.5: 38846230-38846230 on Build GRCh38
rs1049212281 Chr.5: 38846424-38846424 on Build GRCh38
rs944372111 Chr.5: 38846281-38846281 on Build GRCh38
rs188144236 Chr.5: 38846341-38846341 on Build GRCh38
rs1043582823 Chr.5: 38846366-38846366 on Build GRCh38
rs184876382 Chr.5: 38846173-38846173 on Build GRCh38
rs879736252 Chr.5: 38846541-38846541 on Build GRCh38
rs906037749 Chr.5: 38846403-38846403 on Build GRCh38
rs2292018 Chr.5: 38846452-38846452 on Build GRCh38
rs920797481 Chr.5: 38846493-38846493 on Build GRCh38
rs529408408 Chr.5: 38846429-38846429 on Build GRCh38
rs79488292 Chr.5: 38846478-38846478 on Build GRCh38
rs372499997 Chr.5: 38846438-38846438 on Build GRCh38

View Full Product Details