Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 53603270-53603796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFS4
Entrez Gene ID
4724
Gene Name
NADH:ubiquinone oxidoreductase subunit S4
Gene Aliases
AQDQ, CI-18, CI-18 kDa, CI-AQDQ
UniGene
Hs.528222
Gene Chromosome Location
Chr.5: 53560610-53683341 on Build GRCh38
Transcripts
NM_001318051.1 NM_002495.3 NR_134473.1 NR_134474.1 NR_134475.1 XM_017009491.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7379251 c.160A>G p.T54A Chr.5: 53603513-53603513 on Build GRCh38
5344952 c.164T>A p.V55D Chr.5: 53603517-53603517 on Build GRCh38
7156550 c.136C>A p.Q46K Chr.5: 53603489-53603489 on Build GRCh38
482874 c.155T>C p.L52P Chr.5: 53603508-53603508 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs987556423 Chr.5: 53603634-53603634 on Build GRCh38
rs202015682 Chr.5: 53603352-53603352 on Build GRCh38
rs921685455 Chr.5: 53603599-53603599 on Build GRCh38
rs142368721 Chr.5: 53603503-53603503 on Build GRCh38
rs863224108 Chr.5: 53603496-53603496 on Build GRCh38
rs756821245 Chr.5: 53603513-53603513 on Build GRCh38
rs878941723 Chr.5: 53603337-53603337 on Build GRCh38
rs761939656 Chr.5: 53603408-53603408 on Build GRCh38
rs532142629 Chr.5: 53603440-53603440 on Build GRCh38
rs914900719 Chr.5: 53603768-53603768 on Build GRCh38
rs963118375 Chr.5: 53603595-53603595 on Build GRCh38
rs148737893 Chr.5: 53603470-53603470 on Build GRCh38
rs912801201 Chr.5: 53603655-53603655 on Build GRCh38
rs932951532 Chr.5: 53603625-53603625 on Build GRCh38
rs376281345 Chr.5: 53603451-53603451 on Build GRCh38
rs553670267 Chr.5: 53603707-53603707 on Build GRCh38
rs932982574 Chr.5: 53603367-53603367 on Build GRCh38
rs557636203 Chr.5: 53603750-53603750 on Build GRCh38
rs750909425 Chr.5: 53603410-53603410 on Build GRCh38
rs11414085 Chr.5: 53603349-53603350 on Build GRCh38
rs751196164 Chr.5: 53603512-53603512 on Build GRCh38
rs768406660 Chr.5: 53603551-53603551 on Build GRCh38
rs760818714 Chr.5: 53603497-53603497 on Build GRCh38
rs932638487 Chr.5: 53603326-53603326 on Build GRCh38
rs774622848 Chr.5: 53603405-53603405 on Build GRCh38
rs749272992 Chr.5: 53603550-53603550 on Build GRCh38
rs767012684 Chr.5: 53603429-53603429 on Build GRCh38
rs934932956 Chr.5: 53603665-53603665 on Build GRCh38
rs34241409 Chr.5: 53603351-53603352 on Build GRCh38
rs200926524 Chr.5: 53603452-53603452 on Build GRCh38
rs772784623 Chr.5: 53603454-53603454 on Build GRCh38
rs373996902 Chr.5: 53603413-53603413 on Build GRCh38
rs190363728 Chr.5: 53603299-53603299 on Build GRCh38
rs151297702 Chr.5: 53603511-53603511 on Build GRCh38
rs370695630 Chr.5: 53603477-53603477 on Build GRCh38
rs373429283 Chr.5: 53603334-53603336 on Build GRCh38
rs754465699 Chr.5: 53603504-53603504 on Build GRCh38
rs760828666 Chr.5: 53603582-53603582 on Build GRCh38
rs756535726 Chr.5: 53603424-53603424 on Build GRCh38
rs1040260438 Chr.5: 53603524-53603524 on Build GRCh38
rs1053288812 Chr.5: 53603691-53603691 on Build GRCh38
rs138941073 Chr.5: 53603455-53603455 on Build GRCh38
rs754193714 Chr.5: 53603435-53603435 on Build GRCh38
rs746701465 Chr.5: 53603450-53603450 on Build GRCh38
rs1038700066 Chr.5: 53603331-53603331 on Build GRCh38
rs941786478 Chr.5: 53603318-53603318 on Build GRCh38
rs974457367 Chr.5: 53603597-53603597 on Build GRCh38
rs773114153 Chr.5: 53603488-53603488 on Build GRCh38
rs201151224 Chr.5: 53603556-53603556 on Build GRCh38
rs1051317249 Chr.5: 53603384-53603384 on Build GRCh38
rs376119394 Chr.5: 53603335-53603336 on Build GRCh38
rs371577393 Chr.5: 53603336-53603337 on Build GRCh38
rs747994192 Chr.5: 53603565-53603565 on Build GRCh38
rs1003288416 Chr.5: 53603474-53603474 on Build GRCh38
rs535041698 Chr.5: 53603705-53603705 on Build GRCh38
rs905836637 Chr.5: 53603464-53603464 on Build GRCh38
rs773483033 Chr.5: 53603574-53603574 on Build GRCh38
rs78541239 Chr.5: 53603353-53603353 on Build GRCh38
rs755386101 Chr.5: 53603506-53603506 on Build GRCh38
rs938268728 Chr.5: 53603391-53603391 on Build GRCh38
rs189714605 Chr.5: 53603740-53603740 on Build GRCh38
rs75151684 Chr.5: 53603351-53603351 on Build GRCh38
rs1022912416 Chr.5: 53603460-53603460 on Build GRCh38
rs777567835 Chr.5: 53603449-53603449 on Build GRCh38
rs961560008 Chr.5: 53603308-53603308 on Build GRCh38
rs991493479 Chr.5: 53603760-53603760 on Build GRCh38
rs749077486 Chr.5: 53603466-53603466 on Build GRCh38
rs11548867 Chr.5: 53603468-53603468 on Build GRCh38
rs368854518 Chr.5: 53603515-53603515 on Build GRCh38
rs184907822 Chr.5: 53603737-53603737 on Build GRCh38
rs143327974 Chr.5: 53603756-53603756 on Build GRCh38
rs1035188310 Chr.5: 53603725-53603725 on Build GRCh38
rs537190985 Chr.5: 53603521-53603521 on Build GRCh38
rs866475889 Chr.5: 53603728-53603728 on Build GRCh38
rs547401888 Chr.5: 53603297-53603301 on Build GRCh38
rs749845171 Chr.5: 53603520-53603520 on Build GRCh38
rs982356058 Chr.5: 53603343-53603344 on Build GRCh38
rs200789732 Chr.5: 53603544-53603544 on Build GRCh38
rs374180161 Chr.5: 53603484-53603484 on Build GRCh38
rs182554293 Chr.5: 53603703-53603703 on Build GRCh38
rs200060526 Chr.5: 53603476-53603476 on Build GRCh38
rs891392256 Chr.5: 53603395-53603395 on Build GRCh38
rs772124334 Chr.5: 53603567-53603567 on Build GRCh38
rs1015725430 Chr.5: 53603669-53603669 on Build GRCh38

View Full Product Details