Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55852113-55852612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL31RA
Entrez Gene ID
133396
Gene Name
interleukin 31 receptor A
Gene Aliases
CRL, CRL3, GLM-R, GLMR, GPL, IL-31RA, PLCA2, PRO21384, hGLM-R
UniGene
Hs.55378
Gene Chromosome Location
Chr.5: 55840334-55922854 on Build GRCh38
Transcripts
NM_001242637.1 NM_001242638.1 NM_001297570.1 NM_001297572.1 NM_139017.5 XM_011543142.2

SNP Detail

SNP ID Chromosome Location
rs565519899 Chr.5: 55852456-55852456 on Build GRCh38
rs541565409 Chr.5: 55852494-55852494 on Build GRCh38
rs139183041 Chr.5: 55852461-55852461 on Build GRCh38
rs940639538 Chr.5: 55852304-55852311 on Build GRCh38
rs999360611 Chr.5: 55852273-55852273 on Build GRCh38
rs1019636153 Chr.5: 55852196-55852196 on Build GRCh38
rs893316443 Chr.5: 55852162-55852162 on Build GRCh38
rs1031218947 Chr.5: 55852294-55852294 on Build GRCh38
rs1055473657 Chr.5: 55852435-55852435 on Build GRCh38
rs985971510 Chr.5: 55852353-55852353 on Build GRCh38
rs779199553 Chr.5: 55852177-55852177 on Build GRCh38
rs1043903357 Chr.5: 55852504-55852504 on Build GRCh38
rs954031564 Chr.5: 55852337-55852337 on Build GRCh38
rs909965626 Chr.5: 55852359-55852359 on Build GRCh38
rs894787505 Chr.5: 55852216-55852216 on Build GRCh38
rs777048117 Chr.5: 55852215-55852215 on Build GRCh38
rs996888437 Chr.5: 55852560-55852560 on Build GRCh38
rs964042170 Chr.5: 55852542-55852542 on Build GRCh38
rs975502902 Chr.5: 55852209-55852209 on Build GRCh38
rs143355268 Chr.5: 55852140-55852140 on Build GRCh38
rs1045784180 Chr.5: 55852316-55852316 on Build GRCh38
rs867974613 Chr.5: 55852226-55852226 on Build GRCh38
rs964446542 Chr.5: 55852205-55852206 on Build GRCh38
rs987192806 Chr.5: 55852190-55852190 on Build GRCh38
rs1011914337 Chr.5: 55852497-55852497 on Build GRCh38
rs918717730 Chr.5: 55852447-55852447 on Build GRCh38
rs984655212 Chr.5: 55852251-55852251 on Build GRCh38
rs771307211 Chr.5: 55852204-55852204 on Build GRCh38
rs930459496 Chr.5: 55852211-55852211 on Build GRCh38
rs193133910 Chr.5: 55852463-55852463 on Build GRCh38
rs867830063 Chr.5: 55852464-55852464 on Build GRCh38
rs975949216 Chr.5: 55852299-55852299 on Build GRCh38
rs184569045 Chr.5: 55852516-55852516 on Build GRCh38
rs1034453550 Chr.5: 55852157-55852161 on Build GRCh38
rs1003761399 Chr.5: 55852181-55852181 on Build GRCh38
rs531787345 Chr.5: 55852261-55852261 on Build GRCh38
rs1030811635 Chr.5: 55852521-55852521 on Build GRCh38
rs529719147 Chr.5: 55852576-55852576 on Build GRCh38
rs922620060 Chr.5: 55852317-55852317 on Build GRCh38
rs1056143077 Chr.5: 55852210-55852210 on Build GRCh38
rs758944718 Chr.5: 55852272-55852272 on Build GRCh38
rs965937432 Chr.5: 55852295-55852295 on Build GRCh38
rs1037207925 Chr.5: 55852444-55852444 on Build GRCh38
rs903610325 Chr.5: 55852506-55852506 on Build GRCh38
rs965823258 Chr.5: 55852575-55852575 on Build GRCh38
rs747465139 Chr.5: 55852505-55852505 on Build GRCh38
rs753052361 Chr.5: 55852197-55852197 on Build GRCh38
rs907769736 Chr.5: 55852291-55852291 on Build GRCh38
rs531082492 Chr.5: 55852156-55852157 on Build GRCh38
rs894838222 Chr.5: 55852484-55852484 on Build GRCh38
rs549970097 Chr.5: 55852262-55852262 on Build GRCh38
rs9292100 Chr.5: 55852392-55852392 on Build GRCh38
rs906883148 Chr.5: 55852364-55852364 on Build GRCh38
rs1012032979 Chr.5: 55852243-55852243 on Build GRCh38
rs968327609 Chr.5: 55852263-55852263 on Build GRCh38
rs368330932 Chr.5: 55852223-55852223 on Build GRCh38

View Full Product Details