Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 56865638-56866134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K1
Entrez Gene ID
4214
Gene Name
mitogen-activated protein kinase kinase kinase 1
Gene Aliases
MAPKKK1, MEKK, MEKK 1, MEKK1, SRXY6
UniGene
Hs.653654
Gene Chromosome Location
Chr.5: 56815073-56896152 on Build GRCh38
Transcripts
NM_005921.1 XM_017009484.1 XM_017009485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6949963 c.1160_1161insTTTGT p.Q390fs*48 Chr.5: 56865836-56865837 on Build GRCh38
6929575 c.1229A>C p.K410T Chr.5: 56865905-56865905 on Build GRCh38
5029141 c.1214_1215insT p.N406fs*1 Chr.5: 56865890-56865891 on Build GRCh38
6707069 c.1301+2T>G p.? Chr.5: 56865979-56865979 on Build GRCh38
6914273 c.1186C>T p.R396C Chr.5: 56865862-56865862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005132606 Chr.5: 56865937-56865937 on Build GRCh38
rs753903548 Chr.5: 56865956-56865956 on Build GRCh38
rs898262264 Chr.5: 56865963-56865963 on Build GRCh38
rs371897325 Chr.5: 56865820-56865820 on Build GRCh38
rs755573286 Chr.5: 56865958-56865958 on Build GRCh38
rs758890766 Chr.5: 56865964-56865964 on Build GRCh38
rs776091964 Chr.5: 56865800-56865800 on Build GRCh38
rs929628890 Chr.5: 56865990-56865990 on Build GRCh38
rs913140705 Chr.5: 56865694-56865694 on Build GRCh38
rs780173928 Chr.5: 56865780-56865780 on Build GRCh38
rs1024569606 Chr.5: 56866077-56866077 on Build GRCh38
rs545050907 Chr.5: 56866008-56866008 on Build GRCh38
rs768591030 Chr.5: 56866013-56866013 on Build GRCh38
rs767233872 Chr.5: 56865806-56865806 on Build GRCh38
rs374852011 Chr.5: 56865782-56865782 on Build GRCh38
rs745548925 Chr.5: 56865994-56865994 on Build GRCh38
rs774326877 Chr.5: 56866022-56866022 on Build GRCh38
rs748761056 Chr.5: 56866012-56866012 on Build GRCh38
rs942550524 Chr.5: 56865936-56865936 on Build GRCh38
rs368525887 Chr.5: 56865673-56865673 on Build GRCh38
rs778197681 Chr.5: 56865971-56865971 on Build GRCh38
rs191214731 Chr.5: 56866011-56866011 on Build GRCh38
rs761182103 Chr.5: 56865687-56865689 on Build GRCh38
rs186794673 Chr.5: 56865739-56865739 on Build GRCh38
rs541706462 Chr.5: 56866065-56866065 on Build GRCh38
rs750675091 Chr.5: 56865916-56865916 on Build GRCh38
rs556103246 Chr.5: 56865829-56865829 on Build GRCh38
rs748482210 Chr.5: 56865795-56865795 on Build GRCh38
rs1020167899 Chr.5: 56866109-56866109 on Build GRCh38
rs761108519 Chr.5: 56866007-56866007 on Build GRCh38
rs368468756 Chr.5: 56865799-56865799 on Build GRCh38
rs751025307 Chr.5: 56865809-56865812 on Build GRCh38
rs553741923 Chr.5: 56866108-56866108 on Build GRCh38
rs761732807 Chr.5: 56866023-56866023 on Build GRCh38
rs762754628 Chr.5: 56865932-56865932 on Build GRCh38
rs832575 Chr.5: 56865960-56865960 on Build GRCh38
rs767326010 Chr.5: 56865870-56865870 on Build GRCh38
rs761534801 Chr.5: 56865854-56865854 on Build GRCh38
rs765810354 Chr.5: 56865803-56865804 on Build GRCh38
rs886602536 Chr.5: 56865868-56865868 on Build GRCh38
rs775038228 Chr.5: 56865823-56865823 on Build GRCh38
rs760916911 Chr.5: 56865925-56865925 on Build GRCh38
rs763973149 Chr.5: 56865843-56865843 on Build GRCh38
rs1024495024 Chr.5: 56865942-56865942 on Build GRCh38
rs1046737249 Chr.5: 56865999-56865999 on Build GRCh38
rs372700780 Chr.5: 56865961-56865961 on Build GRCh38
rs1039129610 Chr.5: 56865753-56865753 on Build GRCh38
rs779440158 Chr.5: 56865794-56865794 on Build GRCh38
rs957313347 Chr.5: 56866031-56866031 on Build GRCh38
rs376819928 Chr.5: 56865852-56865852 on Build GRCh38
rs745520989 Chr.5: 56865812-56865812 on Build GRCh38
rs944689675 Chr.5: 56865692-56865692 on Build GRCh38

View Full Product Details