Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65665595-65666146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGTB
Entrez Gene ID
54557
Gene Name
small glutamine rich tetratricopeptide repeat containing beta
Gene Aliases
SGT2
UniGene
Hs.482301
Gene Chromosome Location
Chr.5: 65665928-65722821 on Build GRCh38
Transcripts
NM_019072.2 XM_005248548.3 XM_017009597.1 NM_001093755.1 NM_001093756.1 NM_001243737.1 NM_024941.3 XM_011543652.2 XM_017009890.1

Gene Symbol
TRAPPC13
Entrez Gene ID
80006
Gene Name
trafficking protein particle complex 13
Gene Aliases
C5orf44
UniGene
Hs.591760
Gene Chromosome Location
Chr.5: 65624731-65666136 on Build GRCh38
Transcripts
NM_019072.2 XM_005248548.3 XM_017009597.1 NM_001093755.1 NM_001093756.1 NM_001243737.1 NM_024941.3 XM_011543652.2 XM_017009890.1

SNP Detail

SNP ID Chromosome Location
rs545248734 Chr.5: 65666075-65666078 on Build GRCh38
rs920803389 Chr.5: 65666003-65666003 on Build GRCh38
rs576285515 Chr.5: 65666095-65666095 on Build GRCh38
rs200999932 Chr.5: 65665923-65665923 on Build GRCh38
rs777006949 Chr.5: 65665775-65665775 on Build GRCh38
rs747230097 Chr.5: 65665714-65665714 on Build GRCh38
rs183897400 Chr.5: 65665757-65665757 on Build GRCh38
rs918446749 Chr.5: 65665814-65665814 on Build GRCh38
rs987138641 Chr.5: 65665832-65665832 on Build GRCh38
rs907954022 Chr.5: 65665708-65665708 on Build GRCh38
rs1023865808 Chr.5: 65665740-65665740 on Build GRCh38
rs1024316280 Chr.5: 65665967-65665970 on Build GRCh38
rs899310086 Chr.5: 65665737-65665737 on Build GRCh38
rs544284650 Chr.5: 65665874-65665874 on Build GRCh38
rs933302178 Chr.5: 65665667-65665667 on Build GRCh38
rs566173244 Chr.5: 65665693-65665693 on Build GRCh38
rs545878265 Chr.5: 65665715-65665715 on Build GRCh38
rs937633697 Chr.5: 65665741-65665741 on Build GRCh38
rs973468672 Chr.5: 65665987-65665987 on Build GRCh38
rs530614258 Chr.5: 65665854-65665854 on Build GRCh38
rs1029983312 Chr.5: 65665953-65665953 on Build GRCh38
rs1019867922 Chr.5: 65665818-65665818 on Build GRCh38
rs541988455 Chr.5: 65665797-65665797 on Build GRCh38
rs940775083 Chr.5: 65665977-65665977 on Build GRCh38
rs907849164 Chr.5: 65665967-65665967 on Build GRCh38
rs1043359665 Chr.5: 65665886-65665886 on Build GRCh38
rs987469929 Chr.5: 65665964-65665964 on Build GRCh38
rs530626659 Chr.5: 65666016-65666016 on Build GRCh38
rs933805707 Chr.5: 65665820-65665820 on Build GRCh38
rs1013732735 Chr.5: 65665960-65665960 on Build GRCh38
rs999696367 Chr.5: 65665696-65665696 on Build GRCh38
rs561912573 Chr.5: 65665810-65665810 on Build GRCh38
rs999872874 Chr.5: 65665922-65665922 on Build GRCh38
rs950296694 Chr.5: 65666037-65666037 on Build GRCh38
rs975383572 Chr.5: 65665948-65665948 on Build GRCh38
rs1030348617 Chr.5: 65666070-65666070 on Build GRCh38
rs752293382 Chr.5: 65665725-65665726 on Build GRCh38
rs372555825 Chr.5: 65666013-65666013 on Build GRCh38
rs557915959 Chr.5: 65665968-65665968 on Build GRCh38
rs866668296 Chr.5: 65665806-65665806 on Build GRCh38
rs564174995 Chr.5: 65665882-65665882 on Build GRCh38
rs546294989 Chr.5: 65665901-65665901 on Build GRCh38
rs902948810 Chr.5: 65665864-65665864 on Build GRCh38
rs1037888751 Chr.5: 65665719-65665719 on Build GRCh38
rs1016766630 Chr.5: 65665777-65665777 on Build GRCh38
rs954770256 Chr.5: 65665958-65665958 on Build GRCh38
rs773754576 Chr.5: 65666054-65666054 on Build GRCh38
rs528810648 Chr.5: 65665983-65665983 on Build GRCh38
rs984777112 Chr.5: 65665759-65665761 on Build GRCh38
rs548758469 Chr.5: 65666008-65666008 on Build GRCh38
rs188177989 Chr.5: 65665939-65665939 on Build GRCh38
rs896059013 Chr.5: 65666024-65666024 on Build GRCh38
rs532913735 Chr.5: 65665899-65665899 on Build GRCh38
rs371368873 Chr.5: 65665687-65665687 on Build GRCh38
rs62367742 Chr.5: 65665659-65665659 on Build GRCh38
rs563133502 Chr.5: 65665657-65665657 on Build GRCh38
rs902832675 Chr.5: 65666118-65666118 on Build GRCh38
rs1032824438 Chr.5: 65665932-65665932 on Build GRCh38
rs553202195 Chr.5: 65665752-65665752 on Build GRCh38
rs1014335096 Chr.5: 65665792-65665792 on Build GRCh38
rs903514040 Chr.5: 65665920-65665920 on Build GRCh38
rs550612017 Chr.5: 65666101-65666101 on Build GRCh38
rs972116611 Chr.5: 65665779-65665779 on Build GRCh38
rs537255072 Chr.5: 65666093-65666093 on Build GRCh38
rs972144416 Chr.5: 65666049-65666049 on Build GRCh38

View Full Product Details