Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 77432158-77432659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR41
Entrez Gene ID
55255
Gene Name
WD repeat domain 41
Gene Aliases
MSTP048
UniGene
Hs.482573
Gene Chromosome Location
Chr.5: 77426652-77620611 on Build GRCh38
Transcripts
NM_018268.3 XM_005248551.4 XM_011543505.2 XM_017009606.1

SNP Detail

SNP ID Chromosome Location
rs776856876 Chr.5: 77432366-77432366 on Build GRCh38
rs995977998 Chr.5: 77432456-77432456 on Build GRCh38
rs922538 Chr.5: 77432186-77432186 on Build GRCh38
rs952323629 Chr.5: 77432397-77432397 on Build GRCh38
rs145240626 Chr.5: 77432604-77432604 on Build GRCh38
rs3149 Chr.5: 77432611-77432611 on Build GRCh38
rs1006821294 Chr.5: 77432382-77432382 on Build GRCh38
rs1055529133 Chr.5: 77432307-77432307 on Build GRCh38
rs879057581 Chr.5: 77432391-77432391 on Build GRCh38
rs979516706 Chr.5: 77432288-77432288 on Build GRCh38
rs772687883 Chr.5: 77432229-77432229 on Build GRCh38
rs949380646 Chr.5: 77432628-77432628 on Build GRCh38
rs1009196259 Chr.5: 77432466-77432467 on Build GRCh38
rs335632 Chr.5: 77432260-77432260 on Build GRCh38
rs886580405 Chr.5: 77432379-77432379 on Build GRCh38
rs954405476 Chr.5: 77432510-77432510 on Build GRCh38
rs964387635 Chr.5: 77432227-77432227 on Build GRCh38
rs372451527 Chr.5: 77432620-77432620 on Build GRCh38
rs76830603 Chr.5: 77432561-77432561 on Build GRCh38
rs917962934 Chr.5: 77432331-77432331 on Build GRCh38
rs1001940065 Chr.5: 77432213-77432213 on Build GRCh38
rs888050939 Chr.5: 77432583-77432583 on Build GRCh38
rs1043977262 Chr.5: 77432472-77432472 on Build GRCh38
rs116513566 Chr.5: 77432407-77432407 on Build GRCh38
rs759297570 Chr.5: 77432537-77432538 on Build GRCh38
rs913852138 Chr.5: 77432460-77432460 on Build GRCh38
rs867846715 Chr.5: 77432470-77432470 on Build GRCh38
rs771206165 Chr.5: 77432303-77432303 on Build GRCh38
rs965178007 Chr.5: 77432421-77432421 on Build GRCh38
rs927184186 Chr.5: 77432607-77432607 on Build GRCh38
rs917910746 Chr.5: 77432624-77432624 on Build GRCh38
rs140094139 Chr.5: 77432610-77432610 on Build GRCh38
rs956098608 Chr.5: 77432615-77432615 on Build GRCh38
rs865902544 Chr.5: 77432414-77432414 on Build GRCh38
rs937992062 Chr.5: 77432297-77432297 on Build GRCh38
rs139266315 Chr.5: 77432422-77432422 on Build GRCh38
rs757318404 Chr.5: 77432534-77432534 on Build GRCh38
rs968534327 Chr.5: 77432287-77432287 on Build GRCh38
rs937771441 Chr.5: 77432478-77432478 on Build GRCh38
rs925360972 Chr.5: 77432290-77432290 on Build GRCh38
rs1017752970 Chr.5: 77432413-77432413 on Build GRCh38
rs868506420 Chr.5: 77432521-77432521 on Build GRCh38
rs1018204708 Chr.5: 77432599-77432599 on Build GRCh38
rs896239531 Chr.5: 77432550-77432550 on Build GRCh38
rs988710737 Chr.5: 77432277-77432277 on Build GRCh38
rs949598028 Chr.5: 77432365-77432365 on Build GRCh38
rs1020176838 Chr.5: 77432586-77432586 on Build GRCh38
rs934610709 Chr.5: 77432430-77432430 on Build GRCh38
rs1030030940 Chr.5: 77432257-77432257 on Build GRCh38
rs335633 Chr.5: 77432214-77432214 on Build GRCh38
rs747337689 Chr.5: 77432279-77432279 on Build GRCh38
rs904969773 Chr.5: 77432210-77432210 on Build GRCh38
rs1026565386 Chr.5: 77432386-77432386 on Build GRCh38
rs993158740 Chr.5: 77432554-77432554 on Build GRCh38
rs1056105325 Chr.5: 77432485-77432485 on Build GRCh38
rs923276704 Chr.5: 77432428-77432428 on Build GRCh38
rs150001144 Chr.5: 77432589-77432589 on Build GRCh38

View Full Product Details