Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80407878-80408416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFYVE16
Entrez Gene ID
9765
Gene Name
zinc finger FYVE-type containing 16
Gene Aliases
PPP1R69
UniGene
Hs.482660
Gene Chromosome Location
Chr.5: 80407615-80479869 on Build GRCh38
Transcripts
NM_001105251.2 NM_001284236.1 NM_001284237.1 NM_014733.4 XM_005248632.4 XM_011543753.2 XM_017010091.1 XM_017010092.1 XM_017010095.1

SNP Detail

SNP ID Chromosome Location
rs368821911 Chr.5: 80408312-80408312 on Build GRCh38
rs948052727 Chr.5: 80408160-80408161 on Build GRCh38
rs1031552869 Chr.5: 80408101-80408101 on Build GRCh38
rs919061965 Chr.5: 80408198-80408198 on Build GRCh38
rs541541965 Chr.5: 80408236-80408236 on Build GRCh38
rs969574948 Chr.5: 80408353-80408353 on Build GRCh38
rs760222990 Chr.5: 80408273-80408273 on Build GRCh38
rs143477949 Chr.5: 80408212-80408212 on Build GRCh38
rs762459982 Chr.5: 80408181-80408181 on Build GRCh38
rs1006876130 Chr.5: 80408068-80408068 on Build GRCh38
rs924192467 Chr.5: 80407958-80407958 on Build GRCh38
rs1014833596 Chr.5: 80407910-80407910 on Build GRCh38
rs914850043 Chr.5: 80407995-80407995 on Build GRCh38
rs540375177 Chr.5: 80408053-80408053 on Build GRCh38
rs578004099 Chr.5: 80408112-80408112 on Build GRCh38
rs553789783 Chr.5: 80408054-80408054 on Build GRCh38
rs1027368555 Chr.5: 80407936-80407936 on Build GRCh38
rs376174390 Chr.5: 80408001-80408001 on Build GRCh38
rs772677342 Chr.5: 80408205-80408205 on Build GRCh38
rs759058339 Chr.5: 80408365-80408365 on Build GRCh38
rs916821903 Chr.5: 80408349-80408349 on Build GRCh38
rs538373353 Chr.5: 80408012-80408012 on Build GRCh38
rs916604029 Chr.5: 80408157-80408157 on Build GRCh38
rs956895502 Chr.5: 80408114-80408114 on Build GRCh38
rs1022810416 Chr.5: 80407932-80407932 on Build GRCh38
rs951904016 Chr.5: 80408269-80408269 on Build GRCh38
rs985486418 Chr.5: 80407939-80407939 on Build GRCh38
rs983752689 Chr.5: 80407997-80407997 on Build GRCh38
rs569288491 Chr.5: 80408377-80408377 on Build GRCh38
rs979861602 Chr.5: 80408374-80408374 on Build GRCh38
rs995123727 Chr.5: 80408052-80408052 on Build GRCh38
rs948167695 Chr.5: 80408034-80408034 on Build GRCh38
rs552886913 Chr.5: 80408373-80408373 on Build GRCh38
rs372527774 Chr.5: 80408375-80408375 on Build GRCh38
rs538666705 Chr.5: 80407973-80407973 on Build GRCh38
rs992356027 Chr.5: 80408144-80408144 on Build GRCh38
rs1008791031 Chr.5: 80408090-80408090 on Build GRCh38
rs990113341 Chr.5: 80408121-80408121 on Build GRCh38
rs951794154 Chr.5: 80408107-80408107 on Build GRCh38
rs891735875 Chr.5: 80408089-80408089 on Build GRCh38
rs1004604792 Chr.5: 80408270-80408270 on Build GRCh38
rs930805539 Chr.5: 80407991-80407991 on Build GRCh38
rs1018327969 Chr.5: 80407949-80407949 on Build GRCh38
rs186116302 Chr.5: 80408098-80408098 on Build GRCh38
rs939051685 Chr.5: 80408171-80408171 on Build GRCh38
rs998182680 Chr.5: 80408311-80408311 on Build GRCh38
rs371143183 Chr.5: 80408391-80408391 on Build GRCh38
rs888417885 Chr.5: 80408055-80408055 on Build GRCh38
rs969028113 Chr.5: 80408159-80408159 on Build GRCh38
rs530281839 Chr.5: 80407920-80407920 on Build GRCh38
rs960379431 Chr.5: 80408396-80408396 on Build GRCh38
rs749402528 Chr.5: 80408010-80408010 on Build GRCh38
rs976911910 Chr.5: 80407956-80407956 on Build GRCh38
rs567108760 Chr.5: 80407954-80407954 on Build GRCh38
rs1006843338 Chr.5: 80408288-80408288 on Build GRCh38
rs559036461 Chr.5: 80407984-80407984 on Build GRCh38
rs993780574 Chr.5: 80408210-80408210 on Build GRCh38
rs753536412 Chr.5: 80408341-80408341 on Build GRCh38
rs774089128 Chr.5: 80408143-80408143 on Build GRCh38
rs964358316 Chr.5: 80408097-80408097 on Build GRCh38
rs375440579 Chr.5: 80408072-80408072 on Build GRCh38
rs935616039 Chr.5: 80407982-80407982 on Build GRCh38
rs897667789 Chr.5: 80408051-80408051 on Build GRCh38
rs530236236 Chr.5: 80408305-80408305 on Build GRCh38
rs572105750 Chr.5: 80408306-80408306 on Build GRCh38
rs1018295103 Chr.5: 80408091-80408091 on Build GRCh38
rs889063135 Chr.5: 80408266-80408266 on Build GRCh38
rs189628094 Chr.5: 80407937-80407937 on Build GRCh38
rs561795954 Chr.5: 80408294-80408294 on Build GRCh38
rs3805447 Chr.5: 80408022-80408022 on Build GRCh38
rs1011101631 Chr.5: 80408348-80408348 on Build GRCh38
rs780335285 Chr.5: 80407909-80407909 on Build GRCh38
rs994176076 Chr.5: 80407934-80407934 on Build GRCh38
rs770517556 Chr.5: 80408044-80408044 on Build GRCh38
rs965472279 Chr.5: 80408096-80408096 on Build GRCh38
rs891623448 Chr.5: 80408196-80408196 on Build GRCh38
rs577107638 Chr.5: 80408082-80408082 on Build GRCh38
rs574565941 Chr.5: 80408033-80408033 on Build GRCh38
rs531192351 Chr.5: 80408185-80408185 on Build GRCh38
rs532881112 Chr.5: 80408354-80408354 on Build GRCh38
rs939166239 Chr.5: 80408039-80408039 on Build GRCh38
rs897777781 Chr.5: 80407927-80407927 on Build GRCh38
rs974478650 Chr.5: 80407974-80407974 on Build GRCh38
rs538211980 Chr.5: 80408380-80408380 on Build GRCh38
rs927692479 Chr.5: 80408167-80408167 on Build GRCh38
rs969664329 Chr.5: 80408366-80408366 on Build GRCh38
rs906340298 Chr.5: 80407905-80407905 on Build GRCh38
rs1052919253 Chr.5: 80408067-80408067 on Build GRCh38
rs566945454 Chr.5: 80408313-80408313 on Build GRCh38
rs895121938 Chr.5: 80408023-80408023 on Build GRCh38
rs370547347 Chr.5: 80408164-80408164 on Build GRCh38
rs774810208 Chr.5: 80408136-80408136 on Build GRCh38

View Full Product Details